Thermo Scientific™

MAbPac™ HIC-20 HPLC Columns

Catalog number: 02005-259570
Thermo Scientific™

MAbPac™ HIC-20 HPLC Columns

Catalog number: 02005-259570

Use the Thermo Scientific™ MAbPac™ HIC-20 HPLC column for separation of monoclonal antibodies (mAbs), mAb fragments and oxidized variants by hydrophobic interaction chromatography (HIC). The unique chemistry of this silica column offers high resolution and excellent selectivity. The advanced surface bonding technology provides excellent chemical stability, high resolution, and unique selectivity optimized for mAbs and different from other HIC phases.

 
Catalog Number
02005-259570
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
50 mm
Length (Metric)
250 mm
Price (USD)
Full specifications
Column FormatPreparative Column, Standard
Diameter (Metric)50 mm
Length (Metric)250 mm
Max. Pressure8000 psi (550 bar)
Particle Size5 μm
pH2 to 9
Pore Size1000 Å
Temperature60°C
Stationary PhaseProprietary Alkyl Amide
Product LineMAbPac
Packing MaterialSpherical, High Purity Silica
Column TypeSeparation of Monoclonal Antibodies
PhaseHydrophobic Interaction
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
4.6 mm
Length (Metric)
100 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Length (Metric)100 mm
Max. Pressure6000 psi (415 bar)
Particle Size5 μm
pH2 to 9
Pore Size1000 Å
Temperature60°C
Stationary PhaseSilica
Product LineMAbPac
Packing MaterialSpherical, High Purity Silica
Column TypeHydrophobic Interaction
PhaseHydrophobic Interaction
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
4.6 mm
Length (Metric)
250 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Length (Metric)250 mm
Max. Pressure8000 psi (550 bar)
Particle Size5 μm
pH2 to 9
Pore Size1000 Å
Temperature60°C
Stationary PhaseSilica
Product LineMAbPac
Packing MaterialSpherical, High Purity Silica
Column TypeHydrophobic Interaction
PhaseHydrophobic Interaction
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
4.6 mm
Length (Metric)
10 mm
Price (USD)
Full specifications
Column FormatGuard Column
Diameter (Metric)4.6 mm
Length (Metric)10 mm
Max. Pressure6000 psi (415 bar)
Particle Size5 μm
pH2 to 9
Pore Size1000 Å
Temperature60°C
Stationary PhaseSilica
Product LineMAbPac
Packing MaterialSpherical, High Purity Silica
Column TypeHydrophobic Interaction
PhaseHydrophobic Interaction
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
10 mm
Length (Metric)
150 mm
Price (USD)
Full specifications
Column FormatPreparative Column, Standard
Diameter (Metric)10 mm
Length (Metric)150 mm
Max. Pressure8000 psi (550 bar)
Particle Size5 μm
pH2 to 9
Pore Size1000 Å
Temperature60°C
Stationary PhaseAlkyl Amide
Product LineMAbPac
Packing MaterialSpherical, High Purity Silica
Column TypeHydrophobic Interaction
PhaseHydrophobic Interaction
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
10 mm
Length (Metric)
250 mm
Price (USD)
Full specifications
Column FormatPreparative Column, Standard
Diameter (Metric)10 mm
Length (Metric)250 mm
Max. Pressure8000 psi (550 bar)
Particle Size5 μm
pH2 to 9
Pore Size1000 Å
Temperature60°C
Stationary PhaseAlkyl Amide
Product LineMAbPac
Packing MaterialSpherical, High Purity Silica
Column TypeHydrophobic Interaction
PhaseHydrophobic Interaction
Unit SizeEach
Showing 6 of 6
Catalog NumberSpecificationsUnit SizeParticle SizeDiameter (Metric)Length (Metric)Price (USD)
02005-259570Full specifications
Each5 μm50 mm250 mmRequest A Quote
Column FormatPreparative Column, Standard
Diameter (Metric)50 mm
Length (Metric)250 mm
Max. Pressure8000 psi (550 bar)
Particle Size5 μm
pH2 to 9
Pore Size1000 Å
Temperature60°C
Stationary PhaseProprietary Alkyl Amide
Product LineMAbPac
Packing MaterialSpherical, High Purity Silica
Column TypeSeparation of Monoclonal Antibodies
PhaseHydrophobic Interaction
Unit SizeEach
088553Full specifications
Each5 μm4.6 mm100 mmContact Us ›
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Length (Metric)100 mm
Max. Pressure6000 psi (415 bar)
Particle Size5 μm
pH2 to 9
Pore Size1000 Å
Temperature60°C
Stationary PhaseSilica
Product LineMAbPac
Packing MaterialSpherical, High Purity Silica
Column TypeHydrophobic Interaction
PhaseHydrophobic Interaction
Unit SizeEach
088554Full specifications
Each5 μm4.6 mm250 mmContact Us ›
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Length (Metric)250 mm
Max. Pressure8000 psi (550 bar)
Particle Size5 μm
pH2 to 9
Pore Size1000 Å
Temperature60°C
Stationary PhaseSilica
Product LineMAbPac
Packing MaterialSpherical, High Purity Silica
Column TypeHydrophobic Interaction
PhaseHydrophobic Interaction
Unit SizeEach
088555Full specifications
Each5 μm4.6 mm10 mmContact Us ›
Column FormatGuard Column
Diameter (Metric)4.6 mm
Length (Metric)10 mm
Max. Pressure6000 psi (415 bar)
Particle Size5 μm
pH2 to 9
Pore Size1000 Å
Temperature60°C
Stationary PhaseSilica
Product LineMAbPac
Packing MaterialSpherical, High Purity Silica
Column TypeHydrophobic Interaction
PhaseHydrophobic Interaction
Unit SizeEach
SP6961Full specifications
Each5 μm10 mm150 mmRequest A Quote
Column FormatPreparative Column, Standard
Diameter (Metric)10 mm
Length (Metric)150 mm
Max. Pressure8000 psi (550 bar)
Particle Size5 μm
pH2 to 9
Pore Size1000 Å
Temperature60°C
Stationary PhaseAlkyl Amide
Product LineMAbPac
Packing MaterialSpherical, High Purity Silica
Column TypeHydrophobic Interaction
PhaseHydrophobic Interaction
Unit SizeEach
SP6995Full specifications
Each5 μm10 mm250 mmRequest A Quote
Column FormatPreparative Column, Standard
Diameter (Metric)10 mm
Length (Metric)250 mm
Max. Pressure8000 psi (550 bar)
Particle Size5 μm
pH2 to 9
Pore Size1000 Å
Temperature60°C
Stationary PhaseAlkyl Amide
Product LineMAbPac
Packing MaterialSpherical, High Purity Silica
Column TypeHydrophobic Interaction
PhaseHydrophobic Interaction
Unit SizeEach
Showing 6 of 6
Characteristics of the MAbPac HIC-20 Column

This column is based on wide-pore silica particles with proprietary alkyl amide groups offering:

  • Optimal selectivity for mAbs and related variants
  • Excellent separation of mAb fragments and oxidized variants
  • High resolution and high efficiency
  • Compatible with both organic solvent and aqueous mobile phase
  • Rugged column stability
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.