| 063710 | Full specifications | Each | 10 μm | 9 mm | 50 mm | Loading Column | Request A Quote |
Column Format | Loading Column | Diameter (Metric) | 9 mm | Flow Rate | 0.1 to 10 mL/min | For Use With | ProPac IMAC-10 Column | Length (Metric) | 50 mm | Max. Pressure | 3000 psi (207 bar) | Particle Size | 10 μm | pH | 2 to 12 | Pore Size | Nonporous | Temperature | 60°C | Stationary Phase | Lysozyne Poly(IDA) Grafts | Product Line | ProPac | Packing Material | Spherical, Nonporous Polymer | Column Type | Separation of Metal-Binding Proteins | Phase | Affinity Separation | Type | Loading Column | Unit Size | Each |
|
| 063272 | Full specifications | Each | 10 μm | 2 mm | 50 mm | Analytical Column | Contact Us › |
Column Format | Analytical Column | Diameter (Metric) | 2 mm | Flow Rate | 1 mL/min | For Use With | UltiMate 3000 HPLC System | Length (Metric) | 50 mm | Max. Pressure | 3000 psi (207 bar) | Mobile Phase Compatibility | pH 2 to 12, compatible with most reagents used in IMAC protein purifications, including EtOH, Urea, NaCl, non-ionic detergents, glycerol, acetic acid, guanidine, HCl | Particle Size | 10 μm | pH | 2 to 12 | Pore Size | Nonporous | Temperature | 60°C | Stationary Phase | Lysozyne Poly(IDA) Grafts | Product Line | ProPac | Packing Material | Spherical, Nonporous Polymer | Column Type | Separation of Metal-Binding Proteins | Phase | Affinity Separation | Type | Analytical Column | Unit Size | Each |
|
| 063274 | Full specifications | Each | 10 μm | 2 mm | 250 mm | Analytical Column | Request A Quote |
Column Format | Analytical Column | Diameter (Metric) | 2 mm | Flow Rate | 1 mL/min | For Use With | UltiMate 3000 HPLC System | Length (Metric) | 250 mm | Max. Pressure | 3000 psi (207 bar) | Mobile Phase Compatibility | pH 2 to 12, compatible with most reagents used in IMAC protein purifications, including EtOH, Urea, NaCl, non-ionic detergents, glycerol, acetic acid, guanidine, HCl | Particle Size | 10 μm | pH | 2 to 12 | Pore Size | Nonporous | Temperature | 60°C | Stationary Phase | Lysozyne Poly(IDA) Grafts | Product Line | ProPac | Packing Material | Spherical, Nonporous Polymer | Column Type | Separation of Metal-Binding Proteins | Phase | Affinity Separation | Type | Analytical Column | Unit Size | Each |
|
| 063276 | Full specifications | Each | 10 μm | 4 mm | 50 mm | Analytical Column | Contact Us › |
Column Format | Analytical Column | Diameter (Metric) | 4 mm | Flow Rate | 1 mL/min | For Use With | UltiMate 3000 HPLC System | Length (Metric) | 50 mm | Max. Pressure | 3000 psi (207 bar) | Mobile Phase Compatibility | pH 2 to 12, compatible with most reagents used in IMAC protein purifications, including EtOH, Urea, NaCl, non-ionic detergents, glycerol, acetic acid, guanidine, HCl | Particle Size | 10 μm | pH | 2 to 12 | Pore Size | Nonporous | Temperature | 60°C | Stationary Phase | Lysozyne Poly(IDA) Grafts | Product Line | ProPac | Packing Material | Spherical, Nonporous Polymer | Column Type | Separation of Metal-Binding Proteins | Phase | Affinity Separation | Type | Analytical Column | Unit Size | Each |
|
| 063278 | Full specifications | Each | 10 μm | 4 mm | 250 mm | Analytical Column | Contact Us › |
Column Format | Analytical Column | Diameter (Metric) | 4 mm | Flow Rate | 1 mL/min | For Use With | UltiMate 3000 HPLC System | Length (Metric) | 250 mm | Max. Pressure | 3000 psi (207 bar) | Mobile Phase Compatibility | pH 2 to 12, compatible with most reagents used in IMAC protein purifications, including EtOH, Urea, NaCl, non-ionic detergents, glycerol, acetic acid, guanidine, HCl | Particle Size | 10 μm | pH | 2 to 12 | Pore Size | Nonporous | Temperature | 60°C | Stationary Phase | Lysozyne Poly(IDA) Grafts | Product Line | ProPac | Packing Material | Spherical, Nonporous Polymer | Column Type | Separation of Metal-Binding Proteins | Phase | Affinity Separation | Type | Analytical Column | Unit Size | Each |
|
| 063280 | Full specifications | Each | 10 μm | 9 mm | 250 mm | Analytical Column | Request A Quote |
Column Format | Analytical Column | Diameter (Metric) | 9 mm | Flow Rate | 1 mL/min | For Use With | UltiMate 3000 HPLC System | Length (Metric) | 250 mm | Max. Pressure | 3000 psi (207 bar) | Mobile Phase Compatibility | pH 2 to 12, compatible with most reagents used in IMAC protein purifications, including EtOH, Urea, NaCl, non-ionic detergents, glycerol, acetic acid, guanidine, HCl | Particle Size | 10 μm | pH | 2 to 12 | Pore Size | Nonporous | Temperature | 60°C | Stationary Phase | Lysozyne Poly(IDA) Grafts | Product Line | ProPac | Packing Material | Spherical, Nonporous Polymer | Column Type | Separation of Metal-Binding Proteins | Phase | Affinity Separation | Type | Analytical Column | Unit Size | Each |
|
| 063282 | Full specifications | - | 10 μm | 22 mm | 250 mm | Loading Column | Request A Quote |
Column Format | Loading Column | Diameter (Metric) | 22 mm | Flow Rate | 0.1 to 10 mL/min | For Use With | UltiMate 3000 HPLC System | Length (Metric) | 250 mm | Max. Pressure | 3000 psi (207 bar) | Mobile Phase Compatibility | pH 2 to 12, compatible with most reagents used in IMAC protein purifications, including EtOH, Urea, NaCl, non-ionic detergents, glycerol, acetic acid, guanidine, HCl | Particle Size | 10 μm | pH | 2 to 12 | Pore Size | Nonporous | Temperature | 60°C | Stationary Phase | Lysozyne Poly(IDA) Grafts | Packing Material | Spherical, Nonporous Polymer | Column Type | Separation of Metal-Binding Proteins | Phase | Affinity Separation | Type | Analytical Column | Unit Size | - |
|
| 063615 | Full specifications | Each | 10 μm | 9 mm | 50 mm | Analytical Column | Request A Quote |
Column Format | Analytical Column | Diameter (Metric) | 9 mm | Flow Rate | 1 mL/min | For Use With | UltiMate 3000 HPLC System | Length (Metric) | 50 mm | Max. Pressure | 3000 psi (207 bar) | Mobile Phase Compatibility | pH 2 to 12, compatible with most reagents used in IMAC protein purifications, including EtOH, Urea, NaCl, non-ionic detergents, glycerol, acetic acid, guanidine, HCl | Particle Size | 10 μm | pH | 2 to 12 | Pore Size | Nonporous | Temperature | 60°C | Stationary Phase | Lysozyne Poly(IDA) Grafts | Product Line | ProPac | Packing Material | Spherical, Nonporous Polymer | Column Type | Separation of Metal-Binding Proteins | Phase | Affinity Separation | Type | Analytical Column | Unit Size | Each |
|
| 063617 | Full specifications | Each | 10 μm | 1 mm | 50 mm | Analytical Column | Request A Quote |
Column Format | Analytical Column | Diameter (Metric) | 1 mm | Flow Rate | 1 mL/min | For Use With | UltiMate 3000 HPLC System | Length (Metric) | 50 mm | Max. Pressure | 3000 psi (207 bar) | Mobile Phase Compatibility | pH 2 to 12, compatible with most reagents used in IMAC protein purifications, including EtOH, Urea, NaCl, non-ionic detergents, glycerol, acetic acid, guanidine, HCl | Particle Size | 10 μm | pH | 2 to 12 | Pore Size | Nonporous | Temperature | 60°C | Stationary Phase | Lysozyne Poly(IDA) Grafts | Product Line | ProPac | Packing Material | Spherical, Nonporous Polymer | Column Type | Separation of Metal-Binding Proteins | Phase | Affinity Separation | Type | Analytical Column | Unit Size | Each |
|
| 063667 | Full specifications | Each | 10 μm | 4 mm | 50 mm | Loading Column | Contact Us › |
Column Format | Loading Column | Diameter (Metric) | 4 mm | Flow Rate | 0.1 to 10 mL/min | For Use With | ProPac IMAC-10 Column | Length (Metric) | 50 mm | Max. Pressure | 3000 psi (207 bar) | Particle Size | 10 μm | pH | 2 to 12 | Pore Size | Nonporous | Temperature | 60°C | Stationary Phase | Lysozyne Poly(IDA) Grafts | Product Line | ProPac | Packing Material | Spherical, Nonporous Polymer | Column Type | Separation of Metal-Binding Proteins | Phase | Affinity Separation | Type | Loading Column | Unit Size | Each |
|