Thermo Scientific™
Dionex™ ICS-2100 Integrated IC System with Electrolytic Eluent Generation and Sample Preparation
Catalog number: 069573
Thermo Scientific™
Dionex™ ICS-2100 Integrated IC System with Electrolytic Eluent Generation and Sample Preparation
Catalog number: 069573
Run isocratic and gradient IC separations with the Thermo Scientific™ Dionex™ ICS-2100 Integrated IC System with Electrolytic Eluent Generation and Sample Preparation, which combines eluent generation (EG), electrolytic sample preparation (ESP) and conductivity detection. Eliminate the labor of eluent preparation EG and enjoy superior IC performance and ease of use with the convenience, sensitivity and precision EG provides. Remove contaminants prior to analysis by selecting from a variety of electrolytic sample preparation techniques.
Catalog Number
069573
Unit Size
-
Description
ICS-2100 Ion Chromatography System without Degas
Price (USD)
Full specifications
Includeswithout degasser; includes gradient eluent generation, injection valve, column heater, and conductivity detector. (no software, no PC)
Max. PressurePump 5000; ER 3000 psi
TypeWithout Degas
Detector TypeCD, optional UV-Vis
Flow Rate0.00 to 5.00 mL/min.
Gradient ProfilesOption; Combination of unlimited number of linear, convex and concave positive and negative gradient profiles
DescriptionICS-2100 Ion Chromatography System without Degas
Unit Size-
Catalog Number
Unit Size
-
Description
ICS-2100 Ion Chromatography System with Degas
Price (USD)
Full specifications
Includeswith degasser, gradient eluent generation, injection valve, column heater, and conductivity detector. (No software or PC)
Max. PressurePump 5000; ER 3000 psi
TypeWith Degas
Detector TypeCD, optional UV-Vis
Flow Rate0.00 to 5.00 mL/min.
Gradient ProfilesOption; Combination of unlimited number of linear, convex and concave positive and negative gradient profiles
DescriptionICS-2100 Ion Chromatography System with Degas
Unit Size-
Catalog Number
Unit Size
-
Description
ICS-2100 Ion Chromatography System without Degas, Full EG Control, Chromeleon 7 Pk. (no PC)
Price (USD)
Full specifications
IncludesWithout degasser; includes gradient eluent generation, injection valve, column heater, and conductivity detector. (Also includes Chromeleon 7 software, no PC)
Max. PressurePump 5000; ER 3000 psi
TypeWithout Degas, Chromeleon 7 Package (no PC)
Detector TypeCD, optional UV-Vis
Flow Rate0.00 to 5.00 mL/min.
Gradient ProfilesOption; Combination of unlimited number of linear, convex and concave positive and negative gradient profiles
DescriptionICS-2100 Ion Chromatography System without Degas, Full EG Control, Chromeleon 7 Pk. (no PC)
Unit Size-
Catalog Number
Unit Size
-
Description
ICS-2100 Ion Chromatography System with Degas, Full EG Control, Chromeleon 7 Pk. (no PC)
Price (USD)
Full specifications
Includesincludes eluent water degasser, gradient eluent generation, injection valve, column heater, and conductivity detector. (Includes Chromeleon software, no PC)
Max. PressurePump 5000; ER 3000 psi
TypeWith Degas, Chromeleon 7 Package (no PC)
Detector TypeCD, optional UV-Vis
Flow Rate0.00 to 5.00 mL/min.
Gradient ProfilesOption; Combination of unlimited number of linear, convex and concave positive and negative gradient profiles
DescriptionICS-2100 Ion Chromatography System with Degas, Full EG Control, Chromeleon 7 Pk. (no PC)
Unit Size-
Catalog Number
Unit Size
-
Description
ICS-2100 Ion Chromatography System without Degas, Full EG Control, Chromeleon 7, Windows 7 Workstation
Price (USD)
Full specifications
Includeswithout eluent water degasser; includes gradient eluent generation, injection valve, column heater, and conductivity detector. (Also includes Windows 7 computer and Chromeleon software)
Max. PressurePump 5000; ER 3000 psi
TypeWithout Degas, Chromeleon 7, Windows 7 Workstation
Detector TypeCD, optional UV-Vis
Flow Rate0.00 to 5.00 mL/min.
Gradient ProfilesOption; Combination of unlimited number of linear, convex and concave positive and negative gradient profiles
DescriptionICS-2100 Ion Chromatography System without Degas, Full EG Control, Chromeleon 7, Windows 7 Workstation
Unit Size-
Catalog Number
Unit Size
-
Description
ICS-2100 Ion Chromatography System with Degas, Full EG Control, Chromeleon 7, Windows 7 Workstation
Price (USD)
Full specifications
Max. PressurePump 5000; ER 3000 psi
TypeWith Degas, Windows 7 Station
Detector TypeCD, optional UV-Vis
Flow Rate0.00 to 5.00 mL/min.
Gradient ProfilesOption; Combination of unlimited number of linear, convex and concave positive and negative gradient profiles
DescriptionICS-2100 Ion Chromatography System with Degas, Full EG Control, Chromeleon 7, Windows 7 Workstation
Unit Size-
Showing 6 of 6
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
069573Full specifications
-ICS-2100 Ion Chromatography System without DegasRequest A Quote
Includeswithout degasser; includes gradient eluent generation, injection valve, column heater, and conductivity detector. (no software, no PC)
Max. PressurePump 5000; ER 3000 psi
TypeWithout Degas
Detector TypeCD, optional UV-Vis
Flow Rate0.00 to 5.00 mL/min.
Gradient ProfilesOption; Combination of unlimited number of linear, convex and concave positive and negative gradient profiles
DescriptionICS-2100 Ion Chromatography System without Degas
Unit Size-
069576Full specifications
-ICS-2100 Ion Chromatography System with DegasRequest A Quote
Includeswith degasser, gradient eluent generation, injection valve, column heater, and conductivity detector. (No software or PC)
Max. PressurePump 5000; ER 3000 psi
TypeWith Degas
Detector TypeCD, optional UV-Vis
Flow Rate0.00 to 5.00 mL/min.
Gradient ProfilesOption; Combination of unlimited number of linear, convex and concave positive and negative gradient profiles
DescriptionICS-2100 Ion Chromatography System with Degas
Unit Size-
069658Full specifications
-ICS-2100 Ion Chromatography System without Degas, Full EG Control, Chromeleon 7 Pk. (no PC)Request A Quote
IncludesWithout degasser; includes gradient eluent generation, injection valve, column heater, and conductivity detector. (Also includes Chromeleon 7 software, no PC)
Max. PressurePump 5000; ER 3000 psi
TypeWithout Degas, Chromeleon 7 Package (no PC)
Detector TypeCD, optional UV-Vis
Flow Rate0.00 to 5.00 mL/min.
Gradient ProfilesOption; Combination of unlimited number of linear, convex and concave positive and negative gradient profiles
DescriptionICS-2100 Ion Chromatography System without Degas, Full EG Control, Chromeleon 7 Pk. (no PC)
Unit Size-
069659Full specifications
-ICS-2100 Ion Chromatography System with Degas, Full EG Control, Chromeleon 7 Pk. (no PC)Request A Quote
Includesincludes eluent water degasser, gradient eluent generation, injection valve, column heater, and conductivity detector. (Includes Chromeleon software, no PC)
Max. PressurePump 5000; ER 3000 psi
TypeWith Degas, Chromeleon 7 Package (no PC)
Detector TypeCD, optional UV-Vis
Flow Rate0.00 to 5.00 mL/min.
Gradient ProfilesOption; Combination of unlimited number of linear, convex and concave positive and negative gradient profiles
DescriptionICS-2100 Ion Chromatography System with Degas, Full EG Control, Chromeleon 7 Pk. (no PC)
Unit Size-
069656Full specifications
-ICS-2100 Ion Chromatography System without Degas, Full EG Control, Chromeleon 7, Windows 7 WorkstationRequest A Quote
Includeswithout eluent water degasser; includes gradient eluent generation, injection valve, column heater, and conductivity detector. (Also includes Windows 7 computer and Chromeleon software)
Max. PressurePump 5000; ER 3000 psi
TypeWithout Degas, Chromeleon 7, Windows 7 Workstation
Detector TypeCD, optional UV-Vis
Flow Rate0.00 to 5.00 mL/min.
Gradient ProfilesOption; Combination of unlimited number of linear, convex and concave positive and negative gradient profiles
DescriptionICS-2100 Ion Chromatography System without Degas, Full EG Control, Chromeleon 7, Windows 7 Workstation
Unit Size-
083251Full specifications
-ICS-2100 Ion Chromatography System with Degas, Full EG Control, Chromeleon 7, Windows 7 WorkstationRequest A Quote
Max. PressurePump 5000; ER 3000 psi
TypeWith Degas, Windows 7 Station
Detector TypeCD, optional UV-Vis
Flow Rate0.00 to 5.00 mL/min.
Gradient ProfilesOption; Combination of unlimited number of linear, convex and concave positive and negative gradient profiles
DescriptionICS-2100 Ion Chromatography System with Degas, Full EG Control, Chromeleon 7, Windows 7 Workstation
Unit Size-
Showing 6 of 6
This totally integrated Thermo Scientific™ Reagent-Free™ IC (RFIC™) system combines the powerful features of the Dionex ICS-1600 system with the added convenience, sensitivity and precision of integrated EG and ESP technologies for an extra edge in detecting ultralow concentration targets. Get full control and digital data collection from a PC using Thermo Scientific™ Dionex™ Chromeleon™ Chromatography Data System (CDS) software.
Outstanding Sensitivity and Precision
  • Prepare eluent electrolytically from deionized water and a year's worth of concentrate (on average)
  • Optimize separations with electrolytic gradients
  • Eliminate errors and costs of manual eluent preparation
  • Program isocratic or gradient eluent concentrations programmed with a mouse click for simplified, error-free workflow
  • Improve consistency across runs, instruments and labs
  • Get extremely low detection limits using very low background conductivity hydroxide eluents
Electrolytic Sample Preparation Options
  • The auxiliary power supply on the Dionex ICS-2100 RFIC-ESP system allows control of electrolytic sample preparation devices. Options and benefits include:
  • Neutralize acidic or basic samples with an anion or cation Thermo Scientific™ Dionex™ SRN™ Self-Regenerating Neutralizer
  • Reduce ion backgrounds in sample transfer water for ultratrace-level determinations using an optional electrolytic RFIC-ESP water purifier
  • Extend instrument applications to include integrated in-line filtration, matrix elimination, neutralization and ultratrace analysis
Automated Sample Preparation Option
  • Eliminate pre-injection sample processing
  • Save time and effort using automated large loop/small loop reinjection of samples
  • Support automated sample preparation using the six- or 10-port valve
Additional Options
  • In-line vacuum degas increases reproducibility and protects eluents from contamination and decomposition
  • Integrated auxiliary valve enables time-saving automated sample preparation techniques