Thermo Scientific™

Dionex™ ICS-5000+ EG Eluent Generator

Catalog number: 072045
Thermo Scientific™

Dionex™ ICS-5000+ EG Eluent Generator

Catalog number: 072045
Catalog Number
072045
Unit Size
Each
Price (USD)
Catalog NumberUnit SizePrice (USD)
072045EachRequest A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Minimize baselines, maximize consistency, and simplify your ion chromatography (IC) workflow with the Thermo Scientific™ Dionex™ ICS-5000+ EG Eluent Generator. Add the EG module to an ICS-5000+ system to create a Thermo Scientific™ Reagent-Free™ IC (RFIC™) system that produces single or dual eluents on-line electrolytically using only DI water and simple cartridges. Robust electronics generate high-purity eluents and exceptionally stable baselines. Just add water to generate the perfect eluent for almost any IC application—anion or cation, capillary or analytic, isocratic or gradient.

Specifications

Electrical Requirements
90 to 265 VAC 47/63 Hz
Frequency
47/63 Hz
Flow Rate
0.001 to 10 mL/min.
For Use With (Equipment)
Dionex ICS-5000+ System
Max Eluent Concentration
100 mM (Analytical); 200 mM (Capillary)
Max. Pressure
21 MPa (3000 psi)
Type
Eluent Generator
Gradient Profiles
Any combination of an unlimited number of linear, convex, and concave positive and negative gradient profiles
Min. Eluent Concentration
0.01 mM
Max. Solvent Concentration
Cations: None; Anions: 25% Methanol
Standard Features
Built-in leak sensor and operator alert; slide-out tray; Automated Procedure Wizards; System Smart Startup and Shutdown; System Wellness and Predictive Performance; Application Templates; Multivendor Automation Support of 3 rd Party
Height (English)
16 in.
Height (Metric)
41 cm
Voltage
90 to 265 VAC
Weight (English)
40 lb.
Weight (Metric)
25 kg
Width (English)
8.75 in.
Width (Metric)
23 cm
Depth (English)
21.5 in.
Depth (Metric)
56 cm
Description
EG Eluent Generator Module