Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Max. Pressure | Pump: 35 MPa (5000 psi); ER: 20 MPa (3000 psi) |
Type | Single Channel RFIC System (MB) |
Detector Type | CD, ED, UV-Vis |
Flow Rate | 0.00 to 10.00 mL/min. |
Pressure Range | Various |
Gradient Profiles | Any combination of an unlimited number of linear, convex, and concave positive and negative |
Description | ICS-5000+ Single Channel Reagent-Free Ion Chromatography Microbore System |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
072242 | Each | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Outstanding Sensitivity Plus Convenience
Unmatched Variety of Applications
Compatible with New 4μm Columns
Outstanding Flexibility to Configure the System You Need
Flexible Choice of Detectors