Thermo Scientific™
Dionex™ ICS-4000 QD Charge Detector and Cell
Catalog number: 078449
Thermo Scientific™
Dionex™ ICS-4000 QD Charge Detector and Cell
Catalog number: 078449
The Thermo Scientific™ Dionex™ ICS-4000 QD Charge Detector and Cell is the world’s first charge detector for ion chromatography. In combination with conductivity detection, it provides improved peak identification and peak purity analysis. The Dionex QD uses a membrane-based technology and detects ions in proportion to their charge and concentration. It produces up to three times greater signal for triply charged ions, e.g., phosphate, than singly charged ions such as chloride. Ions with the same charge and concentration yield similar response, permitting reliable quantification of known and unknown compounds with a single standard. The Dionex QD it is well suited to the analysis of phosphates in environmental samples, organic acids in food and beverages, and amines in chemicals.
Catalog Number
078449
Unit Size
Each
Description
QD-4000 Charge Detector
Price (USD)
Full specifications
DescriptionQD-4000 Charge Detector
For Use With (Application)Ion Chromatography
TypeCharge Detector
Detection ModeMembrane-Based Ion Detection
Detector TypeConductivity
Electronics TypeMicroprocessor controlled digital signal processing, autoranging
Flow Cell Max. Pressure0.7 MPa (100 psi)
Linearity1%
Output RangeDigital Signal Range: 0 to 20 µA, Analog Signal Range: 0 to 20 µA
Unit SizeEach
Catalog Number
Unit Size
Each 1
Description
QDC 300 Charge Detector Cell (Capillary)
Price (USD)
Full specifications
DescriptionQDC 300 Charge Detector Cell (Capillary)
For Use With (Application)Ion Chromatography
TypeCharge Detector Cell (Capillary)
Detection ModeMembrane-Based Ion Detection
Detector TypeConductivity
Unit SizeEach 1
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
078449Full specifications
EachQD-4000 Charge DetectorRequest A Quote
DescriptionQD-4000 Charge Detector
For Use With (Application)Ion Chromatography
TypeCharge Detector
Detection ModeMembrane-Based Ion Detection
Detector TypeConductivity
Electronics TypeMicroprocessor controlled digital signal processing, autoranging
Flow Cell Max. Pressure0.7 MPa (100 psi)
Linearity1%
Output RangeDigital Signal Range: 0 to 20 µA, Analog Signal Range: 0 to 20 µA
Unit SizeEach
078814Full specifications
Each 1QDC 300 Charge Detector Cell (Capillary)Contact Us ›
DescriptionQDC 300 Charge Detector Cell (Capillary)
For Use With (Application)Ion Chromatography
TypeCharge Detector Cell (Capillary)
Detection ModeMembrane-Based Ion Detection
Detector TypeConductivity
Unit SizeEach 1
Showing 2 of 2
  • Detects all ionizable species
  • Stronger relative response for very weakly dissociated and multivalent ions
  • Universal response simplifies calibration
  • Quantification of unknowns possible
  • Linear Response for weakly dissociated ions
  • Peak purity information
  • Low investment cost
  • Long consumables lifetime, low maintenance cost
  • Easy to use
  • Complements conductivity detection
  • Identification of ions in combination with conductivity detection and retention time
  • CD/QD filling a gap between CD and MS
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.