Thermo Scientific™

Accucore™ 150 C18 HPLC Columns

Catalog number: 16126-157569
Thermo Scientific™

Accucore™ 150 C18 HPLC Columns

Catalog number: 16126-157569

Offer phase characteristics designed for the separation of peptides with Thermo Scientific™ Accucore™ 150 C18 LC Columns. Based on Core Enhanced Technology, Accucore columns provide fast, high-resolution biomolecule separations without the elevated backpressures required by sub-2μm particles.

 
Catalog Number
16126-157569
Unit Size
Each
Particle Size
2.6 μm
Diameter (Metric)
0.075 mm
Length (Metric)
150 mm
Price (USD)
Full specifications
Column FormatAnalytical Column, NanoViper
Diameter (Metric)0.075 mm
For Use WithHPLC
Length (Metric)150 mm
Max. Pressure11600 psi (800 bar)
Particle Size2.6 μm
pH1 to 11
Pore Size150 Å
Surface Area80 m²/g
Temperature60°C
USP TypeL1
Stationary PhaseC18
Product LinenanoLC
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Carbon Load7%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
2.6 μm
Diameter (Metric)
2.1 mm
Length (Metric)
30 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
For Use WithHPLC
Length (Metric)30 mm
Max. Pressure14500 psi (1000 bar)
Particle Size2.6 μm
pH1 to 11
Pore Size150 Å
Surface Area80 m²/g
Temperature60°C
USP TypeL1
Stationary PhaseC18
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Carbon Load7%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit SizeEach
Catalog Number
Unit Size
-
Particle Size
2.6 μm
Diameter (Metric)
3 mm
Length (Metric)
30 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)3 mm
For Use WithHPLC
Length (Metric)30 mm
Max. Pressure14500 psi (1000 bar)
Particle Size2.6 μm
pH1 to 11
Pore Size150 Å
Surface Area80 m²/g
Temperature60°C
USP TypeL1
Stationary PhaseC18
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Carbon Load7%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit Size-
Catalog Number
Unit Size
-
Particle Size
2.6 μm
Diameter (Metric)
4.6 mm
Length (Metric)
30 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
For Use WithHPLC
Length (Metric)30 mm
Max. Pressure14500 psi (1000 bar)
Particle Size2.6 μm
pH1 to 11
Pore Size150 Å
Surface Area80 m²/g
Temperature60°C
USP TypeL1
Stationary PhaseC18
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Carbon Load7%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit Size-
Catalog Number
Unit Size
Each
Particle Size
2.6 μm
Diameter (Metric)
2.1 mm
Length (Metric)
50 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
For Use WithHPLC
Length (Metric)50 mm
Max. Pressure14500 psi (1000 bar)
Particle Size2.6 μm
pH1 to 11
Pore Size150 Å
Surface Area80 m²/g
Temperature60°C
USP TypeL1
Stationary PhaseC18
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Carbon Load7%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit SizeEach
Catalog Number
Unit Size
-
Particle Size
2.6 μm
Diameter (Metric)
3 mm
Length (Metric)
50 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)3 mm
For Use WithHPLC
Length (Metric)50 mm
Max. Pressure14500 psi (1000 bar)
Particle Size2.6 μm
pH1 to 11
Pore Size150 Å
Surface Area80 m²/g
Temperature60°C
USP TypeL1
Stationary PhaseC18
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Carbon Load7%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit Size-
Catalog Number
Unit Size
-
Particle Size
2.6 μm
Diameter (Metric)
4.6 mm
Length (Metric)
50 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
For Use WithHPLC
Length (Metric)50 mm
Max. Pressure14500 psi (1000 bar)
Particle Size2.6 μm
pH1 to 11
Pore Size150 Å
Surface Area80 m²/g
Temperature60°C
USP TypeL1
Stationary PhaseC18
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Carbon Load7%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit Size-
Catalog Number
Unit Size
Each
Particle Size
2.6 μm
Diameter (Metric)
2.1 mm
Length (Metric)
100 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
For Use WithHPLC
Length (Metric)100 mm
Max. Pressure14500 psi (1000 bar)
Particle Size2.6 μm
pH1 to 11
Pore Size150 Å
Surface Area80 m²/g
Temperature60°C
USP TypeL1
Stationary PhaseC18
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Carbon Load7%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit SizeEach
Catalog Number
Unit Size
-
Particle Size
2.6 μm
Diameter (Metric)
3 mm
Length (Metric)
100 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)3 mm
For Use WithHPLC
Length (Metric)100 mm
Max. Pressure14500 psi (1000 bar)
Particle Size2.6 μm
pH1 to 11
Pore Size150 Å
Surface Area80 m²/g
Temperature60°C
USP TypeL1
Stationary PhaseC18
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Carbon Load7%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit Size-
Catalog Number
Unit Size
Each
Particle Size
2.6 μm
Diameter (Metric)
4.6 mm
Length (Metric)
100 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
For Use WithHPLC
Length (Metric)100 mm
Max. Pressure14500 psi (1000 bar)
Particle Size2.6 μm
pH1 to 11
Pore Size150 Å
Surface Area80 m²/g
Temperature60°C
USP TypeL1
Stationary PhaseC18
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Carbon Load7%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit SizeEach
Showing 10 of 17Show more
Catalog NumberSpecificationsUnit SizeParticle SizeDiameter (Metric)Length (Metric)Price (USD)
16126-157569Full specifications
Each 2.6 μm0.075 mm150 mmContact Us ›
Column FormatAnalytical Column, NanoViper
Diameter (Metric)0.075 mm
For Use WithHPLC
Length (Metric)150 mm
Max. Pressure11600 psi (800 bar)
Particle Size2.6 μm
pH1 to 11
Pore Size150 Å
Surface Area80 m²/g
Temperature60°C
USP TypeL1
Stationary PhaseC18
Product LinenanoLC
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Carbon Load7%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit SizeEach
16126-032130Full specifications
Each 2.6 μm2.1 mm30 mmContact Us ›
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
For Use WithHPLC
Length (Metric)30 mm
Max. Pressure14500 psi (1000 bar)
Particle Size2.6 μm
pH1 to 11
Pore Size150 Å
Surface Area80 m²/g
Temperature60°C
USP TypeL1
Stationary PhaseC18
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Carbon Load7%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit SizeEach
16126-033030Full specifications
- 2.6 μm3 mm30 mmRequest A Quote
Column FormatAnalytical Column
Diameter (Metric)3 mm
For Use WithHPLC
Length (Metric)30 mm
Max. Pressure14500 psi (1000 bar)
Particle Size2.6 μm
pH1 to 11
Pore Size150 Å
Surface Area80 m²/g
Temperature60°C
USP TypeL1
Stationary PhaseC18
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Carbon Load7%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit Size-
16126-034630Full specifications
-2.6 μm4.6 mm30 mmRequest A Quote
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
For Use WithHPLC
Length (Metric)30 mm
Max. Pressure14500 psi (1000 bar)
Particle Size2.6 μm
pH1 to 11
Pore Size150 Å
Surface Area80 m²/g
Temperature60°C
USP TypeL1
Stationary PhaseC18
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Carbon Load7%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit Size-
16126-052130Full specifications
Each2.6 μm2.1 mm50 mmContact Us ›
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
For Use WithHPLC
Length (Metric)50 mm
Max. Pressure14500 psi (1000 bar)
Particle Size2.6 μm
pH1 to 11
Pore Size150 Å
Surface Area80 m²/g
Temperature60°C
USP TypeL1
Stationary PhaseC18
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Carbon Load7%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit SizeEach
16126-053030Full specifications
- 2.6 μm3 mm50 mmRequest A Quote
Column FormatAnalytical Column
Diameter (Metric)3 mm
For Use WithHPLC
Length (Metric)50 mm
Max. Pressure14500 psi (1000 bar)
Particle Size2.6 μm
pH1 to 11
Pore Size150 Å
Surface Area80 m²/g
Temperature60°C
USP TypeL1
Stationary PhaseC18
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Carbon Load7%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit Size-
16126-054630Full specifications
- 2.6 μm4.6 mm50 mmRequest A Quote
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
For Use WithHPLC
Length (Metric)50 mm
Max. Pressure14500 psi (1000 bar)
Particle Size2.6 μm
pH1 to 11
Pore Size150 Å
Surface Area80 m²/g
Temperature60°C
USP TypeL1
Stationary PhaseC18
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Carbon Load7%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit Size-
16126-102130Full specifications
Each2.6 μm2.1 mm100 mmContact Us ›
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
For Use WithHPLC
Length (Metric)100 mm
Max. Pressure14500 psi (1000 bar)
Particle Size2.6 μm
pH1 to 11
Pore Size150 Å
Surface Area80 m²/g
Temperature60°C
USP TypeL1
Stationary PhaseC18
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Carbon Load7%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit SizeEach
16126-103030Full specifications
-2.6 μm3 mm100 mmRequest A Quote
Column FormatAnalytical Column
Diameter (Metric)3 mm
For Use WithHPLC
Length (Metric)100 mm
Max. Pressure14500 psi (1000 bar)
Particle Size2.6 μm
pH1 to 11
Pore Size150 Å
Surface Area80 m²/g
Temperature60°C
USP TypeL1
Stationary PhaseC18
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Carbon Load7%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit Size-
16126-104630Full specifications
Each 2.6 μm4.6 mm100 mmContact Us ›
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
For Use WithHPLC
Length (Metric)100 mm
Max. Pressure14500 psi (1000 bar)
Particle Size2.6 μm
pH1 to 11
Pore Size150 Å
Surface Area80 m²/g
Temperature60°C
USP TypeL1
Stationary PhaseC18
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Carbon Load7%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit SizeEach
Showing 10 of 17Show more
  • 150Å pore size solid core particles for fast biomoleucule separations
  • Superb resolution at low backpressures
  • Exceptionally rugged columns
  • Designed for the separation of peptides Outstanding resolution

Ordering Information:

Other custom column dimensions are available. Please call your local Customer Service for more information.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.