Thermo Scientific™

PepMap™ 100 C8 HPLC Columns

Catalog number: 161459
Thermo Scientific™

PepMap™ 100 C8 HPLC Columns

Catalog number: 161459
Choose Thermo Scientific™ Acclaim™ PepMap™ 100 C8 LC Columns when separating very hydrophobic peptides (nontryptic peptides) as an alternative to the PepMap™ C18 column.
 
Catalog Number
161459
Unit Size
Each
Particle Size
3 μm
Diameter (Metric)
100 μm
Length (Metric)
150 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)100 μm
Flow Rate40 µL/min
For Use WithLC/MS, ESI-MS, MALDI-MS
Length (Metric)150 mm
Particle Size3 μm
Pore Size100 Å
USP TypeL1
Stationary PhaseC18
Packing MaterialSilica
Column TypePeptide Analysis
PhaseReversed Phase
MaterialStainless Steel
TypeAnalytical Column
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
3 μm
Diameter (Metric)
0.075 mm
Length (Metric)
150 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)0.075 mm
Flow Rate40 µL/min
For Use WithLC/MS, ESI-MS, MALDI-MS
Length (Metric)150 mm
Max. Pressure7250 psi (500 bar)
Particle Size3 μm
pH2 to 8
Pore Size100 Å
Surface Area450 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LinenanoLC
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load9%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
MaterialStainless Steel
TypeAnalytical Column
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
0.3 mm
Length (Metric)
5 mm
Price (USD)
Full specifications
Column FormatTrap Cartridge
Diameter (Metric)0.3 mm
Length (Metric)5 mm
Max. Pressure7250 psi (500 bar)
Particle Size5 μm
pH2 to 8
Pore Size100 Å
Surface Area450 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LinenanoLC
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load9%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
MaterialStainless Steel
TypeTrap Cartridge
Unit SizeEach
Showing 3 of 3
Catalog NumberSpecificationsUnit SizeParticle SizeDiameter (Metric)Length (Metric)Price (USD)
161459Full specifications
Each3 μm100 μm150 mmRequest A Quote
Column FormatAnalytical Column
Diameter (Metric)100 μm
Flow Rate40 µL/min
For Use WithLC/MS, ESI-MS, MALDI-MS
Length (Metric)150 mm
Particle Size3 μm
Pore Size100 Å
USP TypeL1
Stationary PhaseC18
Packing MaterialSilica
Column TypePeptide Analysis
PhaseReversed Phase
MaterialStainless Steel
TypeAnalytical Column
Unit SizeEach
161185Full specifications
Each3 μm0.075 mm150 mmContact Us ›
Column FormatAnalytical Column
Diameter (Metric)0.075 mm
Flow Rate40 µL/min
For Use WithLC/MS, ESI-MS, MALDI-MS
Length (Metric)150 mm
Max. Pressure7250 psi (500 bar)
Particle Size3 μm
pH2 to 8
Pore Size100 Å
Surface Area450 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LinenanoLC
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load9%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
MaterialStainless Steel
TypeAnalytical Column
Unit SizeEach
161194Full specifications
Each5 μm0.3 mm5 mmContact Us ›
Column FormatTrap Cartridge
Diameter (Metric)0.3 mm
Length (Metric)5 mm
Max. Pressure7250 psi (500 bar)
Particle Size5 μm
pH2 to 8
Pore Size100 Å
Surface Area450 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LinenanoLC
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load9%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
MaterialStainless Steel
TypeTrap Cartridge
Unit SizeEach
Showing 3 of 3
This column provides high-resolution, high-sensitivity protein identification for proteomics and biomarker discovery and verification, and is ideal for:

 

High loading capcity

Acclaim PepMap columns are exceptionally suitable for the analysis of low abundant peptides often found in complex proteomics samples.

The PepMap 100 C8 LC Columns is available in 150mm length with a choice of 75, 300, 1000, and classic 75µm I.D. to allow tuning for throughput, sensitivity and amount of sample available.

Superior protein and peptide separations

Trifluoroacetic acid (TFA)-free LC/MS minimizes suppression effects for enhanced MS sensitivity

Outstanding separation efficiency

Superior resolution and recovery

Easy installation with Thermo Scientific™ Dionex™ nanoViper™ Fingertight Fittings

Columns are available with nanoViper Fingertight Fittings

Requires no tools

Provides virtually zero-dead-volume connections

Withstands pressures up to a 1000 bar

About Nano, capillary, and micro Acclaim PepMap columns

Nano (75µm), capillary (300µm) and micro (1000µm) columns are ideal for protein and peptide analyses. Sample amount is often limited and smaller I.D. columns can increase in sensitivity in sample-limited situations. While nano LC columns are often used in proteomics, the concentrating effects of capillary and micro columns are beneficial for metabolomics or for facilitating LC-MS interfacing. Thermo Scientific Acclaim PepMap columns are offered in nano, capillary and micro column in a variety of chemistries for peptides and protein analysis.