Thermo Scientific™

PepSwift™ Monolithic Capillary HPLC Columns

Catalog number: 164585
Thermo Scientific™

PepSwift™ Monolithic Capillary HPLC Columns

Catalog number: 164585
For fast, high-resolution LC/MS separations in bottom-up protein identification, biomarker discovery, and systems biology applications, use Thermo Scientific™ PepSwift™ Monolithic Capillary LC Columns. Based on a polystyrene divinylbenzene copolymer, the monolithic structure offers a high-quality alternative to traditional microparticulate sorbents, providing important advantages to the chromatographic separation. High-sensitivity proteomics and biotech applications are easily performed using these columns.
 
Catalog Number
164585
Unit Size
Each
Particle Size
Monolith
Diameter (Metric)
0.5 mm
Length (Metric)
50 mm
Price (USD)
Full specifications
Column FormatAnalytical Column, NanoViper
Diameter (Metric)0.5 mm
Flow Rate15 to 25 µL/min
Length (Metric)50 mm
Max. Pressure4350 psi (300 bar)
Particle SizeMonolith
pH1 to 10
Pore SizeMonolith
Temperature80°C
Stationary PhasePSDVB (Polystyrene Divinylbenzene)
Product LinenanoLC
Packing MaterialMonolith Polymer
Column TypeReversed Phase
PhaseReversed Phase
MaterialPolystyrene/Divinylbenzene
TypeAnalytical Column, NanoViper
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
Monolith
Diameter (Metric)
0.1 mm
Length (Metric)
50 mm
Price (USD)
Full specifications
Column FormatCapillary Column, NanoViper
Diameter (Metric)0.1 mm
Flow Rate700 to 1000 nL/min
Length (Metric)50 mm
Max. Pressure4350 psi (300 bar)
Particle SizeMonolith
pH1 to 10
Pore SizeMonolith
Temperature80°C
Stationary PhasePSDVB (Polystyrene Divinylbenzene)
Product LinenanoLC
Packing MaterialMonolith Polymer
Column TypeReversed Phase
PhaseReversed Phase
MaterialPolystyrene/Divinylbenzene
TypeAnalytical Column, NanoViper
Unit SizeEach
Catalog Number
Unit Size
Pack of 2
Particle Size
Monolith
Diameter (Metric)
0.2 mm
Length (Metric)
5 mm
Price (USD)
Full specifications
Column FormatTrap Column, NanoViper
Diameter (Metric)0.2 mm
Length (Metric)5 mm
Max. Pressure4350 psi (300 bar)
Particle SizeMonolith
pH1 to 10
Pore SizeMonolith
Temperature80°C
Stationary PhasePSDVB (Polystyrene Divinylbenzene)
Product LinenanoLC
Packing MaterialMonolith Polymer
Column TypeReversed Phase
PhaseReversed Phase
MaterialPolystyrene/Divinylbenzene
TypenanoViper
Unit SizePack of 2
Catalog Number
Unit Size
Each
Particle Size
Monolith
Diameter (Metric)
0.2 mm
Length (Metric)
50 mm
Price (USD)
Full specifications
Column FormatAnalytical Column, NanoViper
Diameter (Metric)0.2 mm
Flow Rate2 to 3 µL/min
Length (Metric)50 mm
Max. Pressure4350 psi (300 bar)
Particle SizeMonolith
pH1 to 10
Pore SizeMonolith
Temperature80°C
Stationary PhasePSDVB (Polystyrene Divinylbenzene)
Product LinenanoLC
Packing MaterialMonolith Polymer
Column TypeReversed Phase
PhaseReversed Phase
MaterialPolystyrene/Divinylbenzene
TypeAnalytical Column, NanoViper
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
Monolith
Diameter (Metric)
0.1 mm
Length (Metric)
250 mm
Price (USD)
Full specifications
Column FormatAnalytical Column, NanoViper
Diameter (Metric)0.1 mm
Length (Metric)250 mm
Max. Pressure4350 psi (300 bar)
Particle SizeMonolith
pH1 to 10
Pore SizeMonolith
Temperature80°C
Stationary PhasePSDVB (Polystyrene Divinylbenzene)
Product LinenanoLC
Packing MaterialMonolith Polymer
Column TypeReversed Phase
PhaseReversed Phase
MaterialPolystyrene/Divinylbenzene
TypeAnalytical Column, NanoViper
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
Monolith
Diameter (Metric)
0.2 mm
Length (Metric)
250 mm
Price (USD)
Full specifications
Column FormatCapillary Column, NanoViper
Diameter (Metric)0.2 mm
Length (Metric)250 mm
Max. Pressure4350 psi (300 bar)
Particle SizeMonolith
pH1 to 10
Pore SizeMonolith
Temperature80°C
Stationary PhasePSDVB (Polystyrene Divinylbenzene)
Product LinenanoLC
Packing MaterialMonolith Polymer
Column TypeReversed Phase
PhaseReversed Phase
MaterialPolystyrene/Divinylbenzene
TypeAnalytical Column, NanoViper
Unit SizeEach
Showing 6 of 6
Catalog NumberSpecificationsUnit SizeParticle SizeDiameter (Metric)Length (Metric)Price (USD)
164585Full specifications
EachMonolith0.5 mm50 mmContact Us ›
Column FormatAnalytical Column, NanoViper
Diameter (Metric)0.5 mm
Flow Rate15 to 25 µL/min
Length (Metric)50 mm
Max. Pressure4350 psi (300 bar)
Particle SizeMonolith
pH1 to 10
Pore SizeMonolith
Temperature80°C
Stationary PhasePSDVB (Polystyrene Divinylbenzene)
Product LinenanoLC
Packing MaterialMonolith Polymer
Column TypeReversed Phase
PhaseReversed Phase
MaterialPolystyrene/Divinylbenzene
TypeAnalytical Column, NanoViper
Unit SizeEach
164584Full specifications
EachMonolith0.1 mm50 mmContact Us ›
Column FormatCapillary Column, NanoViper
Diameter (Metric)0.1 mm
Flow Rate700 to 1000 nL/min
Length (Metric)50 mm
Max. Pressure4350 psi (300 bar)
Particle SizeMonolith
pH1 to 10
Pore SizeMonolith
Temperature80°C
Stationary PhasePSDVB (Polystyrene Divinylbenzene)
Product LinenanoLC
Packing MaterialMonolith Polymer
Column TypeReversed Phase
PhaseReversed Phase
MaterialPolystyrene/Divinylbenzene
TypeAnalytical Column, NanoViper
Unit SizeEach
164558Full specifications
Pack of 2Monolith0.2 mm5 mmContact Us ›
Column FormatTrap Column, NanoViper
Diameter (Metric)0.2 mm
Length (Metric)5 mm
Max. Pressure4350 psi (300 bar)
Particle SizeMonolith
pH1 to 10
Pore SizeMonolith
Temperature80°C
Stationary PhasePSDVB (Polystyrene Divinylbenzene)
Product LinenanoLC
Packing MaterialMonolith Polymer
Column TypeReversed Phase
PhaseReversed Phase
MaterialPolystyrene/Divinylbenzene
TypenanoViper
Unit SizePack of 2
164557Full specifications
EachMonolith0.2 mm50 mmContact Us ›
Column FormatAnalytical Column, NanoViper
Diameter (Metric)0.2 mm
Flow Rate2 to 3 µL/min
Length (Metric)50 mm
Max. Pressure4350 psi (300 bar)
Particle SizeMonolith
pH1 to 10
Pore SizeMonolith
Temperature80°C
Stationary PhasePSDVB (Polystyrene Divinylbenzene)
Product LinenanoLC
Packing MaterialMonolith Polymer
Column TypeReversed Phase
PhaseReversed Phase
MaterialPolystyrene/Divinylbenzene
TypeAnalytical Column, NanoViper
Unit SizeEach
164543Full specifications
EachMonolith0.1 mm250 mmContact Us ›
Column FormatAnalytical Column, NanoViper
Diameter (Metric)0.1 mm
Length (Metric)250 mm
Max. Pressure4350 psi (300 bar)
Particle SizeMonolith
pH1 to 10
Pore SizeMonolith
Temperature80°C
Stationary PhasePSDVB (Polystyrene Divinylbenzene)
Product LinenanoLC
Packing MaterialMonolith Polymer
Column TypeReversed Phase
PhaseReversed Phase
MaterialPolystyrene/Divinylbenzene
TypeAnalytical Column, NanoViper
Unit SizeEach
164542Full specifications
EachMonolith0.2 mm250 mmContact Us ›
Column FormatCapillary Column, NanoViper
Diameter (Metric)0.2 mm
Length (Metric)250 mm
Max. Pressure4350 psi (300 bar)
Particle SizeMonolith
pH1 to 10
Pore SizeMonolith
Temperature80°C
Stationary PhasePSDVB (Polystyrene Divinylbenzene)
Product LinenanoLC
Packing MaterialMonolith Polymer
Column TypeReversed Phase
PhaseReversed Phase
MaterialPolystyrene/Divinylbenzene
TypeAnalytical Column, NanoViper
Unit SizeEach
Showing 6 of 6
Benefit from:
  • High-speed peptide and protein separations (<15 min)
  • High sensitivity for LC/MS
  • High column-to-column reproducibility
  • Wide range of column I.D. and lengths available
  • Superior lifetime
  • Easy column installation using Thermo Scientific™ Dionex™ nanoViper™ Fingertight Fittings
  • Precolumns available to preconcentrate and desalt samples
You can also use Thermo Scientific™ PepSwift™ Trap Columns for preconcentration and desalting of samples consisting of peptides and proteins without negatively impacting the chromatographic performance or recovery of the compounds. Change the selectivity of the separation or improve the trapping efficiency by employing various ion-pairing agents in the loading solvent and/or mobile phases. The trap columns are used with Thermo Scientific™ PepSwift™ and ProSwift™ Monolithic Columns, which are for fast, high-resolution LC/MS separations in protein identification, biomarker discovery, and systems biology applications.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.