Thermo Scientific™
Accucore™ HILIC HPLC Columns
Catalog number: 17526-103030
Thermo Scientific™
Accucore™ HILIC HPLC Columns
Catalog number: 17526-103030
Offer enhanced retention of polar and hydrophilic analytes with Thermo Scientific™ Accucore™ HILIC LC Columns. Based on Core Enhanced Technology™, Accucore columns provide fast, high-resolution separations without the elevated backpressures required by sub-2μm particles.
Catalog Number
17526-103030
Unit Size
Each
Particle Size
2.6 μm
Diameter (Metric)
3 mm
Length (Metric)
100 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)3 mm
For Use WithHILIC, ESI-MS
Length (Metric)100 mm
Max. Pressure14500 psi (1000 bar)
Particle ShapeSpherical Solid Core
Particle Size2.6 μm
pH2 to 8
Pore Size80 Å
Surface Area130 m²/g
Temperature70°C
USP TypeL3
Stationary PhaseSilica
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Column TypeMixed Mode
PhaseNormal Phase and HILIC
Unit SizeEach
Catalog Number
Unit Size
-
Particle Size
2.6 μm
Diameter (Metric)
2.1 mm
Length (Metric)
30 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
For Use WithHILIC, ESI-MS
Length (Metric)30 mm
Max. Pressure14500 psi (1000 bar)
Particle ShapeSpherical Solid Core
Particle Size2.6 μm
pH2 to 8
Pore Size80 Å
Surface Area130 m²/g
Temperature70°C
USP TypeL3
Stationary PhaseSilica
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Column TypeMixed Mode
PhaseNormal Phase and HILIC
Unit Size-
Catalog Number
Unit Size
Each
Particle Size
2.6 μm
Diameter (Metric)
2.1 mm
Length (Metric)
50 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
For Use WithHILIC, ESI-MS
Length (Metric)50 mm
Max. Pressure14500 psi (1000 bar)
Particle ShapeSpherical Solid Core
Particle Size2.6 μm
pH2 to 8
Pore Size80 Å
Surface Area130 m²/g
Temperature70°C
USP TypeL3
Stationary PhaseSilica
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Column TypeMixed Mode
PhaseNormal Phase and HILIC
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
2.6 μm
Diameter (Metric)
3 mm
Length (Metric)
50 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)3 mm
For Use WithHILIC, ESI-MS
Length (Metric)50 mm
Max. Pressure14500 psi (1000 bar)
Particle ShapeSpherical Solid Core
Particle Size2.6 μm
pH2 to 8
Pore Size80 Å
Surface Area130 m²/g
Temperature70°C
USP TypeL3
Stationary PhaseSilica
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Column TypeMixed Mode
PhaseNormal Phase and HILIC
Unit SizeEach
Catalog Number
Unit Size
-
Particle Size
2.6 μm
Diameter (Metric)
4.6 mm
Length (Metric)
50 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
For Use WithHILIC, ESI-MS
Length (Metric)50 mm
Max. Pressure14500 psi (1000 bar)
Particle ShapeSpherical Solid Core
Particle Size2.6 μm
pH2 to 8
Pore Size80 Å
Surface Area130 m²/g
Temperature70°C
USP TypeL3
Stationary PhaseSilica
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Column TypeMixed Mode
PhaseNormal Phase and HILIC
Unit Size-
Catalog Number
Unit Size
Each
Particle Size
2.6 μm
Diameter (Metric)
2.1 mm
Length (Metric)
100 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
For Use WithHILIC, ESI-MS
Length (Metric)100 mm
Max. Pressure14500 psi (1000 bar)
Particle ShapeSpherical Solid Core
Particle Size2.6 μm
pH2 to 8
Pore Size80 Å
Surface Area130 m²/g
Temperature70°C
USP TypeL3
Stationary PhaseSilica
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Column TypeMixed Mode
PhaseNormal Phase and HILIC
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
2.6 μm
Diameter (Metric)
4.6 mm
Length (Metric)
100 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
For Use WithHILIC, ESI-MS
Length (Metric)100 mm
Max. Pressure14500 psi (1000 bar)
Particle ShapeSpherical Solid Core
Particle Size2.6 μm
pH2 to 8
Pore Size80 Å
Surface Area130 m²/g
Temperature70°C
USP TypeL3
Stationary PhaseSilica
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Column TypeMixed Mode
PhaseNormal Phase and HILIC
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
2.6 μm
Diameter (Metric)
2.1 mm
Length (Metric)
150 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
For Use WithHILIC, ESI-MS
Length (Metric)150 mm
Max. Pressure14500 psi (1000 bar)
Particle ShapeSpherical Solid Core
Particle Size2.6 μm
pH2 to 8
Pore Size80 Å
Surface Area130 m²/g
Temperature70°C
USP TypeL3
Stationary PhaseSilica
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Column TypeMixed Mode
PhaseNormal Phase and HILIC
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
2.6 μm
Diameter (Metric)
3 mm
Length (Metric)
150 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)3 mm
For Use WithHILIC, ESI-MS
Length (Metric)150 mm
Max. Pressure14500 psi (1000 bar)
Particle ShapeSpherical Solid Core
Particle Size2.6 μm
pH2 to 8
Pore Size80 Å
Surface Area130 m²/g
Temperature70°C
USP TypeL3
Stationary PhaseSilica
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Column TypeMixed Mode
PhaseNormal Phase and HILIC
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
2.6 μm
Diameter (Metric)
4.6 mm
Length (Metric)
150 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
For Use WithHILIC, ESI-MS
Length (Metric)150 mm
Max. Pressure14500 psi (1000 bar)
Particle ShapeSpherical Solid Core
Particle Size2.6 μm
pH2 to 8
Pore Size80 Å
Surface Area130 m²/g
Temperature70°C
USP TypeL3
Stationary PhaseSilica
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Column TypeMixed Mode
PhaseNormal Phase and HILIC
Unit SizeEach
Showing 10 of 13Show more
Catalog NumberSpecificationsUnit SizeParticle SizeDiameter (Metric)Length (Metric)Price (USD)
17526-103030Full specifications
Each 2.6 μm3 mm100 mmContact Us ›
Column FormatAnalytical Column
Diameter (Metric)3 mm
For Use WithHILIC, ESI-MS
Length (Metric)100 mm
Max. Pressure14500 psi (1000 bar)
Particle ShapeSpherical Solid Core
Particle Size2.6 μm
pH2 to 8
Pore Size80 Å
Surface Area130 m²/g
Temperature70°C
USP TypeL3
Stationary PhaseSilica
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Column TypeMixed Mode
PhaseNormal Phase and HILIC
Unit SizeEach
17526-032130Full specifications
-2.6 μm2.1 mm30 mmRequest A Quote
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
For Use WithHILIC, ESI-MS
Length (Metric)30 mm
Max. Pressure14500 psi (1000 bar)
Particle ShapeSpherical Solid Core
Particle Size2.6 μm
pH2 to 8
Pore Size80 Å
Surface Area130 m²/g
Temperature70°C
USP TypeL3
Stationary PhaseSilica
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Column TypeMixed Mode
PhaseNormal Phase and HILIC
Unit Size-
17526-052130Full specifications
Each2.6 μm2.1 mm50 mmContact Us ›
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
For Use WithHILIC, ESI-MS
Length (Metric)50 mm
Max. Pressure14500 psi (1000 bar)
Particle ShapeSpherical Solid Core
Particle Size2.6 μm
pH2 to 8
Pore Size80 Å
Surface Area130 m²/g
Temperature70°C
USP TypeL3
Stationary PhaseSilica
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Column TypeMixed Mode
PhaseNormal Phase and HILIC
Unit SizeEach
17526-053030Full specifications
Each2.6 μm3 mm50 mmContact Us ›
Column FormatAnalytical Column
Diameter (Metric)3 mm
For Use WithHILIC, ESI-MS
Length (Metric)50 mm
Max. Pressure14500 psi (1000 bar)
Particle ShapeSpherical Solid Core
Particle Size2.6 μm
pH2 to 8
Pore Size80 Å
Surface Area130 m²/g
Temperature70°C
USP TypeL3
Stationary PhaseSilica
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Column TypeMixed Mode
PhaseNormal Phase and HILIC
Unit SizeEach
17526-054630Full specifications
-2.6 μm4.6 mm50 mmRequest A Quote
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
For Use WithHILIC, ESI-MS
Length (Metric)50 mm
Max. Pressure14500 psi (1000 bar)
Particle ShapeSpherical Solid Core
Particle Size2.6 μm
pH2 to 8
Pore Size80 Å
Surface Area130 m²/g
Temperature70°C
USP TypeL3
Stationary PhaseSilica
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Column TypeMixed Mode
PhaseNormal Phase and HILIC
Unit Size-
17526-102130Full specifications
Each2.6 μm2.1 mm100 mmContact Us ›
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
For Use WithHILIC, ESI-MS
Length (Metric)100 mm
Max. Pressure14500 psi (1000 bar)
Particle ShapeSpherical Solid Core
Particle Size2.6 μm
pH2 to 8
Pore Size80 Å
Surface Area130 m²/g
Temperature70°C
USP TypeL3
Stationary PhaseSilica
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Column TypeMixed Mode
PhaseNormal Phase and HILIC
Unit SizeEach
17526-104630Full specifications
Each 2.6 μm4.6 mm100 mmContact Us ›
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
For Use WithHILIC, ESI-MS
Length (Metric)100 mm
Max. Pressure14500 psi (1000 bar)
Particle ShapeSpherical Solid Core
Particle Size2.6 μm
pH2 to 8
Pore Size80 Å
Surface Area130 m²/g
Temperature70°C
USP TypeL3
Stationary PhaseSilica
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Column TypeMixed Mode
PhaseNormal Phase and HILIC
Unit SizeEach
17526-152130Full specifications
Each2.6 μm2.1 mm150 mmContact Us ›
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
For Use WithHILIC, ESI-MS
Length (Metric)150 mm
Max. Pressure14500 psi (1000 bar)
Particle ShapeSpherical Solid Core
Particle Size2.6 μm
pH2 to 8
Pore Size80 Å
Surface Area130 m²/g
Temperature70°C
USP TypeL3
Stationary PhaseSilica
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Column TypeMixed Mode
PhaseNormal Phase and HILIC
Unit SizeEach
17526-153030Full specifications
Each 2.6 μm3 mm150 mmContact Us ›
Column FormatAnalytical Column
Diameter (Metric)3 mm
For Use WithHILIC, ESI-MS
Length (Metric)150 mm
Max. Pressure14500 psi (1000 bar)
Particle ShapeSpherical Solid Core
Particle Size2.6 μm
pH2 to 8
Pore Size80 Å
Surface Area130 m²/g
Temperature70°C
USP TypeL3
Stationary PhaseSilica
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Column TypeMixed Mode
PhaseNormal Phase and HILIC
Unit SizeEach
17526-154630Full specifications
Each2.6 μm4.6 mm150 mmContact Us ›
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
For Use WithHILIC, ESI-MS
Length (Metric)150 mm
Max. Pressure14500 psi (1000 bar)
Particle ShapeSpherical Solid Core
Particle Size2.6 μm
pH2 to 8
Pore Size80 Å
Surface Area130 m²/g
Temperature70°C
USP TypeL3
Stationary PhaseSilica
Product LineAccucore
Packing MaterialSpherical, Solid Core, Ultrapure Silica
Column TypeMixed Mode
PhaseNormal Phase and HILIC
Unit SizeEach
Showing 10 of 13Show more
  • Fast separations with excellent resolution and sensitivity
  • Lower backpressures so that UHPLC is not required
  • Enhanced Retention of Polar and Hydrophilic Analytes
  • Alternative Selectivity to C18 without Ion-pair or Derivatisation
  • Improved Sensitivity for MS detection
Ordering Information:
Other custom column dimensions are available. Please call your local Customer Service for more information.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.