Thermo Scientific™
TriPlus™ RSH Autosampler
Catalog number: 1R77010-0300
Thermo Scientific™
TriPlus™ RSH Autosampler
Catalog number: 1R77010-0300
Expand automated capabilities beyond liquid, headspace, and solid phase microextraction (SPME) injections. The advanced, built-in robotics of the Thermo Scientific™ TriPlus™ RSH Autosampler deliver exceptional precision, unprecedented sample handling flexibility, and reliable unattended operation. Accurate and repeatable sample preparation and introduction are essential to achieving the highest quality GC/GC-MS results. Automate the entire workflow, from standard dilution to injection, and improve the precision and reproducibility of your data, and your lab productivity.
Catalog Number
1R77010-0300
Unit Size
-
Type
TriPlus RSH Autosampler for Liquid and Headspace Injection with Standard X-Axis Length
Sample Capacity
4608 Well Plates, 840 × 0.5 mL or 0.7 mL Vials, 648 × 2 mL Sample Vials, 240 × 10 mL or 20 mL Vials (Liquid Sampling); 180 × 10 or 20 mL Vials (Headspace Sampling)
Price (USD)
Full specifications
For Use With (Equipment)For Liquid and Headspace Injection with Standard X-Axis Length
TypeTriPlus RSH Autosampler for Liquid and Headspace Injection with Standard X-Axis Length
Sample Capacity4608 Well Plates, 840 × 0.5 mL or 0.7 mL Vials, 648 × 2 mL Sample Vials, 240 × 10 mL or 20 mL Vials (Liquid Sampling); 180 × 10 or 20 mL Vials (Headspace Sampling)
Injection Mode8 Fully Customizable Method-Specific Injection Mode Presets
Injection SpeedSelectable from 0.1 μL/sec. to 2000 μL/sec. (Liquid Sampling); 1 to 100 mL/min, in 1 mL/min Increments (Headspace Sampling)
Injection Volume0.1 to 10,000 μL (Liquid Sampling); 0.1 to 5 mL Depending on Syringe (Headspace Sampling)
DescriptionBasic TriPlus RSH for liquid and headspace injection with standard X-axis length. Complete with liquid syringe tool, heaspace syringe tool, incubation oven, 2 x 10 µL syringes, 2 x 2.5 mL GT syringes, a standard washing station, 2 tray holders, 3 trays for 2 mL vials, one tray for 10/20 mL vials.
Tool ExchangeCompatible with Automatic Tool Change (ATC) or manual Tool Release Station
Unit Size-
Catalog Number
Unit Size
-
Type
TriPlus RSH Autosampler for Liquid Injection with Standard X-Axis Length
Sample Capacity
4608 Well Plates, 840 × 0.5 mL or 0.7 mL Vials, 648 × 2 mL Sample Vials, 240 × 10 mL or 20 mL Vials
Price (USD)
Full specifications
For Use With (Equipment)For Liquid Injection with Standard X-Axis Length
TypeTriPlus RSH Autosampler for Liquid Injection with Standard X-Axis Length
Sample Capacity4608 Well Plates, 840 × 0.5 mL or 0.7 mL Vials, 648 × 2 mL Sample Vials, 240 × 10 mL or 20 mL Vials
Injection Mode8 Fully Customizable Method-Specific Injection Mode Presets
Injection SpeedSelectable from 0.1 μL/sec. to 2000 μL/sec.
Injection Volume0.1 to 10,000 μL
DescriptionBasic TriPlus RSH for liquid injection with standard X-axis length. Complete with liquid syringe tool, 2 x 10 µL syringes, a standard washing station and a single tray holder with 3 trays for 2 mL vials.
Tool ExchangeCompatible with Automatic Tool Change (ATC) or manual Tool Release Station
Unit Size-
Catalog Number
Unit Size
-
Type
TriPlus RSH Autosampler for Headspace Injection with Standard X-Axis Length
Sample Capacity
180 × 10 or 20 mL Vials
Price (USD)
Full specifications
For Use With (Equipment)For Headspace Injection with Standard X-Axis Length
TypeTriPlus RSH Autosampler for Headspace Injection with Standard X-Axis Length
Sample Capacity180 × 10 or 20 mL Vials
Injection Mode8 Fully Customizable Method-Specific Injection Mode Presets
Injection Speed1 to 100 mL/min, in 1 mL/min Increments
Injection Volume0.1 to 5 mL Depending on Syringe
DescriptionBasic TriPlus RSH for headspace injection with standard X-axis length. Complete with headspace syringe tool, 2 x 2.5 mL GT syringes, a standard washing station and one tray holder with 1 tray for 10/20 mL vials.
Tool ExchangeCompatible with Automatic Tool Change (ATC) or manual Tool Release Station
Unit Size-
Catalog Number
Unit Size
-
Type
TriPlus RSH Autosampler for Liquid Injection with Extended X-Axis Length
Sample Capacity
6912 Well Plates, 1260 × 0.5 mL or 0.7 mL Vials, 972 × 2 mL Vials, 360 × 10 mL or 20 mL Vials
Price (USD)
Full specifications
For Use With (Equipment)For Liquid Injection with Extended X-Axis Length
TypeTriPlus RSH Autosampler for Liquid Injection with Extended X-Axis Length
Sample Capacity6912 Well Plates, 1260 × 0.5 mL or 0.7 mL Vials, 972 × 2 mL Vials, 360 × 10 mL or 20 mL Vials
PC Connection4608 Well Plates, 840 × 0.5 mL or 0.7 mL Vials, 648 × 2 mL Sample Vials, 240 × 10 mL or 20 mL Vials
Injection Mode8 Fully Customizable Method-Specific Injection Mode Presets
Injection SpeedSelectable from 0.1 μL/sec. to 2000 μL/sec.
Injection Volume0.1 to 10,000 μL
DescriptionBasic TriPlus RSH for liquid injection with extended X-axis length. Complete with liquid syringe tool, 2 x 10 µL syringes, a standard washing station and a single tray holder with 3 trays for 2 mL vials.
Tool ExchangeCompatible with Automatic Tool Change (ATC) or manual Tool Release Station
Unit Size-
Catalog Number
Unit Size
-
Type
TriPlus RSH Autosampler for Headspace Injection with Extended X-Axis Length
Sample Capacity
300 × 10 or 20 mL Vials
Price (USD)
Full specifications
For Use With (Equipment)For Headspace Injection with Extended X-Axis Length
TypeTriPlus RSH Autosampler for Headspace Injection with Extended X-Axis Length
Sample Capacity300 × 10 or 20 mL Vials
Injection Mode8 Fully Customizable Method-Specific Injection Mode Presets
Injection Speed1 to 100 mL/min, in 1 mL/min Increments
Injection Volume0.1 to 5 mL Depending on Syringe
DescriptionBasic TriPlus RSH for headspace injection with extended X-axis length. Complete with headspace syringe tool, 2 x 2.5 mL GT syringes, a standard washing station and one tray holder with 1 tray for 10/20 mL vials.
Tool ExchangeCompatible with Automatic Tool Change (ATC) or manual Tool Release Station
Unit Size-
Catalog Number
Unit Size
-
Type
TriPlus RSH Autosampler for Liquid and Headspace Injection with Extended X-Axis Length
Sample Capacity
6912 Well Plates, 1260 × 0.5 mL or 0.7 mL Vials, 972 × 2 mL Vials, 360 × 10 mL or 20 mL Vials (Liquid Sampling); 300 × 10 or 20 mL Vials (Headspace Sampling)
Price (USD)
Full specifications
TypeTriPlus RSH Autosampler for Liquid and Headspace Injection with Extended X-Axis Length
Sample Capacity6912 Well Plates, 1260 × 0.5 mL or 0.7 mL Vials, 972 × 2 mL Vials, 360 × 10 mL or 20 mL Vials (Liquid Sampling); 300 × 10 or 20 mL Vials (Headspace Sampling)
Injection Mode8 Fully Customizable Method-Specific Injection Mode Presets
Injection SpeedSelectable from 0.1 μL/sec. to 2000 μL/sec. (Liquid Sampling); 1 to 100 mL/min, in 1 mL/min Increments (Headspace Sampling)
Injection Volume0.1 to 10,000 μL (Liquid Sampling); 0.1 to 5 mL Depending on Syringe (Headspace Sampling)
DescriptionBasic TriPlus RSH for liquid and headspace injection with sextended X-axis length. Complete with liquid syringe tool, heaspace syringe tool, incubation oven, 2 x 10 µL syringes, 2 x 2.5 mL GT syringes, a standard washing station, 2 tray holders, 3 trays for 2 mL vials, one tray for 10/20 mL vials.
Tool ExchangeCompatible with Automatic Tool Change (ATC) or manual Tool Release Station
Unit Size-
Catalog Number
Unit Size
Each
Type
TriPlus RSH Autosampler for Sample Preparation with Standard X-Axis Length
Sample Capacity
4608 Well Plates, 840 × 0.5 mL or 0.7 mL Vials, 648 × 2 mL Sample Vials, 240 × 10 mL or 20 mL Vials (Liquid Sampling); 180 × 10 or 20 mL Vials (Headspace Sampling)
Price (USD)
Full specifications
TypeTriPlus RSH Autosampler for Sample Preparation with Standard X-Axis Length
Sample Capacity4608 Well Plates, 840 × 0.5 mL or 0.7 mL Vials, 648 × 2 mL Sample Vials, 240 × 10 mL or 20 mL Vials (Liquid Sampling); 180 × 10 or 20 mL Vials (Headspace Sampling)
Injection Mode8 Fully Customizable Method-Specific Injection Mode Presets
Injection SpeedSelectable from 0.1 μL/sec. to 2000 μL/sec. (Liquid Sampling); 1 to 100 mL/min, in 1 mL/min Increments (Headspace Sampling)
Injection Volume0.1 to 10,000 μL (Liquid Sampling); 0.1 to 5 mL Depending on Syringe (Headspace Sampling)
DescriptionBasic TriPlus RSH for Sample Preparation with standard X-axis length. Includes two liquid syringe tool kits, incubation oven, vortex mixer, ATC station, one standard washing station, one solvent station, one set of adapters for 2 mL vials and one tray holder with 1 tray for 10/20 mL vials.
Tool ExchangeAutomatic Tool Change (ATC) included
SoftwareTriPlus RSH Sampling Workflow Editor Software included
Unit SizeEach
Catalog Number
Unit Size
Each
Type
TriPlus RSH Autosampler for Sample Preparation with Extended X-Axis Length
Sample Capacity
6912 Well Plates, 1260 × 0.5 mL or 0.7 mL Vials, 972 × 2 mL Vials, 360 × 10 mL or 20 mL Vials (Liquid Sampling); 300 × 10 or 20 mL Vials (Headspace Sampling)
Price (USD)
Full specifications
TypeTriPlus RSH Autosampler for Sample Preparation with Extended X-Axis Length
Sample Capacity6912 Well Plates, 1260 × 0.5 mL or 0.7 mL Vials, 972 × 2 mL Vials, 360 × 10 mL or 20 mL Vials (Liquid Sampling); 300 × 10 or 20 mL Vials (Headspace Sampling)
Injection Mode8 Fully Customizable Method-Specific Injection Mode Presets
Injection SpeedSelectable from 0.1 μL/sec. to 2000 μL/sec. (Liquid Sampling); 1 to 100 mL/min, in 1 mL/min Increments (Headspace Sampling)
Injection Volume0.1 to 10,000 μL (Liquid Sampling); 0.1 to 5 mL Depending on Syringe (Headspace Sampling)
DescriptionBasic TriPlus RSH for Sample Preparation with extended X-axis length. Includes two liquid syringe tool kits, incubation oven, vortex mixer, ATC station, one standard washing station, one solvent station, one set of adapters for 2 mL vials and one tray holder with 1 tray for 10/20 mL vials.
Tool ExchangeAutomatic Tool Change (ATC) included
SoftwareTriPlus RSH Sampling Workflow Editor Software included
Unit SizeEach
Catalog Number
Unit Size
-
Type
TriPlus RSH Autosampler for SPME with Standard X-Axis Length
Sample Capacity
180 × 10 mL or 20 mL Vials
Price (USD)
Full specifications
For Use With (Equipment)SPME Arrow
TypeTriPlus RSH Autosampler for SPME with Standard X-Axis Length
Sample Capacity180 × 10 mL or 20 mL Vials
Injection SpeedNA
Injection VolumeNA
DescriptionBasic TriPlus RSH for SPME Arrow with standard X-axis length. Complete with SPME Arrow tool kit, incubation oven, Heatex Stirrer, a standard washing station and one tray holder with 1 tray for 10/20 mL vials.
Tool ExchangeCompatible with Automatic Tool Change (ATC) or manual Tool Release Station
Sample PreparationSolid Phase Microextraction (SPME)
Unit Size-
Catalog Number
Unit Size
-
Type
TriPlus RSH Autosampler for SPME with Extended X-Axis Length
Sample Capacity
300 × 10 mL or 20 mL Vials
Price (USD)
Full specifications
For Use With (Equipment)SPME Arrow
TypeTriPlus RSH Autosampler for SPME with Extended X-Axis Length
Sample Capacity300 × 10 mL or 20 mL Vials
Injection SpeedNA
Injection VolumeNA
DescriptionBasic TriPlus RSH for SPME Arrow with extended X-axis length. Complete with SPME Arrow tool kit, incubation oven, Heatex Stirrer, a standard washing station and one tray holder with 1 tray for 10/20 mL vials.
Tool ExchangeCompatible with Automatic Tool Change (ATC) or manual Tool Release Station
Sample PreparationSolid Phase Microextraction (SPME)
Unit Size-
Showing 10 of 10
Catalog NumberSpecificationsUnit SizeTypeSample CapacityPrice (USD)
1R77010-0300Full specifications
-TriPlus RSH Autosampler for Liquid and Headspace Injection with Standard X-Axis Length4608 Well Plates, 840 × 0.5 mL or 0.7 mL Vials, 648 × 2 mL Sample Vials, 240 × 10 mL or 20 mL Vials (Liquid Sampling); 180 × 10 or 20 mL Vials (Headspace Sampling)Request A Quote
For Use With (Equipment)For Liquid and Headspace Injection with Standard X-Axis Length
TypeTriPlus RSH Autosampler for Liquid and Headspace Injection with Standard X-Axis Length
Sample Capacity4608 Well Plates, 840 × 0.5 mL or 0.7 mL Vials, 648 × 2 mL Sample Vials, 240 × 10 mL or 20 mL Vials (Liquid Sampling); 180 × 10 or 20 mL Vials (Headspace Sampling)
Injection Mode8 Fully Customizable Method-Specific Injection Mode Presets
Injection SpeedSelectable from 0.1 μL/sec. to 2000 μL/sec. (Liquid Sampling); 1 to 100 mL/min, in 1 mL/min Increments (Headspace Sampling)
Injection Volume0.1 to 10,000 μL (Liquid Sampling); 0.1 to 5 mL Depending on Syringe (Headspace Sampling)
DescriptionBasic TriPlus RSH for liquid and headspace injection with standard X-axis length. Complete with liquid syringe tool, heaspace syringe tool, incubation oven, 2 x 10 µL syringes, 2 x 2.5 mL GT syringes, a standard washing station, 2 tray holders, 3 trays for 2 mL vials, one tray for 10/20 mL vials.
Tool ExchangeCompatible with Automatic Tool Change (ATC) or manual Tool Release Station
Unit Size-
1R77010-0100Full specifications
-TriPlus RSH Autosampler for Liquid Injection with Standard X-Axis Length4608 Well Plates, 840 × 0.5 mL or 0.7 mL Vials, 648 × 2 mL Sample Vials, 240 × 10 mL or 20 mL VialsRequest A Quote
For Use With (Equipment)For Liquid Injection with Standard X-Axis Length
TypeTriPlus RSH Autosampler for Liquid Injection with Standard X-Axis Length
Sample Capacity4608 Well Plates, 840 × 0.5 mL or 0.7 mL Vials, 648 × 2 mL Sample Vials, 240 × 10 mL or 20 mL Vials
Injection Mode8 Fully Customizable Method-Specific Injection Mode Presets
Injection SpeedSelectable from 0.1 μL/sec. to 2000 μL/sec.
Injection Volume0.1 to 10,000 μL
DescriptionBasic TriPlus RSH for liquid injection with standard X-axis length. Complete with liquid syringe tool, 2 x 10 µL syringes, a standard washing station and a single tray holder with 3 trays for 2 mL vials.
Tool ExchangeCompatible with Automatic Tool Change (ATC) or manual Tool Release Station
Unit Size-
1R77010-0200Full specifications
-TriPlus RSH Autosampler for Headspace Injection with Standard X-Axis Length180 × 10 or 20 mL VialsRequest A Quote
For Use With (Equipment)For Headspace Injection with Standard X-Axis Length
TypeTriPlus RSH Autosampler for Headspace Injection with Standard X-Axis Length
Sample Capacity180 × 10 or 20 mL Vials
Injection Mode8 Fully Customizable Method-Specific Injection Mode Presets
Injection Speed1 to 100 mL/min, in 1 mL/min Increments
Injection Volume0.1 to 5 mL Depending on Syringe
DescriptionBasic TriPlus RSH for headspace injection with standard X-axis length. Complete with headspace syringe tool, 2 x 2.5 mL GT syringes, a standard washing station and one tray holder with 1 tray for 10/20 mL vials.
Tool ExchangeCompatible with Automatic Tool Change (ATC) or manual Tool Release Station
Unit Size-
1R77010-0400Full specifications
-TriPlus RSH Autosampler for Liquid Injection with Extended X-Axis Length6912 Well Plates, 1260 × 0.5 mL or 0.7 mL Vials, 972 × 2 mL Vials, 360 × 10 mL or 20 mL VialsRequest A Quote
For Use With (Equipment)For Liquid Injection with Extended X-Axis Length
TypeTriPlus RSH Autosampler for Liquid Injection with Extended X-Axis Length
Sample Capacity6912 Well Plates, 1260 × 0.5 mL or 0.7 mL Vials, 972 × 2 mL Vials, 360 × 10 mL or 20 mL Vials
PC Connection4608 Well Plates, 840 × 0.5 mL or 0.7 mL Vials, 648 × 2 mL Sample Vials, 240 × 10 mL or 20 mL Vials
Injection Mode8 Fully Customizable Method-Specific Injection Mode Presets
Injection SpeedSelectable from 0.1 μL/sec. to 2000 μL/sec.
Injection Volume0.1 to 10,000 μL
DescriptionBasic TriPlus RSH for liquid injection with extended X-axis length. Complete with liquid syringe tool, 2 x 10 µL syringes, a standard washing station and a single tray holder with 3 trays for 2 mL vials.
Tool ExchangeCompatible with Automatic Tool Change (ATC) or manual Tool Release Station
Unit Size-
1R77010-0500Full specifications
-TriPlus RSH Autosampler for Headspace Injection with Extended X-Axis Length300 × 10 or 20 mL VialsRequest A Quote
For Use With (Equipment)For Headspace Injection with Extended X-Axis Length
TypeTriPlus RSH Autosampler for Headspace Injection with Extended X-Axis Length
Sample Capacity300 × 10 or 20 mL Vials
Injection Mode8 Fully Customizable Method-Specific Injection Mode Presets
Injection Speed1 to 100 mL/min, in 1 mL/min Increments
Injection Volume0.1 to 5 mL Depending on Syringe
DescriptionBasic TriPlus RSH for headspace injection with extended X-axis length. Complete with headspace syringe tool, 2 x 2.5 mL GT syringes, a standard washing station and one tray holder with 1 tray for 10/20 mL vials.
Tool ExchangeCompatible with Automatic Tool Change (ATC) or manual Tool Release Station
Unit Size-
1R77010-0600Full specifications
-TriPlus RSH Autosampler for Liquid and Headspace Injection with Extended X-Axis Length6912 Well Plates, 1260 × 0.5 mL or 0.7 mL Vials, 972 × 2 mL Vials, 360 × 10 mL or 20 mL Vials (Liquid Sampling); 300 × 10 or 20 mL Vials (Headspace Sampling)Request A Quote
TypeTriPlus RSH Autosampler for Liquid and Headspace Injection with Extended X-Axis Length
Sample Capacity6912 Well Plates, 1260 × 0.5 mL or 0.7 mL Vials, 972 × 2 mL Vials, 360 × 10 mL or 20 mL Vials (Liquid Sampling); 300 × 10 or 20 mL Vials (Headspace Sampling)
Injection Mode8 Fully Customizable Method-Specific Injection Mode Presets
Injection SpeedSelectable from 0.1 μL/sec. to 2000 μL/sec. (Liquid Sampling); 1 to 100 mL/min, in 1 mL/min Increments (Headspace Sampling)
Injection Volume0.1 to 10,000 μL (Liquid Sampling); 0.1 to 5 mL Depending on Syringe (Headspace Sampling)
DescriptionBasic TriPlus RSH for liquid and headspace injection with sextended X-axis length. Complete with liquid syringe tool, heaspace syringe tool, incubation oven, 2 x 10 µL syringes, 2 x 2.5 mL GT syringes, a standard washing station, 2 tray holders, 3 trays for 2 mL vials, one tray for 10/20 mL vials.
Tool ExchangeCompatible with Automatic Tool Change (ATC) or manual Tool Release Station
Unit Size-
1R77010-0150Full specifications
EachTriPlus RSH Autosampler for Sample Preparation with Standard X-Axis Length4608 Well Plates, 840 × 0.5 mL or 0.7 mL Vials, 648 × 2 mL Sample Vials, 240 × 10 mL or 20 mL Vials (Liquid Sampling); 180 × 10 or 20 mL Vials (Headspace Sampling)Request A Quote
TypeTriPlus RSH Autosampler for Sample Preparation with Standard X-Axis Length
Sample Capacity4608 Well Plates, 840 × 0.5 mL or 0.7 mL Vials, 648 × 2 mL Sample Vials, 240 × 10 mL or 20 mL Vials (Liquid Sampling); 180 × 10 or 20 mL Vials (Headspace Sampling)
Injection Mode8 Fully Customizable Method-Specific Injection Mode Presets
Injection SpeedSelectable from 0.1 μL/sec. to 2000 μL/sec. (Liquid Sampling); 1 to 100 mL/min, in 1 mL/min Increments (Headspace Sampling)
Injection Volume0.1 to 10,000 μL (Liquid Sampling); 0.1 to 5 mL Depending on Syringe (Headspace Sampling)
DescriptionBasic TriPlus RSH for Sample Preparation with standard X-axis length. Includes two liquid syringe tool kits, incubation oven, vortex mixer, ATC station, one standard washing station, one solvent station, one set of adapters for 2 mL vials and one tray holder with 1 tray for 10/20 mL vials.
Tool ExchangeAutomatic Tool Change (ATC) included
SoftwareTriPlus RSH Sampling Workflow Editor Software included
Unit SizeEach
1R77010-0450Full specifications
EachTriPlus RSH Autosampler for Sample Preparation with Extended X-Axis Length6912 Well Plates, 1260 × 0.5 mL or 0.7 mL Vials, 972 × 2 mL Vials, 360 × 10 mL or 20 mL Vials (Liquid Sampling); 300 × 10 or 20 mL Vials (Headspace Sampling)Request A Quote
TypeTriPlus RSH Autosampler for Sample Preparation with Extended X-Axis Length
Sample Capacity6912 Well Plates, 1260 × 0.5 mL or 0.7 mL Vials, 972 × 2 mL Vials, 360 × 10 mL or 20 mL Vials (Liquid Sampling); 300 × 10 or 20 mL Vials (Headspace Sampling)
Injection Mode8 Fully Customizable Method-Specific Injection Mode Presets
Injection SpeedSelectable from 0.1 μL/sec. to 2000 μL/sec. (Liquid Sampling); 1 to 100 mL/min, in 1 mL/min Increments (Headspace Sampling)
Injection Volume0.1 to 10,000 μL (Liquid Sampling); 0.1 to 5 mL Depending on Syringe (Headspace Sampling)
DescriptionBasic TriPlus RSH for Sample Preparation with extended X-axis length. Includes two liquid syringe tool kits, incubation oven, vortex mixer, ATC station, one standard washing station, one solvent station, one set of adapters for 2 mL vials and one tray holder with 1 tray for 10/20 mL vials.
Tool ExchangeAutomatic Tool Change (ATC) included
SoftwareTriPlus RSH Sampling Workflow Editor Software included
Unit SizeEach
1R77010-0800Full specifications
-TriPlus RSH Autosampler for SPME with Standard X-Axis Length180 × 10 mL or 20 mL VialsRequest A Quote
For Use With (Equipment)SPME Arrow
TypeTriPlus RSH Autosampler for SPME with Standard X-Axis Length
Sample Capacity180 × 10 mL or 20 mL Vials
Injection SpeedNA
Injection VolumeNA
DescriptionBasic TriPlus RSH for SPME Arrow with standard X-axis length. Complete with SPME Arrow tool kit, incubation oven, Heatex Stirrer, a standard washing station and one tray holder with 1 tray for 10/20 mL vials.
Tool ExchangeCompatible with Automatic Tool Change (ATC) or manual Tool Release Station
Sample PreparationSolid Phase Microextraction (SPME)
Unit Size-
1R77010-0900Full specifications
-TriPlus RSH Autosampler for SPME with Extended X-Axis Length300 × 10 mL or 20 mL VialsRequest A Quote
For Use With (Equipment)SPME Arrow
TypeTriPlus RSH Autosampler for SPME with Extended X-Axis Length
Sample Capacity300 × 10 mL or 20 mL Vials
Injection SpeedNA
Injection VolumeNA
DescriptionBasic TriPlus RSH for SPME Arrow with extended X-axis length. Complete with SPME Arrow tool kit, incubation oven, Heatex Stirrer, a standard washing station and one tray holder with 1 tray for 10/20 mL vials.
Tool ExchangeCompatible with Automatic Tool Change (ATC) or manual Tool Release Station
Sample PreparationSolid Phase Microextraction (SPME)
Unit Size-
Showing 10 of 10
The TriPlus RSH autosampler brings together technological innovations that further expand productivity, performance and versatility for GC and GC/MS applications. Automated high-precision sample handling eliminates the manual labor and downtime typically required for tasks as standard/sample dilutions, internal standard addition and derivatization.
Automatic Syringe Changeout
  • Automatic Tool Change (ATC) capability automates the use of different syringes for sample preparation steps or different injection techniques within the sequence.
Unprecedented Sample Capacity
  • Capacity for up to 972 2 mL sample vials combined with multiple 100 mL wash/waste bottles allow for long unattended operation maximizing productivity.
Vial Bottom Sensing Expands Application Range
  • Precise detection of vial bottoms enables the autosampler to accurately withdraw from sample amounts as little as 5 µL, enhancing data quality of sample-limited applications.
Flexible liquid sample handling
  • Optimized liquid injection modes and customizable syringe filling techniques, support a wide range of sample types and full GC inlet compatibility.
Automated headspace injection for volatile analysis
  • High temperature gas-tight syringe allows direct headspace injection into the GC inlet for increased injection volume flexibility.
  • Sample vial capacity of up to 300x 10 mL/20 mL vials and sample incubation overlapping, facilitate long unattended operation.
Automated, solvent-free SPME
  • Precise control of all steps—fiber preconditioning, adsorption and desorption, fiber washes—delivers maximum performance for SPME applications.
  • Boost productivity with the optimized SPME Arrow design. It offers optimized fiber geometry to achieve 10x superior sensitivity compared to classical SPME, 2x faster extraction time and superior robustness.
Automated Sample Preparation
The Triplus RSH combines highly versatile sample injection capability with automated sample preparation procedures, eliminating typical GCMS workflow bottlenecks.
Daily operations ranging from sample/standard dilution to more complex derivatization protocols, can be fully automated. Common steps include:
  • Sequential dilution
  • Calibration dilution
  • Standard addition
  • Batch sample derivatization
  • Sequential sample derivatization
  • Heating/mixing
  • Vortexing
  • Ambient temperature headspace
  • Ambient temperature SPME
TriPlus RSH Sampling Workflow Editor Software
In addition to its ready-to-use prep cycles to automate basic sample preparation workflows, the TriPlus RSH can be easily programmed for custom procedures using a visual programming software, which is based on an intuitive drag-and-drop user interface.
Extend productivity with the optional 1D barcode reader
  • Save time: Sample information is automatically read, negating logging by hand.
  • Increase efficiency: Reduce errors from manually recording information and tracking samples.
  • Streamline operations: Smooth and safe internal operations are improved with sample tracking and management.