Thermo Scientific™

Chromacol™ GOLD-Grade Inert Vials and Inserts

Catalog number: 2-SVG
Thermo Scientific™

Chromacol™ GOLD-Grade Inert Vials and Inserts

Catalog number: 2-SVG

Maximize your sample performance and have fewer failed assays with Thermo Scientific™ Chromacol™ GOLD-Grade Inert Glass Vials. These vials offer the most state-of-the-art technology in the industry for the analysis of critical low detection limit analytes, and when sample quality, reproducibility, security, and integrity are of upmost importance.

For easier sample handling and less risk of sample loss, the Chromacol GOLD-Grade Vials are the best-in-class choice for all chromatography users performing ultra-trace analysis.

 
Catalog Number
2-SVG
Unit Size
Pack of 500
Closure Size
8 mm
Description
2mL Screw Thread Vial, Flat Bottom, GOLD Grade Glass, Clear
Price (USD)
Full specifications
Description2mL Screw Thread Vial, Flat Bottom, GOLD Grade Glass, Clear
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)2.0 mL
Volume (Metric) Residual<170 μL
Product LineChromacol Gold
Height (Metric)32 mm
Quantity500
Closure Size8 mm
Marking SpotNo
Unit SizePack of 500
Catalog Number
Unit Size
Pack of 500
Closure Size
8 mm
Description
0.1mL Sci-Vi Crimp Top Vial - GOLD Grade Glass
Price (USD)
Full specifications
Description0.1mL Sci-Vi Crimp Top Vial - GOLD Grade Glass
ColorClear
Diameter (Metric) Outer6 mm
MaterialGlass
Volume (Metric)125 μL
Volume (Metric) Residual<1 μL
Product LineChromacol Gold
Height (Metric)32 mm
Quantity500
Closure Size8 mm
Marking SpotNo
Unit SizePack of 500
Catalog Number
Unit Size
Pack of 500
Closure Size
8 mm
Description
0.3mL Crimp Top, Round Bottom Vial, Clear Gold Grade
Price (USD)
Full specifications
Description0.3mL Crimp Top, Round Bottom Vial, Clear Gold Grade
ColorClear
Diameter (Metric) Outer6 mm
MaterialGlass
Volume (Metric)325 μL
Volume (Metric) Residual<5 mL
Product LineChromacol Gold
Height (Metric)32 mm
Quantity500
Closure Size8 mm
Marking SpotNo
Unit SizePack of 500
Catalog Number
Unit Size
Pack of 500
Closure Size
8 mm
Description
0.2mL Sci-Vi Crimp Top Vial - GOLD Grade Glass
Price (USD)
Full specifications
Description0.2mL Sci-Vi Crimp Top Vial - GOLD Grade Glass
ColorClear
Diameter (Metric) Outer6 mm
MaterialGlass
Volume (Metric)250 μL
Volume (Metric) Residual<5 μL
Product LineChromacol Gold
Height (Metric)32 mm
Quantity500
Closure Size8 mm
Marking SpotNo
Unit SizePack of 500
Catalog Number
Unit Size
Pack of 500
Closure Size
8 mm
Description
8-425 Screw Thread 1.1mL Vial - GOLD Grade Glass
Price (USD)
Full specifications
Description8-425 Screw Thread 1.1mL Vial - GOLD Grade Glass
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)1.2 mL
Volume (Metric) Residual<5 mL
Product LineChromacol Gold
Height (Metric)32 mm
Quantity500
Closure Size8 mm
Marking SpotNo
Unit SizePack of 500
Catalog Number
Unit Size
Pack of 100
Closure Size
9 mm
Description
2mL Wide Opening SureStop Screw Thread Vial with Write-on Patch, Flat Bottom, GOLD Grade Glass, Clear
Price (USD)
Full specifications
Description2mL Wide Opening SureStop Screw Thread Vial with Write-on Patch, Flat Bottom, GOLD Grade Glass, Clear
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)2.0 mL
Volume (Metric) Residual<170 μL
Product LineChromacol Gold
Height (Metric)32 mm
Quantity100
Closure Size9 mm
Marking SpotYes
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 500
Closure Size
9 mm
Description
0.2mL Wide Opening Screw Thread Vial, Insert Vial, Fused Insert-GOLD Grade Glass, Clear
Price (USD)
Full specifications
Description0.2mL Wide Opening Screw Thread Vial, Insert Vial, Fused Insert-GOLD Grade Glass, Clear
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)0.2 mL
Volume (Metric) Residual<2 μL
Product LineChromacol Gold
Height (Metric)32 mm
Quantity500
Closure Size9 mm
Marking SpotYes
Unit SizePack of 500
Catalog Number
Unit Size
Pack of 500
Closure Size
11 mm
Description
2mL Wide Opening Crimp Top Vial, Flat Bottom, GOLD Grade Glass, Clear
Price (USD)
Full specifications
Description2mL Wide Opening Crimp Top Vial, Flat Bottom, GOLD Grade Glass, Clear
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)2.0 mL
Volume (Metric) Residual<170 μL
Product LineChromacol Gold
Height (Metric)32 mm
Quantity500
Closure Size11 mm
Marking SpotYes
Unit SizePack of 500
Catalog Number
Unit Size
Pack of 500
Closure Size
11 mm
Description
11mm Crimp Top 1.1mL Vial, Wide Opening - GOLD Grade Glass
Price (USD)
Full specifications
Description11mm Crimp Top 1.1mL Vial, Wide Opening - GOLD Grade Glass
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)1.4 mL
Volume (Metric) Residual<5 μL
Product LineChromacol Gold
Height (Metric)32 mm
Quantity500
Closure Size11 mm
Marking SpotNo
Unit SizePack of 500
Catalog Number
Unit Size
Pack of 500
Closure Size
11 mm
Description
0.2mL Crimp Top Vial with Write-on Patch, Insert Vial, Fused Insert-GOLD Grade Glass, Clear
Price (USD)
Full specifications
Description0.2mL Crimp Top Vial with Write-on Patch, Insert Vial, Fused Insert-GOLD Grade Glass, Clear
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)0.2 mL
Volume (Metric) Residual<2 μL
Product LineChromacol Gold
Height (Metric)32 mm
Quantity500
Closure Size11 mm
Marking SpotYes
Unit SizePack of 500
Showing 10 of 11Show more
Catalog NumberSpecificationsUnit SizeClosure SizeDescriptionPrice (USD)
2-SVGFull specifications
Pack of 5008 mm2mL Screw Thread Vial, Flat Bottom, GOLD Grade Glass, ClearRequest A Quote
Description2mL Screw Thread Vial, Flat Bottom, GOLD Grade Glass, Clear
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)2.0 mL
Volume (Metric) Residual<170 μL
Product LineChromacol Gold
Height (Metric)32 mm
Quantity500
Closure Size8 mm
Marking SpotNo
Unit SizePack of 500
01-CVGFull specifications
Pack of 5008 mm0.1mL Sci-Vi Crimp Top Vial - GOLD Grade GlassRequest A Quote
Description0.1mL Sci-Vi Crimp Top Vial - GOLD Grade Glass
ColorClear
Diameter (Metric) Outer6 mm
MaterialGlass
Volume (Metric)125 μL
Volume (Metric) Residual<1 μL
Product LineChromacol Gold
Height (Metric)32 mm
Quantity500
Closure Size8 mm
Marking SpotNo
Unit SizePack of 500
03-CVGFull specifications
Pack of 5008 mm0.3mL Crimp Top, Round Bottom Vial, Clear Gold GradeContact Us ›
Description0.3mL Crimp Top, Round Bottom Vial, Clear Gold Grade
ColorClear
Diameter (Metric) Outer6 mm
MaterialGlass
Volume (Metric)325 μL
Volume (Metric) Residual<5 mL
Product LineChromacol Gold
Height (Metric)32 mm
Quantity500
Closure Size8 mm
Marking SpotNo
Unit SizePack of 500
02-CTVGFull specifications
Pack of 5008 mm0.2mL Sci-Vi Crimp Top Vial - GOLD Grade GlassRequest A Quote
Description0.2mL Sci-Vi Crimp Top Vial - GOLD Grade Glass
ColorClear
Diameter (Metric) Outer6 mm
MaterialGlass
Volume (Metric)250 μL
Volume (Metric) Residual<5 μL
Product LineChromacol Gold
Height (Metric)32 mm
Quantity500
Closure Size8 mm
Marking SpotNo
Unit SizePack of 500
1.1-STVGFull specifications
Pack of 5008 mm8-425 Screw Thread 1.1mL Vial - GOLD Grade GlassContact Us ›
Description8-425 Screw Thread 1.1mL Vial - GOLD Grade Glass
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)1.2 mL
Volume (Metric) Residual<5 mL
Product LineChromacol Gold
Height (Metric)32 mm
Quantity500
Closure Size8 mm
Marking SpotNo
Unit SizePack of 500
2-SVWGKFull specifications
Pack of 1009 mm2mL Wide Opening SureStop Screw Thread Vial with Write-on Patch, Flat Bottom, GOLD Grade Glass, ClearRequest A Quote
Description2mL Wide Opening SureStop Screw Thread Vial with Write-on Patch, Flat Bottom, GOLD Grade Glass, Clear
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)2.0 mL
Volume (Metric) Residual<170 μL
Product LineChromacol Gold
Height (Metric)32 mm
Quantity100
Closure Size9 mm
Marking SpotYes
Unit SizePack of 100
02-FISVGFull specifications
Pack of 5009 mm0.2mL Wide Opening Screw Thread Vial, Insert Vial, Fused Insert-GOLD Grade Glass, ClearRequest A Quote
Description0.2mL Wide Opening Screw Thread Vial, Insert Vial, Fused Insert-GOLD Grade Glass, Clear
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)0.2 mL
Volume (Metric) Residual<2 μL
Product LineChromacol Gold
Height (Metric)32 mm
Quantity500
Closure Size9 mm
Marking SpotYes
Unit SizePack of 500
2-CVGFull specifications
Pack of 50011 mm2mL Wide Opening Crimp Top Vial, Flat Bottom, GOLD Grade Glass, ClearRequest A Quote
Description2mL Wide Opening Crimp Top Vial, Flat Bottom, GOLD Grade Glass, Clear
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)2.0 mL
Volume (Metric) Residual<170 μL
Product LineChromacol Gold
Height (Metric)32 mm
Quantity500
Closure Size11 mm
Marking SpotYes
Unit SizePack of 500
1.1-CTVGFull specifications
Pack of 50011 mm11mm Crimp Top 1.1mL Vial, Wide Opening - GOLD Grade GlassContact Us ›
Description11mm Crimp Top 1.1mL Vial, Wide Opening - GOLD Grade Glass
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)1.4 mL
Volume (Metric) Residual<5 μL
Product LineChromacol Gold
Height (Metric)32 mm
Quantity500
Closure Size11 mm
Marking SpotNo
Unit SizePack of 500
02-FIVGFull specifications
Pack of 50011 mm0.2mL Crimp Top Vial with Write-on Patch, Insert Vial, Fused Insert-GOLD Grade Glass, ClearRequest A Quote
Description0.2mL Crimp Top Vial with Write-on Patch, Insert Vial, Fused Insert-GOLD Grade Glass, Clear
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)0.2 mL
Volume (Metric) Residual<2 μL
Product LineChromacol Gold
Height (Metric)32 mm
Quantity500
Closure Size11 mm
Marking SpotYes
Unit SizePack of 500
Showing 10 of 11Show more
Achieve confidence in results
  • Made of 1st hydrolytic class, special 33 expansion glass; meeting current USP requirements for pharmacopeias.
  • Ultra-low adsorption surface for critical polar compounds, enabling ultra trace-level analysis for strongly adsorbing analytes.
  • The high-class standard in Europe and Asia for decades.
  • Ultra-low pH shift due to the minimized surface activity.
  • Lowest alkaline content for less glass-wall interactions.
Increase productivity and save time
  • Highest recovery rates, even with critical compounds (e.g., trisubstituted N-atoms, tertiary amines) in trace concentrations.
  • Peaks show the same heights and symmetry and feature high recovery and reproducibility.
Reduce cost of analysis
  • Stable analysis and higher reproducibility means increased security and integrity of analytical results.
  • Reduced amount of buffer saves money.
  • No risk of run-stops during overnight autosampler runs.
  • Lot-independent, optimal autosampler fit.