Thermo Scientific™

Orion™ 2109XP Fluoride Analyzer

Catalog number: 2109RBA
Thermo Scientific™

Orion™ 2109XP Fluoride Analyzer

Catalog number: 2109RBA
Maximize up-time and keep your plant in pristine operation with the versatility and performance of the Thermo Scientific™ Orion™ 2109XP Fluoride Analyzer. It offers accuracy, reliability and ease of use to best meet the demands of your fluoride analysis.
 
Catalog Number
2109RBA
Unit Size
Each
Description
2109XP Reagent Bottle Adapter
Price (USD)
Full specifications
Description2109XP Reagent Bottle Adapter
Temperature (Metric) Operating5°C to 45°C (Ambient)
TypeFluoride Analyzer
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Fluoride Consumables Kit - 1 year operation
Price (USD)
Full specifications
DescriptionFluoride Consumables Kit - 1 year operation
Outputs0 to 20 mA; 4 to 20 mA
Temperature (Metric) Operating5°C to 35°C
TypeFluoride Analyzer
Response Time95% within 2 minutes of injecting a standard solution
Enclosure RatingWaterproof; IP66; NEMA 4X
Flow Rate40mL/min.
Relay Details3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
Alarm Output3
Analog Outputs2, Galvanically Isolated
Case MaterialValox 364
Certifications/ComplianceCE, CSA, UL, FCC Class A limits
IncludesReagent and diffusion tubing, calibration standards, sensing electrode and reference electrode with filling solution
Inlet Pressure8 to 100psig
Inputs85 to 132VAC; 170 to 264VAC
Sample Inlet1/4 in. NPTF tube fitting
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Reagentless Fluoride Consumables Kit - 1 year operation
Price (USD)
Full specifications
DescriptionReagentless Fluoride Consumables Kit - 1 year operation
Outputs0-20; 4-20 mA
Temperature (Metric) Operating5°C to 35°C
TypeFluoride Analyzer
Response Time95% within 2 minutes of injecting a standard solution
Enclosure RatingWaterproof; IP66; NEMA 4X
Flow Rate40 mL/min.
Relay Details3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
Alarm Output3
Analog Outputs210960XPF
Case MaterialValox 364
Certifications/Compliance210960XPF
IncludesDiffusion tubing, calibration standards, sensing electrode and reference electrode with filling solution
Inlet Pressure8 to 100 psig
Inputs85-132; 170-264 VAC
Sample Inlet1/4 NPTF tube fitting in.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
2109XP Fluoride Analyzer
Price (USD)
Full specifications
Current Loops2
Description2109XP Fluoride Analyzer
Frequency0 to 60 Hz
Height (English)26 in.
Height (Metric)65 cm
Length (English)11 in.
Length (Metric)27 cm
Outputs0 to 20 mA; 4 to 20 mA
Voltage85 to 132 VAC; 170 to 264 VAC
Temperature (Metric) Operating5°C to 35°C
TypeFluoride Analyzer
Accuracy±10% or 10 ppb, whichever is greater
Response Time95% within 2 minutes of injecting a standard solution
Width (English)17 in.
Width (Metric)45 cm
CalibrationDouble Known Addition
Enclosure RatingWaterproof; IP66; NEMA 4X
Flow Rate40mL/min.
Channels1
Communication DetailsModbus with Additional Module
Display TypeCustom backlit LCD with temperature, concentration, mV, error codes and menu driven prompts
Keypad TypeMembrane
Max. Relative Humidity5 to 95%, non-condensing
Mounting OptionsWall mount
Options for Channel 1Fluoride
Options for Channel 2Optional ph/ORP or Conductivity
Relay Details3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
Accuracy (mV)± (0.5 mV + 0.1%)
Accuracy (Temperature)±0.5°C
Alarm Output3
Amperage5A
Analog Outputs2, Galvanically Isolated
Case MaterialValox 364
Certifications/ComplianceCE, CSA, UL, FCC Class A limits
IncludesFluoride Analyzer Instrument Only
Inlet Pressure8 to 100psig
Inputs85-132; 170-264 VAC
Measurement Ranges10 ppb to 200 ppm
Sample Inlet1/4 in. NPTF tube fitting
Sample Streams1
Dimensions (L x W x H)11 x 17 x 26 in. (27 x 45 x 65cm)
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Fluoride Calibration Kit with Carrying Case
Price (USD)
Full specifications
DescriptionFluoride Calibration Kit with Carrying Case
Outputs0-20; 4-20 mA
Temperature (Metric) Operating5°C to 35°C
TypeFluoride Analyzer
Response Time95% within 2 minutes of injecting a standard solution
Enclosure RatingWaterproof; IP66; NEMA 4X
Flow Rate40 mL/min.
Relay Details3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
Alarm Output3
Analog Outputs2109XPCAL
Case MaterialValox 364
Certifications/Compliance2109XPCAL
Includes2109XPCAL
Inlet Pressure8 to 100 psig
Inputs85-132; 170-264 VAC
Sample Inlet1/4 NPTF tube fitting in.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Fluoride Electrode Kit
Price (USD)
Full specifications
DescriptionFluoride Electrode Kit
Outputs0-20; 4-20 mA
Temperature (Metric) Operating5°C to 35°C
TypeFluoride Analyzer
Response Time95% within 2 minutes of injecting a standard solution
Enclosure RatingWaterproof; IP66; NEMA 4X
Flow Rate40 mL/min.
Relay Details3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
Alarm Output3
Analog Outputs2, Galvanically Isolated
Case MaterialValox 364
Certifications/ComplianceCE, CSA, UL, FCC Class A limits
Includesfluoride electrode (100029), reference electrode (100056) and 2 oz. reference electrode filling solution (181073)
Inlet Pressure8 to 100 psig
Inputs85-132; 170-264 VAC
Sample Inlet1/4 NPTF tube fitting in.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
2109XP Fluoride Analyzer With Enclosure
Price (USD)
Full specifications
Current Loops2
Description2109XP Fluoride Analyzer With Enclosure
Frequency0 to 60 Hz
Height (English)26 in.
Height (Metric)65 cm
Length (English)11 in.
Length (Metric)27 cm
Outputs0 to 20 mA; 4 to 20 mA
Voltage85 to 132 VAC; 170 to 264 VAC
Temperature (Metric) Operating5°C to 35°C
TypeFluoride Analyzer
Accuracy±10% or 10 ppb, whichever is greater
Response Time95% within 2 minutes of injecting a standard solution
Width (English)17 in.
Width (Metric)45 cm
CalibrationDouble Known Addition
Enclosure RatingWaterproof; IP66; NEMA 4X
Flow Rate40mL/min.
Channels1
Communication DetailsModbus with Additional Module
Display TypeCustom backlit LCD with temperature, concentration, mV, error codes and menu driven prompts
Keypad TypeMembrane
Max. Relative Humidity5 to 95%, non-condensing
Mounting OptionsWall mount
Options for Channel 1Fluoride
Options for Channel 2Optional ph/ORP or Conductivity
Relay Details3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
Accuracy (mV)± (0.5 mV + 0.1%)
Accuracy (Temperature)±0.5°C
Alarm Output3
Amperage5A
Analog Outputs2, Galvanically Isolated
Case MaterialValox 364
Certifications/ComplianceCE, CSA, UL, FCC Class A limits
IncludesAnalyzer with Protective Enclosure
Inlet Pressure8 to 100psig
Inputs85-132; 170-264 VAC
Measurement Ranges10 ppb to 200 ppm
Sample Inlet1/4 in. NPTF tube fitting
Sample Streams1
Dimensions (L x W x H)11 x 17 x 26 in. (27 x 45 x 65cm)
Unit SizeEach
Catalog Number
Unit Size
Each
Description
2109XP Electronics Faceplate
Price (USD)
Full specifications
Description2109XP Electronics Faceplate
Temperature (Metric) Operating5°C to 45°C (Ambient)
TypeFluoride Analyzer
Unit SizeEach
Catalog Number
Unit Size
Each
Description
2109XP Service Kit
Price (USD)
Full specifications
Description2109XP Service Kit
Outputs0-20; 4-20 mA
Temperature (Metric) Operating5°C to 35°C
TypeFluoride Analyzer
Response Time95% within 2 minutes of injecting a standard solution
Enclosure RatingWaterproof; IP66; NEMA 4X
Flow Rate40 mL/min.
Relay Details3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
Alarm Output3
Analog Outputs2109XPSK
Case MaterialValox 364
Certifications/Compliance2109XPSK
Includeselectronics faceplate (2109XPEP), air pump assembly (2100PA), regulator assembly (2100RG), 2 electrode cables with gland assemblies for sensing and reference electrodes (2100EC), power supply assembly (2100PS) and fuse kit assembly (2100FK115 and 2100FK230)
Inlet Pressure8 to 100 psig
Inputs85-132; 170-264 VAC
Sample Inlet1/4 NPTF tube fitting in.
Unit SizeEach
Showing 9 of 9
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
2109RBAFull specifications
Each2109XP Reagent Bottle AdapterRequest A Quote
Description2109XP Reagent Bottle Adapter
Temperature (Metric) Operating5°C to 45°C (Ambient)
TypeFluoride Analyzer
Unit SizeEach
210960XPFull specifications
EachFluoride Consumables Kit - 1 year operationContact Us ›
DescriptionFluoride Consumables Kit - 1 year operation
Outputs0 to 20 mA; 4 to 20 mA
Temperature (Metric) Operating5°C to 35°C
TypeFluoride Analyzer
Response Time95% within 2 minutes of injecting a standard solution
Enclosure RatingWaterproof; IP66; NEMA 4X
Flow Rate40mL/min.
Relay Details3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
Alarm Output3
Analog Outputs2, Galvanically Isolated
Case MaterialValox 364
Certifications/ComplianceCE, CSA, UL, FCC Class A limits
IncludesReagent and diffusion tubing, calibration standards, sensing electrode and reference electrode with filling solution
Inlet Pressure8 to 100psig
Inputs85 to 132VAC; 170 to 264VAC
Sample Inlet1/4 in. NPTF tube fitting
Unit SizeEach
210960XPFFull specifications
EachReagentless Fluoride Consumables Kit - 1 year operationRequest A Quote
DescriptionReagentless Fluoride Consumables Kit - 1 year operation
Outputs0-20; 4-20 mA
Temperature (Metric) Operating5°C to 35°C
TypeFluoride Analyzer
Response Time95% within 2 minutes of injecting a standard solution
Enclosure RatingWaterproof; IP66; NEMA 4X
Flow Rate40 mL/min.
Relay Details3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
Alarm Output3
Analog Outputs210960XPF
Case MaterialValox 364
Certifications/Compliance210960XPF
IncludesDiffusion tubing, calibration standards, sensing electrode and reference electrode with filling solution
Inlet Pressure8 to 100 psig
Inputs85-132; 170-264 VAC
Sample Inlet1/4 NPTF tube fitting in.
Unit SizeEach
2109XPFull specifications
Each2109XP Fluoride AnalyzerRequest A Quote
Current Loops2
Description2109XP Fluoride Analyzer
Frequency0 to 60 Hz
Height (English)26 in.
Height (Metric)65 cm
Length (English)11 in.
Length (Metric)27 cm
Outputs0 to 20 mA; 4 to 20 mA
Voltage85 to 132 VAC; 170 to 264 VAC
Temperature (Metric) Operating5°C to 35°C
TypeFluoride Analyzer
Accuracy±10% or 10 ppb, whichever is greater
Response Time95% within 2 minutes of injecting a standard solution
Width (English)17 in.
Width (Metric)45 cm
CalibrationDouble Known Addition
Enclosure RatingWaterproof; IP66; NEMA 4X
Flow Rate40mL/min.
Channels1
Communication DetailsModbus with Additional Module
Display TypeCustom backlit LCD with temperature, concentration, mV, error codes and menu driven prompts
Keypad TypeMembrane
Max. Relative Humidity5 to 95%, non-condensing
Mounting OptionsWall mount
Options for Channel 1Fluoride
Options for Channel 2Optional ph/ORP or Conductivity
Relay Details3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
Accuracy (mV)± (0.5 mV + 0.1%)
Accuracy (Temperature)±0.5°C
Alarm Output3
Amperage5A
Analog Outputs2, Galvanically Isolated
Case MaterialValox 364
Certifications/ComplianceCE, CSA, UL, FCC Class A limits
IncludesFluoride Analyzer Instrument Only
Inlet Pressure8 to 100psig
Inputs85-132; 170-264 VAC
Measurement Ranges10 ppb to 200 ppm
Sample Inlet1/4 in. NPTF tube fitting
Sample Streams1
Dimensions (L x W x H)11 x 17 x 26 in. (27 x 45 x 65cm)
Unit SizeEach
2109XPCALFull specifications
EachFluoride Calibration Kit with Carrying CaseContact Us ›
DescriptionFluoride Calibration Kit with Carrying Case
Outputs0-20; 4-20 mA
Temperature (Metric) Operating5°C to 35°C
TypeFluoride Analyzer
Response Time95% within 2 minutes of injecting a standard solution
Enclosure RatingWaterproof; IP66; NEMA 4X
Flow Rate40 mL/min.
Relay Details3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
Alarm Output3
Analog Outputs2109XPCAL
Case MaterialValox 364
Certifications/Compliance2109XPCAL
Includes2109XPCAL
Inlet Pressure8 to 100 psig
Inputs85-132; 170-264 VAC
Sample Inlet1/4 NPTF tube fitting in.
Unit SizeEach
2109XPEKFull specifications
EachFluoride Electrode KitContact Us ›
DescriptionFluoride Electrode Kit
Outputs0-20; 4-20 mA
Temperature (Metric) Operating5°C to 35°C
TypeFluoride Analyzer
Response Time95% within 2 minutes of injecting a standard solution
Enclosure RatingWaterproof; IP66; NEMA 4X
Flow Rate40 mL/min.
Relay Details3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
Alarm Output3
Analog Outputs2, Galvanically Isolated
Case MaterialValox 364
Certifications/ComplianceCE, CSA, UL, FCC Class A limits
Includesfluoride electrode (100029), reference electrode (100056) and 2 oz. reference electrode filling solution (181073)
Inlet Pressure8 to 100 psig
Inputs85-132; 170-264 VAC
Sample Inlet1/4 NPTF tube fitting in.
Unit SizeEach
2109XPENFull specifications
Each2109XP Fluoride Analyzer With EnclosureRequest A Quote
Current Loops2
Description2109XP Fluoride Analyzer With Enclosure
Frequency0 to 60 Hz
Height (English)26 in.
Height (Metric)65 cm
Length (English)11 in.
Length (Metric)27 cm
Outputs0 to 20 mA; 4 to 20 mA
Voltage85 to 132 VAC; 170 to 264 VAC
Temperature (Metric) Operating5°C to 35°C
TypeFluoride Analyzer
Accuracy±10% or 10 ppb, whichever is greater
Response Time95% within 2 minutes of injecting a standard solution
Width (English)17 in.
Width (Metric)45 cm
CalibrationDouble Known Addition
Enclosure RatingWaterproof; IP66; NEMA 4X
Flow Rate40mL/min.
Channels1
Communication DetailsModbus with Additional Module
Display TypeCustom backlit LCD with temperature, concentration, mV, error codes and menu driven prompts
Keypad TypeMembrane
Max. Relative Humidity5 to 95%, non-condensing
Mounting OptionsWall mount
Options for Channel 1Fluoride
Options for Channel 2Optional ph/ORP or Conductivity
Relay Details3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
Accuracy (mV)± (0.5 mV + 0.1%)
Accuracy (Temperature)±0.5°C
Alarm Output3
Amperage5A
Analog Outputs2, Galvanically Isolated
Case MaterialValox 364
Certifications/ComplianceCE, CSA, UL, FCC Class A limits
IncludesAnalyzer with Protective Enclosure
Inlet Pressure8 to 100psig
Inputs85-132; 170-264 VAC
Measurement Ranges10 ppb to 200 ppm
Sample Inlet1/4 in. NPTF tube fitting
Sample Streams1
Dimensions (L x W x H)11 x 17 x 26 in. (27 x 45 x 65cm)
Unit SizeEach
2109XPEPFull specifications
Each2109XP Electronics FaceplateRequest A Quote
Description2109XP Electronics Faceplate
Temperature (Metric) Operating5°C to 45°C (Ambient)
TypeFluoride Analyzer
Unit SizeEach
2109XPSKFull specifications
Each2109XP Service KitRequest A Quote
Description2109XP Service Kit
Outputs0-20; 4-20 mA
Temperature (Metric) Operating5°C to 35°C
TypeFluoride Analyzer
Response Time95% within 2 minutes of injecting a standard solution
Enclosure RatingWaterproof; IP66; NEMA 4X
Flow Rate40 mL/min.
Relay Details3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
Alarm Output3
Analog Outputs2109XPSK
Case MaterialValox 364
Certifications/Compliance2109XPSK
Includeselectronics faceplate (2109XPEP), air pump assembly (2100PA), regulator assembly (2100RG), 2 electrode cables with gland assemblies for sensing and reference electrodes (2100EC), power supply assembly (2100PS) and fuse kit assembly (2100FK115 and 2100FK230)
Inlet Pressure8 to 100 psig
Inputs85-132; 170-264 VAC
Sample Inlet1/4 NPTF tube fitting in.
Unit SizeEach
Showing 9 of 9

Features:

  • Uses a recognized EPA method for fluoride analysis using an ion selective electrode (ISE)
  • Eliminates problems with turbidity and suspended solids in samples
  • Provides true accuracy in measuring total fluoride, with a detection limit of 10ppb
  • Offers continuous fluoride monitoring in a wide variety of samples
  • Protects against over-fluoridation of samples with sensitive, selective, reliable and verifiable measurements
  • Provides early detection of fluoride
  • Limits unnecessary calibration cycles due to drift with superior Orion fluoride electrode technology
  • Minimizes operator time and maintenance without the use of complicated moving parts or pumps that often require frequent attention and expensive spare part expenditures

Advanced User Interface:

  • Detailed calibration, measurement and diagnostic logs for early diagnostic and action level notification
  • Configurable to your facility's desired level of performance
  • Password protected
  • Easy to install, allowing your plant to be up and running in minutes

Options:

  • Retrofitable to the Orion 1809AO panel mounting footprint
  • Available in several configurations including the analyzer instrument only or with a protective enclosure
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.