TriPlus™ 500 GC Headspace Autosampler
Catalog number: 25118000
TriPlus™ 500 GC Headspace Autosampler
Catalog number: 25118000
Powered by a completely new design, the Thermo Scientific™ TriPlus™ 500 Headspace Autosampler delivers reliability and robustness for testing laboratories performing volatiles analysis. Valve-and-loop technology and other innovative features ensure that this autosampler streamlines daily workflows, saving time with reliable unattended operation, optimizing sample throughput with expanded vial capability, and facilitating method portability with powerful software. Smartly integrated with the Thermo Scientific™ TRACE™ 1300 Series GC, this modular platform provides today’s analytical testing and services laboratories with the productivity needed to succeed.
Catalog Number
25118000
Unit Size
Each
Description
Vial loader and vial tray for upgrade from 12 to 120 vial capacity
Software
Thermo Scientific Chromeleon CDS, Thermo Scientific TraceFinder, and Thermo Scientific XCalibur Software
Injection Mode
Vial tray only
Price (USD)
Full specifications
For Use With (Equipment)TriPlus 500 GC Headspace Autosampler
TypeVial Loader and Vial Tray Upgrade
Sample Capacity120 vials
Injection ModeVial tray only
DescriptionVial loader and vial tray for upgrade from 12 to 120 vial capacity
SoftwareThermo Scientific Chromeleon CDS, Thermo Scientific TraceFinder, and Thermo Scientific XCalibur Software
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Vial tray for upgrade from 120 to 240 vial capacity
Software
Thermo Scientific Chromeleon CDS, Thermo Scientific TraceFinder, and Thermo Scientific XCalibur Software
Injection Mode
Vial tray only
Price (USD)
Full specifications
For Use With (Equipment)TriPlus 500 GC Headspace Autosampler
TypeVial Tray Upgrade
Sample Capacity240 vials
Injection ModeVial tray only
DescriptionVial tray for upgrade from 120 to 240 vial capacity
SoftwareThermo Scientific Chromeleon CDS, Thermo Scientific TraceFinder, and Thermo Scientific XCalibur Software
Unit SizeEach
Catalog Number
Unit Size
Each
Description
TriPlus 500 Headspace Autosampler with direct column connection
Software
Thermo Scientific Chromeleon CDS, Thermo Scientific TraceFinder, and Thermo Scientific XCalibur Software
Injection Mode
Standard; Multiple Headspace Extraction (MHE); Multiple Headspace Injection (MHI)
Price (USD)
Full specifications
TypeTriPlus 500 GC Headspace Autosampler
Sample Capacity12 vials, upgradable to 120 and 240 vials
Injection ModeStandard; Multiple Headspace Extraction (MHE); Multiple Headspace Injection (MHI)
DescriptionTriPlus 500 Headspace Autosampler with direct column connection
SoftwareThermo Scientific Chromeleon CDS, Thermo Scientific TraceFinder, and Thermo Scientific XCalibur Software
Unit SizeEach
Showing 3 of 3
Catalog NumberSpecificationsUnit SizeDescriptionSoftwareInjection ModePrice (USD)
25118000Full specifications
EachVial loader and vial tray for upgrade from 12 to 120 vial capacityThermo Scientific Chromeleon CDS, Thermo Scientific TraceFinder, and Thermo Scientific XCalibur SoftwareVial tray onlyRequest A Quote
For Use With (Equipment)TriPlus 500 GC Headspace Autosampler
TypeVial Loader and Vial Tray Upgrade
Sample Capacity120 vials
Injection ModeVial tray only
DescriptionVial loader and vial tray for upgrade from 12 to 120 vial capacity
SoftwareThermo Scientific Chromeleon CDS, Thermo Scientific TraceFinder, and Thermo Scientific XCalibur Software
Unit SizeEach
25118200Full specifications
EachVial tray for upgrade from 120 to 240 vial capacityThermo Scientific Chromeleon CDS, Thermo Scientific TraceFinder, and Thermo Scientific XCalibur SoftwareVial tray onlyRequest A Quote
For Use With (Equipment)TriPlus 500 GC Headspace Autosampler
TypeVial Tray Upgrade
Sample Capacity240 vials
Injection ModeVial tray only
DescriptionVial tray for upgrade from 120 to 240 vial capacity
SoftwareThermo Scientific Chromeleon CDS, Thermo Scientific TraceFinder, and Thermo Scientific XCalibur Software
Unit SizeEach
25118100Full specifications
EachTriPlus 500 Headspace Autosampler with direct column connectionThermo Scientific Chromeleon CDS, Thermo Scientific TraceFinder, and Thermo Scientific XCalibur SoftwareStandard; Multiple Headspace Extraction (MHE); Multiple Headspace Injection (MHI)Request A Quote
TypeTriPlus 500 GC Headspace Autosampler
Sample Capacity12 vials, upgradable to 120 and 240 vials
Injection ModeStandard; Multiple Headspace Extraction (MHE); Multiple Headspace Injection (MHI)
DescriptionTriPlus 500 Headspace Autosampler with direct column connection
SoftwareThermo Scientific Chromeleon CDS, Thermo Scientific TraceFinder, and Thermo Scientific XCalibur Software
Unit SizeEach
Showing 3 of 3
Reliability for analytical testing and services laboratories and versatility for research and method development.
The new design of the TriPlus 500 Headspace Autosampler incorporates innovative features for highly accurate analytical results, including:
New pneumatic controls for high precision and robustness
  • Best injection repeatability for consistent results at every run.
  • No carryover due to effective sample path purging.
  • Automatic leak check during injection to ensure safe runs.
Best sample integrity and ease-of-use via direct column connection
  • A shorter sample path for high inertness towards polar compounds.
  • Expanded high temperature applicability with higher recovery of high boiling point compounds.
  • Simplified method set up with no transfer line and injector temperature settings required.
 Compact yet scalable design for better laboratory efficiency and productivity
  • Compact HS-GC design saves 30% of valuable bench space.
  • Tailored throughput requirements with in-field scalability from 12 to 240 vials, requiring no additional space and no compromise on performance.
  • Faster sample equilibration with the new Quick Spin Shaking design for maximal sensitivity in a shorter cycle time.
 Comprehensive HS-GC configuration solution
  • Available with direct column connection or transfer line for maximum configuration flexibility.
  • When a direct column connection is not possible, the autosampler configured with a transfer line maintains high precision, efficient sample transfer.
  • When configured with a transfer line, the TriPlus 500 HS autosampler is ideal for  connecting to the Thermo Scientific™ TRACE™ 1310 GC with auxiliary oven or for connection to third-party GC instruments
  • The TriPlus 500 HS with transfer line allows as well combined headspace and liquid injection in the same column, through operation with the TriPlus 100 LS or TriPlus RSH autosampler on the same GC. 
Software and system checks for compliance and data integrity
  • Integrated with Thermo Scientific™ Chromeleon™ CDS software, Title 21 CFR Part 11-compliant.
  • Easy analytical system performance  qualification with the IQ/OQ/PQ Qualification Wizard and daily sequence performance checks with the System Suitability Test Wizard.