Thermo Scientific™
TraceGOLD™ TG-OCP I and II GC Columns
Catalog number: 26078-2120
Thermo Scientific™
TraceGOLD™ TG-OCP I and II GC Columns
Catalog number: 26078-2120
Analyze organochlorine pesticides and herbicides with ultralow bleed, superior sensitivity, excellent peak shape and the highest levels of reproducibility using Thermo Scientific™ TraceGOLD™ TG-OCP I and TG-OCP II gas chromatography columns.
Catalog Number
26078-2120
Unit Size
-
Length (Metric)
15 m
Film Thickness
0.5 µm
Diameter (Metric)
0.32 mm
Stationary Phase
TraceGOLD™ TG-OCP I
Price (USD)
Full specifications
Diameter (Metric)0.32 mm
Diameter (Metric) Inner0.32 mm
For Use With (Application)GC/MS for Organochlorine Pesticides, GC/MS for Herbicides
Stationary PhaseTraceGOLD™ TG-OCP I
Max. Temperature340°C
Film Thickness0.5 µm
Length (Metric)15 m
PolarityNon-Polar
Description15m x 0.32mm x 0.50um
Unit Size-
Catalog Number
Unit Size
-
Length (Metric)
10 m
Film Thickness
0.1 µm
Diameter (Metric)
0.1 mm
Stationary Phase
TraceGOLD™ TG-OCP I
Price (USD)
Full specifications
Diameter (Metric)0.1 mm
Diameter (Metric) Inner0.1 mm
For Use With (Application)GC/MS for Organochlorine Pesticides, GC/MS for Herbicides
Stationary PhaseTraceGOLD™ TG-OCP I
Max. Temperature340°C
Film Thickness0.1 µm
Length (Metric)10 m
PolarityNon-Polar
Description10m x 0.10mm x 0.10um
Unit Size-
Catalog Number
Unit Size
-
Length (Metric)
15 m
Film Thickness
0.25 µm
Diameter (Metric)
0.25 mm
Stationary Phase
TraceGOLD™ TG-OCP I
Price (USD)
Full specifications
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS for Organochlorine Pesticides, GC/MS for Herbicides
Stationary PhaseTraceGOLD™ TG-OCP I
Max. Temperature340°C
Film Thickness0.25 µm
Length (Metric)15 m
PolarityNon-Polar
Description15m x 0.25mm x 0.25um
Unit Size-
Catalog Number
Unit Size
Each
Length (Metric)
30 m
Film Thickness
0.25 µm
Diameter (Metric)
0.25 mm
Stationary Phase
TraceGOLD™ TG-OCP I
Price (USD)
Full specifications
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS for Organochlorine Pesticides, GC/MS for Herbicides
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-OCP I
Max. Temperature340°C
Film Thickness0.25 µm
Length (Metric)30 m
PolarityNon-Polar
Description30 m x 0.25 mm x 0.25 µm
Unit SizeEach
Catalog Number
Unit Size
-
Length (Metric)
10 m
Film Thickness
0.18 µm
Diameter (Metric)
0.18 mm
Stationary Phase
TraceGOLD™ TG-OCP I
Price (USD)
Full specifications
Diameter (Metric)0.18 mm
Diameter (Metric) Inner0.18 mm
For Use With (Application)GC/MS for Organochlorine Pesticides, GC/MS for Herbicides
Stationary PhaseTraceGOLD™ TG-OCP I
Max. Temperature340°C
Film Thickness0.18 µm
Length (Metric)10 m
PolarityNon-Polar
Description10m x 0.18mm x 0.18um
Unit Size-
Catalog Number
Unit Size
-
Length (Metric)
60 m
Film Thickness
0.25 µm
Diameter (Metric)
0.25 mm
Stationary Phase
TraceGOLD™ TG-OCP I
Price (USD)
Full specifications
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS for Organochlorine Pesticides, GC/MS for Herbicides
Stationary PhaseTraceGOLD™ TG-OCP I
Max. Temperature340°C
Film Thickness0.25 µm
Length (Metric)60 m
PolarityNon-Polar
Description60m x 0.25mm x 0.25um
Unit Size-
Catalog Number
Unit Size
-
Length (Metric)
15 m
Film Thickness
0.5 µm
Diameter (Metric)
0.53 mm
Stationary Phase
TraceGOLD™ TG-OCP I
Price (USD)
Full specifications
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
For Use With (Application)GC/MS for Organochlorine Pesticides, GC/MS for Herbicides
Stationary PhaseTraceGOLD™ TG-OCP I
Max. Temperature340°C
Film Thickness0.5 µm
Length (Metric)15 m
PolarityNon-Polar
Description15m x 0.53mm x 0.50um
Unit Size-
Catalog Number
Unit Size
-
Length (Metric)
30 m
Film Thickness
0.5 µm
Diameter (Metric)
0.53 mm
Stationary Phase
TraceGOLD™ TG-OCP I
Price (USD)
Full specifications
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
For Use With (Application)GC/MS for Organochlorine Pesticides, GC/MS for Herbicides
Stationary PhaseTraceGOLD™ TG-OCP I
Max. Temperature340°C
Film Thickness0.5 µm
Length (Metric)30 m
PolarityNon-Polar
Description30m x 0.53mm x 0.50um
Unit Size-
Catalog Number
Unit Size
Each
Length (Metric)
30 m
Film Thickness
0.32 µm
Diameter (Metric)
0.32 mm
Stationary Phase
TraceGOLD™ TG-OCP I
Price (USD)
Full specifications
Diameter (Metric)0.32 mm
Diameter (Metric) Inner0.32 mm
For Use With (Application)GC/MS for Organochlorine Pesticides, GC/MS for Herbicides
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-OCP I
Max. Temperature340°C
Film Thickness0.32 µm
Length (Metric)30 m
PolarityNon-Polar
Description30 m x 0.32 mm x 0.32 µm
Unit SizeEach
Catalog Number
Unit Size
Each
Length (Metric)
20 m
Film Thickness
0.18 µm
Diameter (Metric)
0.18 mm
Stationary Phase
TraceGOLD™ TG-OCP I
Price (USD)
Full specifications
Diameter (Metric)0.18 mm
Diameter (Metric) Inner0.18 mm
For Use With (Application)GC/MS for Organochlorine Pesticides, GC/MS for Herbicides
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-OCP I
Max. Temperature340°C
Film Thickness0.18 µm
Length (Metric)20 m
PolarityNon-Polar
Description20 m x 0.18 mm x 0.18 µm
Unit SizeEach
Showing 10 of 22Show more
Catalog NumberSpecificationsUnit SizeLength (Metric)Film ThicknessDiameter (Metric)Stationary PhasePrice (USD)
26078-2120Full specifications
- 15 m0.5 µm0.32 mmTraceGOLD™ TG-OCP IRequest A Quote
Diameter (Metric)0.32 mm
Diameter (Metric) Inner0.32 mm
For Use With (Application)GC/MS for Organochlorine Pesticides, GC/MS for Herbicides
Stationary PhaseTraceGOLD™ TG-OCP I
Max. Temperature340°C
Film Thickness0.5 µm
Length (Metric)15 m
PolarityNon-Polar
Description15m x 0.32mm x 0.50um
Unit Size-
26078-0200Full specifications
- 10 m0.1 µm0.1 mmTraceGOLD™ TG-OCP IRequest A Quote
Diameter (Metric)0.1 mm
Diameter (Metric) Inner0.1 mm
For Use With (Application)GC/MS for Organochlorine Pesticides, GC/MS for Herbicides
Stationary PhaseTraceGOLD™ TG-OCP I
Max. Temperature340°C
Film Thickness0.1 µm
Length (Metric)10 m
PolarityNon-Polar
Description10m x 0.10mm x 0.10um
Unit Size-
26078-1300Full specifications
- 15 m0.25 µm0.25 mmTraceGOLD™ TG-OCP IRequest A Quote
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS for Organochlorine Pesticides, GC/MS for Herbicides
Stationary PhaseTraceGOLD™ TG-OCP I
Max. Temperature340°C
Film Thickness0.25 µm
Length (Metric)15 m
PolarityNon-Polar
Description15m x 0.25mm x 0.25um
Unit Size-
26078-1420Full specifications
Each30 m0.25 µm0.25 mmTraceGOLD™ TG-OCP IContact Us ›
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS for Organochlorine Pesticides, GC/MS for Herbicides
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-OCP I
Max. Temperature340°C
Film Thickness0.25 µm
Length (Metric)30 m
PolarityNon-Polar
Description30 m x 0.25 mm x 0.25 µm
Unit SizeEach
26078-1510Full specifications
- 10 m0.18 µm0.18 mmTraceGOLD™ TG-OCP IRequest A Quote
Diameter (Metric)0.18 mm
Diameter (Metric) Inner0.18 mm
For Use With (Application)GC/MS for Organochlorine Pesticides, GC/MS for Herbicides
Stationary PhaseTraceGOLD™ TG-OCP I
Max. Temperature340°C
Film Thickness0.18 µm
Length (Metric)10 m
PolarityNon-Polar
Description10m x 0.18mm x 0.18um
Unit Size-
26078-1540Full specifications
- 60 m0.25 µm0.25 mmTraceGOLD™ TG-OCP IRequest A Quote
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS for Organochlorine Pesticides, GC/MS for Herbicides
Stationary PhaseTraceGOLD™ TG-OCP I
Max. Temperature340°C
Film Thickness0.25 µm
Length (Metric)60 m
PolarityNon-Polar
Description60m x 0.25mm x 0.25um
Unit Size-
26078-2130Full specifications
- 15 m0.5 µm0.53 mmTraceGOLD™ TG-OCP IRequest A Quote
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
For Use With (Application)GC/MS for Organochlorine Pesticides, GC/MS for Herbicides
Stationary PhaseTraceGOLD™ TG-OCP I
Max. Temperature340°C
Film Thickness0.5 µm
Length (Metric)15 m
PolarityNon-Polar
Description15m x 0.53mm x 0.50um
Unit Size-
26078-2250Full specifications
- 30 m0.5 µm0.53 mmTraceGOLD™ TG-OCP IRequest A Quote
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
For Use With (Application)GC/MS for Organochlorine Pesticides, GC/MS for Herbicides
Stationary PhaseTraceGOLD™ TG-OCP I
Max. Temperature340°C
Film Thickness0.5 µm
Length (Metric)30 m
PolarityNon-Polar
Description30m x 0.53mm x 0.50um
Unit Size-
26078-5760Full specifications
Each30 m0.32 µm0.32 mmTraceGOLD™ TG-OCP IContact Us ›
Diameter (Metric)0.32 mm
Diameter (Metric) Inner0.32 mm
For Use With (Application)GC/MS for Organochlorine Pesticides, GC/MS for Herbicides
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-OCP I
Max. Temperature340°C
Film Thickness0.32 µm
Length (Metric)30 m
PolarityNon-Polar
Description30 m x 0.32 mm x 0.32 µm
Unit SizeEach
26078-5780Full specifications
Each20 m0.18 µm0.18 mmTraceGOLD™ TG-OCP IRequest A Quote
Diameter (Metric)0.18 mm
Diameter (Metric) Inner0.18 mm
For Use With (Application)GC/MS for Organochlorine Pesticides, GC/MS for Herbicides
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-OCP I
Max. Temperature340°C
Film Thickness0.18 µm
Length (Metric)20 m
PolarityNon-Polar
Description20 m x 0.18 mm x 0.18 µm
Unit SizeEach
Showing 10 of 22Show more
A Leap Forward in Performance with Low Bleed and Superior Inertness:
  • This low-bleed, stable column produces consistent results; the low baseline noise enables improved limits of detection with enhanced resolution of low level analytes.
  • Minimizing column bleed extends the interval between scheduled maintenance, leading to higher productivity.
  • Reducing column bleed increases reproducibility of the stationary phase over time, resulting in greater run-to-run and batch-to-batch reproducibility.
  • Superior inertness ensures excellent peak shape and sensitivity, especially for highly active or difficult-to-analyze compounds.
Redefining Your Expectations for Gas Chromatography:
  • Low bleed with excellent signal-to-noise ratio, sensitivity and mass spectral integrity
  • Fast analysis time to facilitate full separation of chlorinated pesticides
  • Ideal for U.S. EPA methods 8081, 608 and CLP
Proprietary Formulation Facilitates Analysis of Difficult Compounds:
  • The acidic nature of the silanol groups prevalent within many GC columns can result in the adsorption of basic compounds, which frequently leads to peak tailing.
  • In TraceGOLD GC columns, basic functional groups are bonded onto the column surface to reduce the effects of tailing and improve run-to-run reproducibility and sensitivity.
Recommended Use:
  • Organochlorine pesticides
  • Herbicides
Similar To:
  • Rtx™-CLPesticides
  • Rtx™-CLPesticides2
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.