Thermo Scientific™
TraceGOLD TG-POLAR GC Columns
Catalog number: 26082-0490
Thermo Scientific™
TraceGOLD TG-POLAR GC Columns
Catalog number: 26082-0490
Refine your expectations for high-polarity GC/MS with the Thermo Scientific™ TraceGOLD™ TG-POLAR columns, which are specifically designed with a polymer and surface treatment to overcome traditional problems with high-polarity columns.
Engineered for Superior Performance
  • Highly polar phase, 90% biscyanopropyl/10% phenylcyanopropyl polysiloxane, not bonded
  • Strong dipole moment and high selectivity for cis/trans compounds or compounds with conjugated double bonds
  • Equivalent to U.S.P G8 and G48
Applications:
  • Cis/Trans fatty acid methyl esters (FAMEs)
  • Dioxins
Catalog Number
26082-0490
Unit Size
Each
Diameter (Metric)
0.53 mm
Film Thickness
0.1 µm
Length (Metric)
30 m
Price (USD)
Full specifications
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
For Use With (Application)GC/MS of Cis/Trans FAMEs, GC/MS of Dioxins
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
Film Thickness0.1 µm
Length (Metric)30 m
PolarityHighly Polar
Description0.1 um Thickness; 0.53 mm ID; 30 m Length
Unit SizeEach
Catalog Number
Unit Size
Each
Diameter (Metric)
0.25 mm
Film Thickness
0.1 µm
Length (Metric)
30 m
Price (USD)
Full specifications
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS of Cis/Trans FAMEs, GC/MS of Dioxins
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
Film Thickness0.1 µm
Length (Metric)30 m
PolarityHighly Polar
Description0.1 um Thickness; 0.25 mm ID; 30 m Length
Unit SizeEach
Catalog Number
Unit Size
Each
Diameter (Metric)
0.25 mm
Film Thickness
0.1 µm
Length (Metric)
60 m
Price (USD)
Full specifications
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS of Cis/Trans FAMEs, GC/MS of Dioxins
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
Film Thickness0.1 µm
Length (Metric)60 m
PolarityHighly Polar
Description0.1 um Thickness; 0.25 mm ID; 60 m Length
Unit SizeEach
Catalog Number
Unit Size
Each
Diameter (Metric)
0.53 mm
Film Thickness
0.1 µm
Length (Metric)
60 m
Price (USD)
Full specifications
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
For Use With (Application)GC/MS of Cis/Trans FAMEs, GC/MS of Dioxins
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
Film Thickness0.1 µm
Length (Metric)60 m
PolarityHighly Polar
Description0.1 um Thickness; 0.53 mm ID; 60 m Length
Unit SizeEach
Catalog Number
Unit Size
Each
Diameter (Metric)
0.25 mm
Film Thickness
0.1 µm
Length (Metric)
105 m
Price (USD)
Full specifications
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS of Cis/Trans FAMEs, GC/MS of Dioxins
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
Film Thickness0.1 µm
Length (Metric)105 m
PolarityHighly Polar
Description0.1 um Thickness; 0.25 mm ID; 105 m Length
Unit SizeEach
Catalog Number
Unit Size
Each 1
Diameter (Metric)
0.25 mm
Film Thickness
0.2 µm
Length (Metric)
30 m
Price (USD)
Full specifications
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS of Cis/Trans FAMEs, GC/MS of Dioxins
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
Film Thickness0.2 µm
Length (Metric)30 m
PolarityHighly Polar
Description0.2 um Thickness; 0.25 mm ID; 30 m Length
Unit SizeEach 1
Catalog Number
Unit Size
-
Diameter (Metric)
0.25 mm
Film Thickness
0.2 µm
Length (Metric)
60 m
Price (USD)
Full specifications
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS of Cis/Trans FAMEs, GC/MS of Dioxins
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
Film Thickness0.2 µm
Length (Metric)60 m
PolarityHighly Polar
Description0.2 um Thickness; 0.25 mm ID; 60 m Length
Unit Size-
Catalog Number
Unit Size
Each
Diameter (Metric)
0.25 mm
Film Thickness
0.2 µm
Length (Metric)
105 m
Price (USD)
Full specifications
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS of Cis/Trans FAMEs, GC/MS of Dioxins
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
Film Thickness0.2 µm
Length (Metric)105 m
PolarityHighly Polar
Description0.2 um Thickness; 0.25 mm ID; 105 m Length
Unit SizeEach
Catalog Number
Unit Size
-
Diameter (Metric)
0.32 mm
Film Thickness
0.2 µm
Length (Metric)
30 m
Price (USD)
Full specifications
Diameter (Metric)0.32 mm
Diameter (Metric) Inner0.32 mm
For Use With (Application)GC/MS of Cis/Trans FAMEs, GC/MS of Dioxins
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
Film Thickness0.2 µm
Length (Metric)30 m
PolarityHighly Polar
Description0.2 um Thickness; 0.32 mm ID; 30 m Length
Unit Size-
Catalog Number
Unit Size
Each
Diameter (Metric)
0.32 mm
Film Thickness
0.2 µm
Length (Metric)
60 m
Price (USD)
Full specifications
Diameter (Metric)0.32 mm
Diameter (Metric) Inner0.32 mm
For Use With (Application)GC/MS of Cis/Trans FAMEs, GC/MS of Dioxins
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
Film Thickness0.2 µm
Length (Metric)60 m
PolarityHighly Polar
Description0.2 um Thickness; 0.32 mm ID; 60 m Length
Unit SizeEach
Showing 10 of 14Show more
Catalog NumberSpecificationsUnit SizeDiameter (Metric)Film ThicknessLength (Metric)Price (USD)
26082-0490Full specifications
Each0.53 mm0.1 µm30 mRequest A Quote
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
For Use With (Application)GC/MS of Cis/Trans FAMEs, GC/MS of Dioxins
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
Film Thickness0.1 µm
Length (Metric)30 m
PolarityHighly Polar
Description0.1 um Thickness; 0.53 mm ID; 30 m Length
Unit SizeEach
26082-0470Full specifications
Each0.25 mm0.1 µm30 mRequest A Quote
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS of Cis/Trans FAMEs, GC/MS of Dioxins
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
Film Thickness0.1 µm
Length (Metric)30 m
PolarityHighly Polar
Description0.1 um Thickness; 0.25 mm ID; 30 m Length
Unit SizeEach
26082-0590Full specifications
Each0.25 mm0.1 µm60 mRequest A Quote
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS of Cis/Trans FAMEs, GC/MS of Dioxins
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
Film Thickness0.1 µm
Length (Metric)60 m
PolarityHighly Polar
Description0.1 um Thickness; 0.25 mm ID; 60 m Length
Unit SizeEach
26082-0610Full specifications
Each0.53 mm0.1 µm60 mRequest A Quote
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
For Use With (Application)GC/MS of Cis/Trans FAMEs, GC/MS of Dioxins
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
Film Thickness0.1 µm
Length (Metric)60 m
PolarityHighly Polar
Description0.1 um Thickness; 0.53 mm ID; 60 m Length
Unit SizeEach
26082-5000Full specifications
Each 0.25 mm0.1 µm105 mContact Us ›
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS of Cis/Trans FAMEs, GC/MS of Dioxins
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
Film Thickness0.1 µm
Length (Metric)105 m
PolarityHighly Polar
Description0.1 um Thickness; 0.25 mm ID; 105 m Length
Unit SizeEach
26082-5010Full specifications
Each 10.25 mm0.2 µm30 mContact Us ›
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS of Cis/Trans FAMEs, GC/MS of Dioxins
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
Film Thickness0.2 µm
Length (Metric)30 m
PolarityHighly Polar
Description0.2 um Thickness; 0.25 mm ID; 30 m Length
Unit SizeEach 1
26082-5020Full specifications
- 0.25 mm0.2 µm60 mRequest A Quote
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS of Cis/Trans FAMEs, GC/MS of Dioxins
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
Film Thickness0.2 µm
Length (Metric)60 m
PolarityHighly Polar
Description0.2 um Thickness; 0.25 mm ID; 60 m Length
Unit Size-
26082-5030Full specifications
Each 0.25 mm0.2 µm105 mContact Us ›
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS of Cis/Trans FAMEs, GC/MS of Dioxins
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
Film Thickness0.2 µm
Length (Metric)105 m
PolarityHighly Polar
Description0.2 um Thickness; 0.25 mm ID; 105 m Length
Unit SizeEach
26082-5040Full specifications
- 0.32 mm0.2 µm30 mRequest A Quote
Diameter (Metric)0.32 mm
Diameter (Metric) Inner0.32 mm
For Use With (Application)GC/MS of Cis/Trans FAMEs, GC/MS of Dioxins
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
Film Thickness0.2 µm
Length (Metric)30 m
PolarityHighly Polar
Description0.2 um Thickness; 0.32 mm ID; 30 m Length
Unit Size-
26082-5050Full specifications
Each0.32 mm0.2 µm60 mRequest A Quote
Diameter (Metric)0.32 mm
Diameter (Metric) Inner0.32 mm
For Use With (Application)GC/MS of Cis/Trans FAMEs, GC/MS of Dioxins
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
Film Thickness0.2 µm
Length (Metric)60 m
PolarityHighly Polar
Description0.2 um Thickness; 0.32 mm ID; 60 m Length
Unit SizeEach
Showing 10 of 14Show more
A Leap Forward in Performance with Low Bleed and Superior Inertness:
  • A low bleed, stable column produces consistent results.
  • Low baseline noise enables improved limits of detection with enhanced resolution of low-level analytes.
  • Minimizing column bleed extends column lifetime, leading to higher productivity.
  • Reducing column bleed increases reproducibility of the stationary phase over time, resulting in greater run-to-run and batch-to-batch reproducibility.
  • Superior intertness ensures excellent peak shape and sensitivity, especially for highly active or difficult-to-analyze compounds.
Alternative Uses:
  • Alcohols
  • Aldehydes
  • Biodiesel–FAMEs
Similar To:
  • DB-23
  • HP-23
  • Rtx™™-2330
  • SP™-2330
  • SP™-2380
  • SPB™-2560
  • HP-88
  • Silar™™ 90c
  • CP-Sil 88 FAME
  • CP-Sil 88
  • BPX 70
  • BPX 90
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.