Thermo Scientific™
Hypersil GOLD™ AX HPLC Columns
Catalog number: 26105-254630
Thermo Scientific™
Hypersil GOLD™ AX HPLC Columns
Catalog number: 26105-254630
Efficiently separate smaller proteins, peptides, anionic species and polar molecules with Thermo Scientific™ Hypersil GOLD™ AX LC Columns. The weak anion-exchange phase, created by a polymeric amine ligand bonded to highly pure base-deactivated silica, separates multiple charged analytes. The bonded phase offers additional chromatographic options, including hydrophilic interaction chromatography (HILIC), which is ideal for the separation of sugars and other highly polar species.
Catalog Number
26105-254630
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
4.6 mm
Length (Metric)
250 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Flow Rate1 mL/min
Length (Metric)250 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolymeric Amine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeMixed Mode
EndcappedNo
PhaseHILIC and Ion Exchange
TypeAnalytical Column
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
2.1 mm
Length (Metric)
50 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Flow Rate0.2 mL/min
Length (Metric)50 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolymeric Amine
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeMixed Mode
EndcappedNo
PhaseHILIC and Ion Exchange
TypeAnalytical Column
Unit SizeEach
Catalog Number
Unit Size
-
Particle Size
5 μm
Diameter (Metric)
4.6 mm
Length (Metric)
50 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Flow Rate1 mL/min
Length (Metric)50 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolymeric Amine
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeMixed Mode
EndcappedNo
PhaseHILIC and Ion Exchange
TypeAnalytical Column
Unit Size-
Catalog Number
Unit Size
-
Particle Size
5 μm
Diameter (Metric)
2.1 mm
Length (Metric)
100 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Flow Rate0.2 mL/min
Length (Metric)100 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolymeric Amine
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeMixed Mode
EndcappedNo
PhaseHILIC and Ion Exchange
TypeAnalytical Column
Unit Size-
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
3 mm
Length (Metric)
100 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)3 mm
Flow Rate0.4 mL/min
Length (Metric)100 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolymeric Amine
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeMixed Mode
EndcappedNo
PhaseHILIC and Ion Exchange
TypeAnalytical Column
Unit SizeEach
Catalog Number
Unit Size
-
Particle Size
5 μm
Diameter (Metric)
4.6 mm
Length (Metric)
100 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Flow Rate1 mL/min
Length (Metric)100 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolymeric Amine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeMixed Mode
EndcappedNo
PhaseHILIC and Ion Exchange
TypeAnalytical Column
Unit Size-
Catalog Number
Unit Size
-
Particle Size
5 μm
Diameter (Metric)
2.1 mm
Length (Metric)
150 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Flow Rate0.2 mL/min
Length (Metric)150 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolymeric Amine
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeMixed Mode
EndcappedNo
PhaseHILIC and Ion Exchange
TypeAnalytical Column
Unit Size-
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
4.6 mm
Length (Metric)
150 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Flow Rate1 mL/min
Length (Metric)150 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolymeric Amine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeMixed Mode
EndcappedNo
PhaseHILIC and Ion Exchange
TypeAnalytical Column
Unit SizeEach
Catalog Number
Unit Size
-
Particle Size
5 μm
Diameter (Metric)
2.1 mm
Length (Metric)
250 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Flow Rate0.2 mL/min
Length (Metric)250 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolymeric Amine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeMixed Mode
EndcappedNo
PhaseHILIC and Ion Exchange
TypeAnalytical Column
Unit Size-
Catalog Number
Unit Size
-
Particle Size
5 μm
Diameter (Metric)
3 mm
Length (Metric)
250 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)3 mm
Flow Rate0.4 mL/min
Length (Metric)250 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolymeric Amine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeMixed Mode
EndcappedNo
PhaseHILIC and Ion Exchange
TypeAnalytical Column
Unit Size-
Showing 10 of 31Show more
Catalog NumberSpecificationsUnit SizeParticle SizeDiameter (Metric)Length (Metric)Price (USD)
26105-254630Full specifications
Each5 μm4.6 mm250 mmContact Us ›
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Flow Rate1 mL/min
Length (Metric)250 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolymeric Amine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeMixed Mode
EndcappedNo
PhaseHILIC and Ion Exchange
TypeAnalytical Column
Unit SizeEach
26105-052130Full specifications
Each5 μm2.1 mm50 mmContact Us ›
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Flow Rate0.2 mL/min
Length (Metric)50 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolymeric Amine
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeMixed Mode
EndcappedNo
PhaseHILIC and Ion Exchange
TypeAnalytical Column
Unit SizeEach
26105-054630Full specifications
- 5 μm4.6 mm50 mmRequest A Quote
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Flow Rate1 mL/min
Length (Metric)50 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolymeric Amine
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeMixed Mode
EndcappedNo
PhaseHILIC and Ion Exchange
TypeAnalytical Column
Unit Size-
26105-102130Full specifications
- 5 μm2.1 mm100 mmRequest A Quote
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Flow Rate0.2 mL/min
Length (Metric)100 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolymeric Amine
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeMixed Mode
EndcappedNo
PhaseHILIC and Ion Exchange
TypeAnalytical Column
Unit Size-
26105-103030Full specifications
Each5 μm3 mm100 mmContact Us ›
Column FormatAnalytical Column
Diameter (Metric)3 mm
Flow Rate0.4 mL/min
Length (Metric)100 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolymeric Amine
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeMixed Mode
EndcappedNo
PhaseHILIC and Ion Exchange
TypeAnalytical Column
Unit SizeEach
26105-104630Full specifications
-5 μm4.6 mm100 mmRequest A Quote
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Flow Rate1 mL/min
Length (Metric)100 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolymeric Amine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeMixed Mode
EndcappedNo
PhaseHILIC and Ion Exchange
TypeAnalytical Column
Unit Size-
26105-152130Full specifications
- 5 μm2.1 mm150 mmRequest A Quote
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Flow Rate0.2 mL/min
Length (Metric)150 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolymeric Amine
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeMixed Mode
EndcappedNo
PhaseHILIC and Ion Exchange
TypeAnalytical Column
Unit Size-
26105-154630Full specifications
Each5 μm4.6 mm150 mmContact Us ›
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Flow Rate1 mL/min
Length (Metric)150 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolymeric Amine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeMixed Mode
EndcappedNo
PhaseHILIC and Ion Exchange
TypeAnalytical Column
Unit SizeEach
26105-252130Full specifications
-5 μm2.1 mm250 mmRequest A Quote
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Flow Rate0.2 mL/min
Length (Metric)250 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolymeric Amine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeMixed Mode
EndcappedNo
PhaseHILIC and Ion Exchange
TypeAnalytical Column
Unit Size-
26105-253030Full specifications
-5 μm3 mm250 mmRequest A Quote
Column FormatAnalytical Column
Diameter (Metric)3 mm
Flow Rate0.4 mL/min
Length (Metric)250 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolymeric Amine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeMixed Mode
EndcappedNo
PhaseHILIC and Ion Exchange
TypeAnalytical Column
Unit Size-
Showing 10 of 31Show more
Excellent Chromatographic Performance
  • Outstanding peak shape and selectivity
  • Superb stability under demanding pH conditions
  • Higher efficiency than polymer-based ion exchange columns
  • Improved speed with 1.9µm particle size columns
About the Hypersil GOLD Family of HPLC Columns
Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC or normal phase chromatography.
  • Excellent peak symmetry
  • Narrow peaks for outstanding efficiency
  • Enhanced sensitivity and improved resolution
  • Exceptional reproducibility for reliable chromatography, column after column
  • Solutions for high-throughput screening
  • 1.9, 3 and 5µm particle sizes for all your separation needs
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.