Thermo Scientific™
TraceGOLD™ TG-5MT Metal GC Columns
Catalog number: 26M982980
Thermo Scientific™
TraceGOLD™ TG-5MT Metal GC Columns
Catalog number: 26M982980
Give greater stability and inertness at elevated temperatures and when working with highly active compounds with Thermo Scientific™ TraceGOLD™ TG-5MT Metal GC Columns. It provides greater robustness under increased stress.
Catalog Number
26M982980
Unit Size
-
Length (Metric)
30 m
Diameter (Metric)
0.53 mm
Film Thickness
1 µm
Price (USD)
Full specifications
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
For Use With (Application)GC/MS for Semi-Volatiles, Phenols, and Amines, GC/MS for Residual Solvents and Solvent Impurities, GC/MS for Drugs of Abuse, Pesticides, PCB Cogeners, and Aroclor Mixes
Stationary PhaseTraceGOLD™ TG-5 MT
USP TypeG27, G36
Max. Temperature330/350°C (Standard Columns), 430°C (Metal Columns)
Particle Size1.00 μm
Temperature Range-60°C to +430°C
Film Thickness1 µm
Length (Metric)30 m
PolarityLow-Polar
Unit Size-
Catalog Number
Unit Size
Each
Length (Metric)
15 m
Diameter (Metric)
0.25 mm
Film Thickness
0.25 µm
Price (USD)
Full specifications
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS for Semi-Volatiles, Phenols, and Amines, GC/MS for Residual Solvents and Solvent Impurities, GC/MS for Drugs of Abuse, Pesticides, PCB Cogeners, and Aroclor Mixes
Stationary PhaseTraceGOLD™ TG-5 MT
USP TypeG27, G36
Max. Temperature330/350°C (Standard Columns), 430°C (Metal Columns)
Particle Size0.25 μm
Temperature Range-60°C to +430°C
Film Thickness0.25 µm
Length (Metric)15 m
PolarityLow-Polar
Unit SizeEach
Catalog Number
Unit Size
Each
Length (Metric)
30 m
Diameter (Metric)
0.25 mm
Film Thickness
0.25 µm
Price (USD)
Full specifications
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS for Semi-Volatiles, Phenols, and Amines, GC/MS for Residual Solvents and Solvent Impurities, GC/MS for Drugs of Abuse, Pesticides, PCB Cogeners, and Aroclor Mixes
Stationary PhaseTraceGOLD™ TG-5 MT
USP TypeG27, G36
Max. Temperature330/350°C (Standard Columns), 430°C (Metal Columns)
Particle Size0.25 μm
Temperature Range-60°C to +430°C
Film Thickness0.25 µm
Length (Metric)30 m
PolarityLow-Polar
Unit SizeEach
Catalog Number
Unit Size
-
Length (Metric)
30 m
Diameter (Metric)
0.25 mm
Film Thickness
1 µm
Price (USD)
Full specifications
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS for Semi-Volatiles, Phenols, and Amines, GC/MS for Residual Solvents and Solvent Impurities, GC/MS for Drugs of Abuse, Pesticides, PCB Cogeners, and Aroclor Mixes
Stationary PhaseTraceGOLD™ TG-5 MT
USP TypeG27, G36
Max. Temperature330/350°C (Standard Columns), 430°C (Metal Columns)
Particle Size1.00 μm
Temperature Range-60°C to +430°C
Film Thickness1 µm
Length (Metric)30 m
PolarityLow-Polar
Unit Size-
Catalog Number
Unit Size
-
Length (Metric)
30 m
Diameter (Metric)
0.25 mm
Film Thickness
0.5 µm
Price (USD)
Full specifications
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS for Semi-Volatiles, Phenols, and Amines, GC/MS for Residual Solvents and Solvent Impurities, GC/MS for Drugs of Abuse, Pesticides, PCB Cogeners, and Aroclor Mixes
Stationary PhaseTraceGOLD™ TG-5 MT
USP TypeG27, G36
Max. Temperature330/350°C (Standard Columns), 430°C (Metal Columns)
Particle Size0.50 μm
Temperature Range-60°C to +430°C
Film Thickness0.5 µm
Length (Metric)30 m
PolarityLow-Polar
Unit Size-
Catalog Number
Unit Size
-
Length (Metric)
15 m
Diameter (Metric)
0.53 mm
Film Thickness
1 µm
Price (USD)
Full specifications
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
For Use With (Application)GC/MS for Semi-Volatiles, Phenols, and Amines, GC/MS for Residual Solvents and Solvent Impurities, GC/MS for Drugs of Abuse, Pesticides, PCB Cogeners, and Aroclor Mixes
Stationary PhaseTraceGOLD™ TG-5 MT
USP TypeG27, G36
Max. Temperature330/350°C (Standard Columns), 430°C (Metal Columns)
Particle Size1.00 μm
Temperature Range-60°C to +430°C
Film Thickness1 µm
Length (Metric)15 m
PolarityLow-Polar
Unit Size-
Catalog Number
Unit Size
Each
Length (Metric)
60 m
Diameter (Metric)
0.25 mm
Film Thickness
0.25 µm
Price (USD)
Full specifications
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS for Semi-Volatiles, Phenols, and Amines, GC/MS for Residual Solvents and Solvent Impurities, GC/MS for Drugs of Abuse, Pesticides, PCB Cogeners, and Aroclor Mixes
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-5 MT
USP TypeG27, G36
Max. Temperature330/350°C (Standard Columns), 430°C (Metal Columns)
Particle Size0.25 μm
Temperature Range-60°C to +430°C
Film Thickness0.25 µm
Length (Metric)60 m
PolarityLow-Polar
Unit SizeEach
Catalog Number
Unit Size
-
Length (Metric)
30 m
Diameter (Metric)
0.53 mm
Film Thickness
0.5 µm
Price (USD)
Full specifications
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
For Use With (Application)GC/MS for Semi-Volatiles, Phenols, and Amines, GC/MS for Residual Solvents and Solvent Impurities, GC/MS for Drugs of Abuse, Pesticides, PCB Cogeners, and Aroclor Mixes
Stationary PhaseTraceGOLD™ TG-5 MT
USP TypeG27, G36
Max. Temperature330/350°C (Standard Columns), 430°C (Metal Columns)
Particle Size0.50 μm
Temperature Range-60°C to +430°C
Film Thickness0.5 µm
Length (Metric)30 m
PolarityLow-Polar
Unit Size-
Catalog Number
Unit Size
Each
Length (Metric)
30 m
Diameter (Metric)
0.53 mm
Film Thickness
1.5 µm
Price (USD)
Full specifications
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
For Use With (Application)GC/MS for Semi-Volatiles, Phenols, and Amines, GC/MS for Residual Solvents and Solvent Impurities, GC/MS for Drugs of Abuse, Pesticides, PCB Cogeners, and Aroclor Mixes
Stationary PhaseTraceGOLD™ TG-5 MT
USP TypeG27, G36
Max. Temperature330/350°C (Standard Columns), 430°C (Metal Columns)
Particle Size1.50 μm
Temperature Range-60°C to +430°C
Film Thickness1.5 µm
Length (Metric)30 m
PolarityLow-Polar
Unit SizeEach
Catalog Number
Unit Size
-
Length (Metric)
30 m
Diameter (Metric)
0.53 mm
Film Thickness
0.25 µm
Price (USD)
Full specifications
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
For Use With (Application)GC/MS for Semi-Volatiles, Phenols, and Amines, GC/MS for Residual Solvents and Solvent Impurities, GC/MS for Drugs of Abuse, Pesticides, PCB Cogeners, and Aroclor Mixes
Stationary PhaseTraceGOLD™ TG-5 MT
USP TypeG27, G36
Max. Temperature330/350°C (Standard Columns), 430°C (Metal Columns)
Particle Size0.25 μm
Temperature Range-60°C to +430°C
Film Thickness0.25 µm
Length (Metric)30 m
PolarityLow-Polar
Unit Size-
Showing 10 of 11Show more
Catalog NumberSpecificationsUnit SizeLength (Metric)Diameter (Metric)Film ThicknessPrice (USD)
26M982980Full specifications
- 30 m0.53 mm1 µmRequest A Quote
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
For Use With (Application)GC/MS for Semi-Volatiles, Phenols, and Amines, GC/MS for Residual Solvents and Solvent Impurities, GC/MS for Drugs of Abuse, Pesticides, PCB Cogeners, and Aroclor Mixes
Stationary PhaseTraceGOLD™ TG-5 MT
USP TypeG27, G36
Max. Temperature330/350°C (Standard Columns), 430°C (Metal Columns)
Particle Size1.00 μm
Temperature Range-60°C to +430°C
Film Thickness1 µm
Length (Metric)30 m
PolarityLow-Polar
Unit Size-
26M981300Full specifications
Each15 m0.25 mm0.25 µmRequest A Quote
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS for Semi-Volatiles, Phenols, and Amines, GC/MS for Residual Solvents and Solvent Impurities, GC/MS for Drugs of Abuse, Pesticides, PCB Cogeners, and Aroclor Mixes
Stationary PhaseTraceGOLD™ TG-5 MT
USP TypeG27, G36
Max. Temperature330/350°C (Standard Columns), 430°C (Metal Columns)
Particle Size0.25 μm
Temperature Range-60°C to +430°C
Film Thickness0.25 µm
Length (Metric)15 m
PolarityLow-Polar
Unit SizeEach
26M981420Full specifications
Each30 m0.25 mm0.25 µmRequest A Quote
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS for Semi-Volatiles, Phenols, and Amines, GC/MS for Residual Solvents and Solvent Impurities, GC/MS for Drugs of Abuse, Pesticides, PCB Cogeners, and Aroclor Mixes
Stationary PhaseTraceGOLD™ TG-5 MT
USP TypeG27, G36
Max. Temperature330/350°C (Standard Columns), 430°C (Metal Columns)
Particle Size0.25 μm
Temperature Range-60°C to +430°C
Film Thickness0.25 µm
Length (Metric)30 m
PolarityLow-Polar
Unit SizeEach
26M982960Full specifications
- 30 m0.25 mm1 µmRequest A Quote
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS for Semi-Volatiles, Phenols, and Amines, GC/MS for Residual Solvents and Solvent Impurities, GC/MS for Drugs of Abuse, Pesticides, PCB Cogeners, and Aroclor Mixes
Stationary PhaseTraceGOLD™ TG-5 MT
USP TypeG27, G36
Max. Temperature330/350°C (Standard Columns), 430°C (Metal Columns)
Particle Size1.00 μm
Temperature Range-60°C to +430°C
Film Thickness1 µm
Length (Metric)30 m
PolarityLow-Polar
Unit Size-
26M982230Full specifications
- 30 m0.25 mm0.5 µmRequest A Quote
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS for Semi-Volatiles, Phenols, and Amines, GC/MS for Residual Solvents and Solvent Impurities, GC/MS for Drugs of Abuse, Pesticides, PCB Cogeners, and Aroclor Mixes
Stationary PhaseTraceGOLD™ TG-5 MT
USP TypeG27, G36
Max. Temperature330/350°C (Standard Columns), 430°C (Metal Columns)
Particle Size0.50 μm
Temperature Range-60°C to +430°C
Film Thickness0.5 µm
Length (Metric)30 m
PolarityLow-Polar
Unit Size-
26M982860Full specifications
- 15 m0.53 mm1 µmRequest A Quote
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
For Use With (Application)GC/MS for Semi-Volatiles, Phenols, and Amines, GC/MS for Residual Solvents and Solvent Impurities, GC/MS for Drugs of Abuse, Pesticides, PCB Cogeners, and Aroclor Mixes
Stationary PhaseTraceGOLD™ TG-5 MT
USP TypeG27, G36
Max. Temperature330/350°C (Standard Columns), 430°C (Metal Columns)
Particle Size1.00 μm
Temperature Range-60°C to +430°C
Film Thickness1 µm
Length (Metric)15 m
PolarityLow-Polar
Unit Size-
26M981540Full specifications
Each60 m0.25 mm0.25 µmRequest A Quote
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
For Use With (Application)GC/MS for Semi-Volatiles, Phenols, and Amines, GC/MS for Residual Solvents and Solvent Impurities, GC/MS for Drugs of Abuse, Pesticides, PCB Cogeners, and Aroclor Mixes
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-5 MT
USP TypeG27, G36
Max. Temperature330/350°C (Standard Columns), 430°C (Metal Columns)
Particle Size0.25 μm
Temperature Range-60°C to +430°C
Film Thickness0.25 µm
Length (Metric)60 m
PolarityLow-Polar
Unit SizeEach
26M982250Full specifications
- 30 m0.53 mm0.5 µmRequest A Quote
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
For Use With (Application)GC/MS for Semi-Volatiles, Phenols, and Amines, GC/MS for Residual Solvents and Solvent Impurities, GC/MS for Drugs of Abuse, Pesticides, PCB Cogeners, and Aroclor Mixes
Stationary PhaseTraceGOLD™ TG-5 MT
USP TypeG27, G36
Max. Temperature330/350°C (Standard Columns), 430°C (Metal Columns)
Particle Size0.50 μm
Temperature Range-60°C to +430°C
Film Thickness0.5 µm
Length (Metric)30 m
PolarityLow-Polar
Unit Size-
26M983360Full specifications
Each30 m0.53 mm1.5 µmRequest A Quote
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
For Use With (Application)GC/MS for Semi-Volatiles, Phenols, and Amines, GC/MS for Residual Solvents and Solvent Impurities, GC/MS for Drugs of Abuse, Pesticides, PCB Cogeners, and Aroclor Mixes
Stationary PhaseTraceGOLD™ TG-5 MT
USP TypeG27, G36
Max. Temperature330/350°C (Standard Columns), 430°C (Metal Columns)
Particle Size1.50 μm
Temperature Range-60°C to +430°C
Film Thickness1.5 µm
Length (Metric)30 m
PolarityLow-Polar
Unit SizeEach
26M981440Full specifications
- 30 m0.53 mm0.25 µmRequest A Quote
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
For Use With (Application)GC/MS for Semi-Volatiles, Phenols, and Amines, GC/MS for Residual Solvents and Solvent Impurities, GC/MS for Drugs of Abuse, Pesticides, PCB Cogeners, and Aroclor Mixes
Stationary PhaseTraceGOLD™ TG-5 MT
USP TypeG27, G36
Max. Temperature330/350°C (Standard Columns), 430°C (Metal Columns)
Particle Size0.25 μm
Temperature Range-60°C to +430°C
Film Thickness0.25 µm
Length (Metric)30 m
PolarityLow-Polar
Unit Size-
Showing 10 of 11Show more
  • Can be used at elevated temperature without risk of bleed or breakdown
  • Excellent inertness when working with highly active compounds
  • Robust under stress
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.