Applied Biosystems™
HID Real-Time PCR Analysis Software v1.1
Catalog number: 4413973
Applied Biosystems™
HID Real-Time PCR Analysis Software v1.1
Catalog number: 4413973
Catalog Number
4413973
Unit Size
Price (USD)
Price: 11,310.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
4413973
Price: 11,310.00
Your Price:
-
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
The HID Real-Time PCR Analysis Software v1.1 was designed specifically for the 7500 Real-Time PCR System and for the Quantifiler® Human, Quantifiler® Y Human Male, and Quantifiler® Duo DNA Quantification Kits. Quantifiler®-specific templates and quality flags, as well as STR reaction and dilution calculation tools, enable streamlined data and sample assessment within the software. New features of the HID Real-Time PCR Analysis Software v1.1 include streamlined sample entry, enhanced reporting and export capability, and improved data analysis tools.