Thermo Scientific™
HyperSep™ Dispersive SPE Extraction Products (QuEChERS)
Catalog number: 60105-212
Thermo Scientific™
HyperSep™ Dispersive SPE Extraction Products (QuEChERS)
Catalog number: 60105-212
QuEChERS Dispersive SPE methods offer a convenient and effective approach for extraction and clean-up of pesticide residues in food and other complex matrices. The robust procedure offers a number of compelling advantages: high recoveries, accurate results, high sample throughput, low solvent and glassware usage, reduced labor and bench space and lower reagent costs.
Thermo Scientific™ HyperSep™ Dispersive SPE Extraction Products are available in a range of pre-prepared sorbent combinations, each containing the proper sorbents for optimum extraction of analytes using QuEChERS methods.
Catalog Number
60105-212
Unit Size
Pack of 250
Description
50 mL Centrifuge Tubes: 6000 mg Anhydrous Magnesium Sulfate, 1500 mg Sodium Chloride, 1500 mg Sodium Citrate Tribasic Dihydrate, 750 mg Disodium Citrate Sesquihydrate
Price (USD)
Full specifications
TypeQuEChERS Sample Prep
Description50 mL Centrifuge Tubes: 6000 mg Anhydrous Magnesium Sulfate, 1500 mg Sodium Chloride, 1500 mg Sodium Citrate Tribasic Dihydrate, 750 mg Disodium Citrate Sesquihydrate
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Sodium Chloride, Sodium Citrate Tribasic Dihydrate, Disodium Citrate Sesquihydrate
Volume (Metric)50 mL
For Use With (Application)QuEChERS Method
Unit SizePack of 250
Catalog Number
Unit Size
Pack of 250
Description
50 mL Centrifuge Tubes: 6000 mg Anhydrous Magnesium Sulfate, 1500 mg Anhydrous Sodium Acetate
Price (USD)
Full specifications
TypeQuEChERS Sample Prep
Description50 mL Centrifuge Tubes: 6000 mg Anhydrous Magnesium Sulfate, 1500 mg Anhydrous Sodium Acetate
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Anhydrous Sodium Acetate
Volume (Metric)50 mL
For Use With (Application)QuEChERS Method
Unit SizePack of 250
Catalog Number
Unit Size
Pack of 250
Description
50 mL Centrifuge Tubes: 4000 mg Anhydrous Magnesium Sulfate, 1000 mg Anhydrous Sodium Chloride
Price (USD)
Full specifications
TypeQuEChERS Sample Prep
Description50 mL Centrifuge Tubes: 4000 mg Anhydrous Magnesium Sulfate, 1000 mg Anhydrous Sodium Chloride
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Anhydrous Sodium Chloride
Volume (Metric)50 mL
For Use With (Application)QuEChERS Method
Unit SizePack of 250
Catalog Number
Unit Size
Pack of 250
Description
50 mL Centrifuge Tubes: 4000 mg Anhydrous Magnesium Sulfate, 1000 mg Sodium Chloride, 1000 mg Sodium Citrate Tribasic Dihydrate, 500 mg Disodium Citrate Sesquihydrate
Price (USD)
Full specifications
TypeQuEChERS Sample Prep
Description50 mL Centrifuge Tubes: 4000 mg Anhydrous Magnesium Sulfate, 1000 mg Sodium Chloride, 1000 mg Sodium Citrate Tribasic Dihydrate, 500 mg Disodium Citrate Sesquihydrate
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Sodium Chloride, Sodium Citrate Tribasic Dihydrate, Disodium Citrate Sesquihydrate
Volume (Metric)50 mL
For Use With (Application)QuEChERS Method
Unit SizePack of 250
Catalog Number
Unit Size
Pack of 25
Description
50 mL Centrifuge Tubes: 6000 mg Anhydrous Magnesium Sulfate, 1500 mg Anhydrous Sodium Acetate
Price (USD)
Full specifications
TypeQuEChERS Sample Prep
Description50 mL Centrifuge Tubes: 6000 mg Anhydrous Magnesium Sulfate, 1500 mg Anhydrous Sodium Acetate
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Anhydrous Sodium Acetate
Volume (Metric)50 mL
For Use With (Application)QuEChERS Method
Unit SizePack of 25
Catalog Number
Unit Size
Pack of 25
Description
50 mL Centrifuge Tubes: 4000 mg Anhydrous Magnesium Sulfate, 1000 mg Sodium Chloride, 1000 mg Sodium Citrate Tribasic Dihydrate, 500 mg Disodium Citrate Sesquihydrate
Price (USD)
Full specifications
TypeQuEChERS Sample Prep
Description50 mL Centrifuge Tubes: 4000 mg Anhydrous Magnesium Sulfate, 1000 mg Sodium Chloride, 1000 mg Sodium Citrate Tribasic Dihydrate, 500 mg Disodium Citrate Sesquihydrate
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Sodium Chloride, Sodium Citrate Tribasic Dihydrate, Disodium Citrate Sesquihydrate
Volume (Metric)50 mL
For Use With (Application)QuEChERS Method
Unit SizePack of 25
Catalog Number
Unit Size
Pack of 25
Description
50 mL Centrifuge Tubes: 4000 mg Anhydrous Magnesium Sulfate, 1000 mg Sodium Chloride
Price (USD)
Full specifications
TypeQuEChERS Sample Prep
Description50 mL Centrifuge Tubes: 4000 mg Anhydrous Magnesium Sulfate, 1000 mg Sodium Chloride
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Anhydrous Sodium Chloride
Volume (Metric)50 mL
For Use With (Application)QuEChERS Method
Unit SizePack of 25
Catalog Number
Unit Size
Pack of 25
Description
50 mL Centrifuge Tubes: 6000 mg Anhydrous Magnesium Sulfate, 1500 mg Sodium Chloride, 1500 mg Sodium Citrate Tribasic Dihydrate, 750 mg Disodium Citrate Sesquihydrate
Price (USD)
Full specifications
TypeQuEChERS Sample Prep
Description50 mL Centrifuge Tubes: 6000 mg Anhydrous Magnesium Sulfate, 1500 mg Sodium Chloride, 1500 mg Sodium Citrate Tribasic Dihydrate, 750 mg Disodium Citrate Sesquihydrate
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Sodium Chloride, Sodium Citrate Tribasic Dihydrate, Disodium Citrate Sesquihydrate
Volume (Metric)50 mL
For Use With (Application)QuEChERS Method
Unit SizePack of 25
Catalog Number
Unit Size
Pack of 250
Description
50 mL Centrifuge Tubes: 900 mg Anhydrous Magnesium Sulfate, 300 mg PSA, 150 mg GCB
Price (USD)
Full specifications
TypeQuEChERS Sample Prep
Description50 mL Centrifuge Tubes: 900 mg Anhydrous Magnesium Sulfate, 300 mg PSA, 150 mg GCB
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, PSA, GCB
Volume (Metric)50 mL
For Use With (Application)QuEChERS Method
Unit SizePack of 250
Catalog Number
Unit Size
Each
Description
50 mL Centrifuge Tubes: 1500 mg Magnesium Sulfate, 500 mg C18
Price (USD)
Full specifications
TypeQuEChERS Sample Prep
Description50 mL Centrifuge Tubes: 1500 mg Magnesium Sulfate, 500 mg C18
Product LineHyperSep
SorbentMagnesium Sulfate, C18
Volume (Metric)50 mL
For Use With (Application)QuEChERS Method
Unit SizeEach
Showing 10 of 11Show more
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
60105-212Full specifications
Pack of 25050 mL Centrifuge Tubes: 6000 mg Anhydrous Magnesium Sulfate, 1500 mg Sodium Chloride, 1500 mg Sodium Citrate Tribasic Dihydrate, 750 mg Disodium Citrate SesquihydrateContact Us ›
TypeQuEChERS Sample Prep
Description50 mL Centrifuge Tubes: 6000 mg Anhydrous Magnesium Sulfate, 1500 mg Sodium Chloride, 1500 mg Sodium Citrate Tribasic Dihydrate, 750 mg Disodium Citrate Sesquihydrate
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Sodium Chloride, Sodium Citrate Tribasic Dihydrate, Disodium Citrate Sesquihydrate
Volume (Metric)50 mL
For Use With (Application)QuEChERS Method
Unit SizePack of 250
60105-210Full specifications
Pack of 25050 mL Centrifuge Tubes: 6000 mg Anhydrous Magnesium Sulfate, 1500 mg Anhydrous Sodium AcetateContact Us ›
TypeQuEChERS Sample Prep
Description50 mL Centrifuge Tubes: 6000 mg Anhydrous Magnesium Sulfate, 1500 mg Anhydrous Sodium Acetate
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Anhydrous Sodium Acetate
Volume (Metric)50 mL
For Use With (Application)QuEChERS Method
Unit SizePack of 250
60105-211Full specifications
Pack of 25050 mL Centrifuge Tubes: 4000 mg Anhydrous Magnesium Sulfate, 1000 mg Anhydrous Sodium ChlorideContact Us ›
TypeQuEChERS Sample Prep
Description50 mL Centrifuge Tubes: 4000 mg Anhydrous Magnesium Sulfate, 1000 mg Anhydrous Sodium Chloride
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Anhydrous Sodium Chloride
Volume (Metric)50 mL
For Use With (Application)QuEChERS Method
Unit SizePack of 250
60105-216Full specifications
Pack of 25050 mL Centrifuge Tubes: 4000 mg Anhydrous Magnesium Sulfate, 1000 mg Sodium Chloride, 1000 mg Sodium Citrate Tribasic Dihydrate, 500 mg Disodium Citrate SesquihydrateContact Us ›
TypeQuEChERS Sample Prep
Description50 mL Centrifuge Tubes: 4000 mg Anhydrous Magnesium Sulfate, 1000 mg Sodium Chloride, 1000 mg Sodium Citrate Tribasic Dihydrate, 500 mg Disodium Citrate Sesquihydrate
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Sodium Chloride, Sodium Citrate Tribasic Dihydrate, Disodium Citrate Sesquihydrate
Volume (Metric)50 mL
For Use With (Application)QuEChERS Method
Unit SizePack of 250
60105-310Full specifications
Pack of 2550 mL Centrifuge Tubes: 6000 mg Anhydrous Magnesium Sulfate, 1500 mg Anhydrous Sodium AcetateContact Us ›
TypeQuEChERS Sample Prep
Description50 mL Centrifuge Tubes: 6000 mg Anhydrous Magnesium Sulfate, 1500 mg Anhydrous Sodium Acetate
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Anhydrous Sodium Acetate
Volume (Metric)50 mL
For Use With (Application)QuEChERS Method
Unit SizePack of 25
60105-316Full specifications
Pack of 2550 mL Centrifuge Tubes: 4000 mg Anhydrous Magnesium Sulfate, 1000 mg Sodium Chloride, 1000 mg Sodium Citrate Tribasic Dihydrate, 500 mg Disodium Citrate SesquihydrateContact Us ›
TypeQuEChERS Sample Prep
Description50 mL Centrifuge Tubes: 4000 mg Anhydrous Magnesium Sulfate, 1000 mg Sodium Chloride, 1000 mg Sodium Citrate Tribasic Dihydrate, 500 mg Disodium Citrate Sesquihydrate
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Sodium Chloride, Sodium Citrate Tribasic Dihydrate, Disodium Citrate Sesquihydrate
Volume (Metric)50 mL
For Use With (Application)QuEChERS Method
Unit SizePack of 25
60105-311Full specifications
Pack of 2550 mL Centrifuge Tubes: 4000 mg Anhydrous Magnesium Sulfate, 1000 mg Sodium ChlorideContact Us ›
TypeQuEChERS Sample Prep
Description50 mL Centrifuge Tubes: 4000 mg Anhydrous Magnesium Sulfate, 1000 mg Sodium Chloride
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Anhydrous Sodium Chloride
Volume (Metric)50 mL
For Use With (Application)QuEChERS Method
Unit SizePack of 25
60105-312Full specifications
Pack of 2550 mL Centrifuge Tubes: 6000 mg Anhydrous Magnesium Sulfate, 1500 mg Sodium Chloride, 1500 mg Sodium Citrate Tribasic Dihydrate, 750 mg Disodium Citrate SesquihydrateContact Us ›
TypeQuEChERS Sample Prep
Description50 mL Centrifuge Tubes: 6000 mg Anhydrous Magnesium Sulfate, 1500 mg Sodium Chloride, 1500 mg Sodium Citrate Tribasic Dihydrate, 750 mg Disodium Citrate Sesquihydrate
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Sodium Chloride, Sodium Citrate Tribasic Dihydrate, Disodium Citrate Sesquihydrate
Volume (Metric)50 mL
For Use With (Application)QuEChERS Method
Unit SizePack of 25
60105-237Full specifications
Pack of 25050 mL Centrifuge Tubes: 900 mg Anhydrous Magnesium Sulfate, 300 mg PSA, 150 mg GCBContact Us ›
TypeQuEChERS Sample Prep
Description50 mL Centrifuge Tubes: 900 mg Anhydrous Magnesium Sulfate, 300 mg PSA, 150 mg GCB
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, PSA, GCB
Volume (Metric)50 mL
For Use With (Application)QuEChERS Method
Unit SizePack of 250
60105-234Full specifications
Each50 mL Centrifuge Tubes: 1500 mg Magnesium Sulfate, 500 mg C18Request A Quote
TypeQuEChERS Sample Prep
Description50 mL Centrifuge Tubes: 1500 mg Magnesium Sulfate, 500 mg C18
Product LineHyperSep
SorbentMagnesium Sulfate, C18
Volume (Metric)50 mL
For Use With (Application)QuEChERS Method
Unit SizeEach
Showing 10 of 11Show more
HyperSep Dispersive SPE Extraction Products use magnesium sulfate to aid extraction, along with either sodium chloride, sodium citrate, or anhydrous sodium acetate for base-sensitive compounds (e.g., folpet or captan). They are supplied in a 50mL polypropylene centrifuge tube for convenient extractions.
The dispersive SPE products are pre-prepared and optimized for the method at hand, so you will:
  • Save valuable preparation time
  • Achieve more consistent analytical results due to consistency in products and rigorous quality control
  • Get cleaner extracts and lower peak counts as a result of using highest quality, pre-prepared products
QuEChERS Technique
  • Developed by the USDA Eastern Regional Research Center in 2003, the QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) technique is increasingly becoming the technique of choice for extraction and clean-up of pesticide residues.
The QuEChERS method is a two-step process: extraction followed by clean-up. There are four variations of the QuEChERS method currently in use:
  • The original QuEChERS method uses sodium chloride to enhance extraction
  • Dispersive AOAC 2007.01 uses sodium acetate as a buffer, replacing sodium chloride
  • The Dual-Phase variation introduces PSA and GCB to remove high levels of chlorophyll and plant sterols in the final extract without the loss of planar pesticides (polar aromatics) using an acetone:toluene solvent mix (3:1)
  • European Version is similar to the AOAC method, except the extraction uses sodium chloride, sodium citrate dihydrate and disodium citrate sesquihydrate instead of sodium acetate
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.