Thermo Scientific™
HyperSep™ Dispersive SPE Clean-up Products (QuEChERS)
Catalog number: 60105-323
Thermo Scientific™
HyperSep™ Dispersive SPE Clean-up Products (QuEChERS)
Catalog number: 60105-323
QuEChERS Dispersive SPE methods offer a convenient and effective approach for extraction and clean-up of pesticide residues in food and other complex matrices. The robust procedure offers a number of compelling advantages: high recoveries, accurate results, high sample throughput, low solvent and glassware usage, reduced labor and bench space and lower reagent costs.
Thermo Scientific™ HyperSep Dispersive SPE Clean-Up Products are available in a range of pre-prepared sorbent combinations, each containing the proper sorbents for optimum clean-up of analytes extracted using QuEChERS methods.
Catalog Number
60105-323
Unit Size
Pack of,10
Description
150 mg MgSO4, 50 mg PSA, 50 mg C18, 50 mg GCB
For Use With (Application)
QuEChERS Method
Price (USD)
Full specifications
TypeQuEChERS Clean-up
Description150 mg MgSO4, 50 mg PSA, 50 mg C18, 50 mg GCB
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Primary/Secondary Amine, Endcapped C18, Graphitized Carbon Black
Volume (Metric)2 mL Centrifuge Tubes
For Use With (Application)QuEChERS Method
Unit SizePack of,10
Catalog Number
Unit Size
Pack of 100
Description
150 mg MgSO4, 50 mg PSA, 50 mg GCB
For Use With (Application)
QuEChERS Method
Price (USD)
Full specifications
TypeQuEChERS Clean-up
Description150 mg MgSO4, 50 mg PSA, 50 mg GCB
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Primary/Secondary Amine, Graphitized Carbon Black
Volume (Metric)2 mL Centrifuge Tubes
For Use With (Application)QuEChERS Method
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Description
150 mg MgSO4, 50 mg PSA
For Use With (Application)
QuEChERS Method
Price (USD)
Full specifications
TypeQuEChERS Clean-up
Description150 mg MgSO4, 50 mg PSA
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Primary/Secondary Amine
Volume (Metric)2 mL Centrifuge Tubes
For Use With (Application)QuEChERS Method
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Description
150 mg MgSO4, 50 mg PSA, 50 mg C18
For Use With (Application)
QuEChERS Method
Price (USD)
Full specifications
TypeQuEChERS Clean-up
Description150 mg MgSO4, 50 mg PSA, 50 mg C18
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Primary/Secondary Amine, Endcapped C18
Volume (Metric)2 mL Centrifuge Tubes
For Use With (Application)QuEChERS Method
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Description
900 mg MgSO4, 300 mg PSA, 150 mg GCB
For Use With (Application)
QuEChERS Method
Price (USD)
Full specifications
TypeQuEChERS Clean-up
Description900 mg MgSO4, 300 mg PSA, 150 mg GCB
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Primary/Secondary Amine, Graphitized Carbon Black
Volume (Metric)15 mL Centrifuge Tubes
For Use With (Application)QuEChERS Method
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 50
Description
900 mg MgSO4, 300 mg PSA, 150 mg C18
For Use With (Application)
QuEChERS Method
Price (USD)
Full specifications
TypeQuEChERS Clean-up
Description900 mg MgSO4, 300 mg PSA, 150 mg C18
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Primary/Secondary Amine, Endcapped C18
Volume (Metric)15 mL Centrifuge Tubes
For Use With (Application)QuEChERS Method
Unit SizePack of 50
Catalog Number
Unit Size
Pack of 30
Description
400 mg PSA on Bottom, 200 mg GCB on Top, Separated by a Frit
For Use With (Application)
Schenck Method
Price (USD)
Full specifications
TypeQuEChERS Clean-up
Description400 mg PSA on Bottom, 200 mg GCB on Top, Separated by a Frit
Base MaterialPrimary/Secondary Amine, Graphitized Carbon Black
Product LineHyperSep
SorbentPrimary/Secondary Amine, Graphitized Carbon Black
Volume (Metric)6 mL Centrifuge Tubes
For Use With (Application)Schenck Method
Unit SizePack of 30
Catalog Number
Unit Size
Pack of,30
Description
500 mg PSA on Bottom, 250 mg GCB on Top, Separated by a Frit
For Use With (Application)
Schenck Method
Price (USD)
Full specifications
TypeQuEChERS Clean-up
Description500 mg PSA on Bottom, 250 mg GCB on Top, Separated by a Frit
Base MaterialPrimary/Secondary Amine, Graphitized Carbon Black
Product LineHyperSep
SorbentPrimary/Secondary Amine, Graphitized Carbon Black
Volume (Metric)6 mL Centrifuge Tubes
For Use With (Application)Schenck Method
Unit SizePack of,30
Catalog Number
Unit Size
Pack of 30
Description
500 mg PSA on Bottom, 500 mg GCB on Top, Separated by a Frit
For Use With (Application)
Schenck Method
Price (USD)
Full specifications
TypeQuEChERS Clean-up
Description500 mg PSA on Bottom, 500 mg GCB on Top, Separated by a Frit
Base MaterialPrimary/Secondary Amine, Graphitized Carbon Black
Product LineHyperSep
SorbentPrimary/Secondary Amine, Graphitized Carbon Black
Volume (Metric)6 mL Centrifuge Tubes
For Use With (Application)Schenck Method
Unit SizePack of 30
Catalog Number
Unit Size
Pack of 50
Description
900 mg MgSO4, 150 mg PSA
For Use With (Application)
QuEChERS Method
Price (USD)
Full specifications
TypeQuEChERS Clean-up
Description900 mg MgSO4, 150 mg PSA
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Primary/Secondary Amine
Volume (Metric)15 mL Centrifuge Tubes
For Use With (Application)QuEChERS Method
Unit SizePack of 50
Showing 10 of 51Show more
Catalog NumberSpecificationsUnit SizeDescriptionFor Use With (Application)Price (USD)
60105-323Full specifications
Pack of,10150 mg MgSO4, 50 mg PSA, 50 mg C18, 50 mg GCBQuEChERS MethodRequest A Quote
TypeQuEChERS Clean-up
Description150 mg MgSO4, 50 mg PSA, 50 mg C18, 50 mg GCB
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Primary/Secondary Amine, Endcapped C18, Graphitized Carbon Black
Volume (Metric)2 mL Centrifuge Tubes
For Use With (Application)QuEChERS Method
Unit SizePack of,10
60105-202Full specifications
Pack of 100150 mg MgSO4, 50 mg PSA, 50 mg GCBQuEChERS MethodContact Us ›
TypeQuEChERS Clean-up
Description150 mg MgSO4, 50 mg PSA, 50 mg GCB
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Primary/Secondary Amine, Graphitized Carbon Black
Volume (Metric)2 mL Centrifuge Tubes
For Use With (Application)QuEChERS Method
Unit SizePack of 100
60105-203Full specifications
Pack of 100150 mg MgSO4, 50 mg PSAQuEChERS MethodContact Us ›
TypeQuEChERS Clean-up
Description150 mg MgSO4, 50 mg PSA
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Primary/Secondary Amine
Volume (Metric)2 mL Centrifuge Tubes
For Use With (Application)QuEChERS Method
Unit SizePack of 100
60105-204Full specifications
Pack of 100150 mg MgSO4, 50 mg PSA, 50 mg C18QuEChERS MethodContact Us ›
TypeQuEChERS Clean-up
Description150 mg MgSO4, 50 mg PSA, 50 mg C18
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Primary/Secondary Amine, Endcapped C18
Volume (Metric)2 mL Centrifuge Tubes
For Use With (Application)QuEChERS Method
Unit SizePack of 100
60105-205Full specifications
Pack of 100900 mg MgSO4, 300 mg PSA, 150 mg GCBQuEChERS MethodContact Us ›
TypeQuEChERS Clean-up
Description900 mg MgSO4, 300 mg PSA, 150 mg GCB
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Primary/Secondary Amine, Graphitized Carbon Black
Volume (Metric)15 mL Centrifuge Tubes
For Use With (Application)QuEChERS Method
Unit SizePack of 100
60105-206Full specifications
Pack of 50900 mg MgSO4, 300 mg PSA, 150 mg C18QuEChERS MethodContact Us ›
TypeQuEChERS Clean-up
Description900 mg MgSO4, 300 mg PSA, 150 mg C18
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Primary/Secondary Amine, Endcapped C18
Volume (Metric)15 mL Centrifuge Tubes
For Use With (Application)QuEChERS Method
Unit SizePack of 50
60105-207Full specifications
Pack of 30400 mg PSA on Bottom, 200 mg GCB on Top, Separated by a FritSchenck MethodContact Us ›
TypeQuEChERS Clean-up
Description400 mg PSA on Bottom, 200 mg GCB on Top, Separated by a Frit
Base MaterialPrimary/Secondary Amine, Graphitized Carbon Black
Product LineHyperSep
SorbentPrimary/Secondary Amine, Graphitized Carbon Black
Volume (Metric)6 mL Centrifuge Tubes
For Use With (Application)Schenck Method
Unit SizePack of 30
60105-208Full specifications
Pack of,30500 mg PSA on Bottom, 250 mg GCB on Top, Separated by a FritSchenck MethodRequest A Quote
TypeQuEChERS Clean-up
Description500 mg PSA on Bottom, 250 mg GCB on Top, Separated by a Frit
Base MaterialPrimary/Secondary Amine, Graphitized Carbon Black
Product LineHyperSep
SorbentPrimary/Secondary Amine, Graphitized Carbon Black
Volume (Metric)6 mL Centrifuge Tubes
For Use With (Application)Schenck Method
Unit SizePack of,30
60105-209Full specifications
Pack of 30500 mg PSA on Bottom, 500 mg GCB on Top, Separated by a FritSchenck MethodContact Us ›
TypeQuEChERS Clean-up
Description500 mg PSA on Bottom, 500 mg GCB on Top, Separated by a Frit
Base MaterialPrimary/Secondary Amine, Graphitized Carbon Black
Product LineHyperSep
SorbentPrimary/Secondary Amine, Graphitized Carbon Black
Volume (Metric)6 mL Centrifuge Tubes
For Use With (Application)Schenck Method
Unit SizePack of 30
60105-215Full specifications
Pack of 50900 mg MgSO4, 150 mg PSAQuEChERS MethodContact Us ›
TypeQuEChERS Clean-up
Description900 mg MgSO4, 150 mg PSA
Product LineHyperSep
SorbentAnhydrous Magnesium Sulfate, Primary/Secondary Amine
Volume (Metric)15 mL Centrifuge Tubes
For Use With (Application)QuEChERS Method
Unit SizePack of 50
Showing 10 of 51Show more
HyperSep Dispersive SPE Clean-Up Products contain PSA (primary/secondary amine) for the removal of organic acids and polar pigments among other compounds. Some products couple the PSA with endcapped C18 for the removal of most lipids and sterols, or graphitized carbon black for the removal of sterols and pigments such as chlorophyll. A variety of tube formats and bed weights are available to accommodate large and small sample sizes.
All HyperSep Dispersive SPE products are pre-prepared and optimized for the method at hand, so you will:
  • Save valuable preparation time
  • Achieve more consistent analytical results due to consistency in products and rigorous quality control
  • Get cleaner extracts and lower peak counts as a result of using highest quality, pre-prepared products
QuEChERS Technique
Developed by the USDA Eastern Regional Research Center in 2003, the QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) technique is increasingly becoming the technique of choice for extraction and clean-up of pesticide residues.
The QuEChERS method is a two-step process: extraction followed by clean-up. There are four variations of the QuEChERS method currently in use:
  • The original QuEChERS method uses sodium chloride to enhance extraction
  • Dispersive AOAC 2007.01 uses sodium acetate as a buffer, replacing sodium chloride
  • The Dual-Phase variation introduces PSA and GCB to remove high levels of chlorophyll and plant sterols in the final extract without the loss of planar pesticides (polar aromatics) using an acetone:toluene solvent mix (3:1)
  • European Version is similar to the AOAC method, except the extraction uses sodium chloride, sodium citrate dihydrate and disodium citrate sesquihydrate instead of sodium acetate
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.