Thermo Scientific™
HyperSep™ SpinTip Microscale SPE Extraction Tips, 1-10μL
Catalog number: 60109-410
Thermo Scientific™
HyperSep™ SpinTip Microscale SPE Extraction Tips, 1-10μL
Catalog number: 60109-410
Get faster sample preparation with minimal sample loss with the revolutionary Thermo Scientific™ HyperSep™ SpinTip Microscale SPE Tips and achieve separation in volumes as low as 100nL.
Catalog Number
60109-410
Unit Size
Pack of 96
Description
Strong Cation Exchanger; 1-10 uL Vol.
Price (USD)
Full specifications
DescriptionStrong Cation Exchanger; 1-10 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Particle Size40 to 60 μm
Product LineHyperSep
SorbentHyperSep Poros SCX
TypeSpinTip
Volume (Metric)1 to 10 µL
Unit SizePack of 96
Catalog Number
Unit Size
Pack of 96
Description
C18; 1-10 uL Vol.
Price (USD)
Full specifications
DescriptionC18; 1-10 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Particle Size40 to 60 μm
Product LineHyperSep
SorbentBioBasic C18
TypeSpinTip
Volume (Metric)1 to 10 µL
Unit SizePack of 96
Catalog Number
Unit Size
Pack of 96
Description
C8; 1-10 uL Vol.
Price (USD)
Full specifications
DescriptionC8; 1-10 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Particle Size40 to 60 μm
Product LineHyperSep
SorbentBioBasic C8
TypeSpinTip
Volume (Metric)1 to 10 µL
Unit SizePack of 96
Catalog Number
Unit Size
Pack of 96
Description
C4; 1-10 uL Vol.
Price (USD)
Full specifications
DescriptionC4; 1-10 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Product LineHyperSep
SorbentBioBasic C4
TypeSpinTip
Volume (Metric)1 to 10 µL
Unit SizePack of 96
Catalog Number
Unit Size
Pack of 96
Description
Hypercarb; 1-10 uL Vol.;
Price (USD)
Full specifications
DescriptionHypercarb; 1-10 uL Vol.;
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Particle Size30 μm
Product LineHyperSep
SorbentHyperSep Hypercarb
TypeSpinTip
Volume (Metric)1 to 10 µL
Unit SizePack of 96
Catalog Number
Unit Size
Pack of 96
Description
C18 plus Hypercarb; 1-10 uL Vol.
Price (USD)
Full specifications
DescriptionC18 plus Hypercarb; 1-10 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Product LineHyperSep
SorbentHyperSep Hypercarb and C18 (Mix Mode)
TypeSpinTip
Volume (Metric)1 to 10 µL
Unit SizePack of 96
Catalog Number
Unit Size
Each
Description
HILIC; 1-10 uL Vol.
Price (USD)
Full specifications
DescriptionHILIC; 1-10 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Product LineHyperSep
SorbentHyperSep HILIC
TypeSpinTip
Volume (Metric)1 to 10 µL
Unit SizeEach
Catalog Number
Unit Size
Pack of 96
Description
Trypsin; 1-10 uL Vol.
Price (USD)
Full specifications
DescriptionTrypsin; 1-10 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Product LineHyperSep
SorbentHyperSep Trypsin
TypeSpinTip
Volume (Metric)1 to 10 µL
Unit SizePack of 96
Catalog Number
Unit Size
Pack of 96
Description
Weak Anion Exchanger; 1-10 uL Vol.
Price (USD)
Full specifications
DescriptionWeak Anion Exchanger; 1-10 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Product LineHyperSep
SorbentHyperSep Poros WAX
TypeSpinTip
Volume (Metric)1 to 10 µL
Unit SizePack of 96
Catalog Number
Unit Size
Each
Description
Strong Anion Exchanger; 1-10 uL Vol.
Price (USD)
Full specifications
DescriptionStrong Anion Exchanger; 1-10 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Particle Size40 to 60 μm
Product LineHyperSep
SorbentHyperSep Poros SAX
TypeSpinTip
Volume (Metric)1 to 10 µL
Unit SizeEach
Showing 10 of 14Show more
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
60109-410Full specifications
Pack of 96Strong Cation Exchanger; 1-10 uL Vol.Contact Us ›
DescriptionStrong Cation Exchanger; 1-10 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Particle Size40 to 60 μm
Product LineHyperSep
SorbentHyperSep Poros SCX
TypeSpinTip
Volume (Metric)1 to 10 µL
Unit SizePack of 96
60109-401Full specifications
Pack of 96C18; 1-10 uL Vol.Contact Us ›
DescriptionC18; 1-10 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Particle Size40 to 60 μm
Product LineHyperSep
SorbentBioBasic C18
TypeSpinTip
Volume (Metric)1 to 10 µL
Unit SizePack of 96
60109-402Full specifications
Pack of 96C8; 1-10 uL Vol.Contact Us ›
DescriptionC8; 1-10 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Particle Size40 to 60 μm
Product LineHyperSep
SorbentBioBasic C8
TypeSpinTip
Volume (Metric)1 to 10 µL
Unit SizePack of 96
60109-403Full specifications
Pack of 96C4; 1-10 uL Vol.Contact Us ›
DescriptionC4; 1-10 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Product LineHyperSep
SorbentBioBasic C4
TypeSpinTip
Volume (Metric)1 to 10 µL
Unit SizePack of 96
60109-404Full specifications
Pack of 96Hypercarb; 1-10 uL Vol.;Contact Us ›
DescriptionHypercarb; 1-10 uL Vol.;
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Particle Size30 μm
Product LineHyperSep
SorbentHyperSep Hypercarb
TypeSpinTip
Volume (Metric)1 to 10 µL
Unit SizePack of 96
60109-405Full specifications
Pack of 96C18 plus Hypercarb; 1-10 uL Vol.Request A Quote
DescriptionC18 plus Hypercarb; 1-10 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Product LineHyperSep
SorbentHyperSep Hypercarb and C18 (Mix Mode)
TypeSpinTip
Volume (Metric)1 to 10 µL
Unit SizePack of 96
60109-406Full specifications
EachHILIC; 1-10 uL Vol.Request A Quote
DescriptionHILIC; 1-10 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Product LineHyperSep
SorbentHyperSep HILIC
TypeSpinTip
Volume (Metric)1 to 10 µL
Unit SizeEach
60109-407Full specifications
Pack of 96Trypsin; 1-10 uL Vol.Contact Us ›
DescriptionTrypsin; 1-10 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Product LineHyperSep
SorbentHyperSep Trypsin
TypeSpinTip
Volume (Metric)1 to 10 µL
Unit SizePack of 96
60109-408Full specifications
Pack of 96Weak Anion Exchanger; 1-10 uL Vol.Request A Quote
DescriptionWeak Anion Exchanger; 1-10 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Product LineHyperSep
SorbentHyperSep Poros WAX
TypeSpinTip
Volume (Metric)1 to 10 µL
Unit SizePack of 96
60109-409Full specifications
EachStrong Anion Exchanger; 1-10 uL Vol.Request A Quote
DescriptionStrong Anion Exchanger; 1-10 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Particle Size40 to 60 μm
Product LineHyperSep
SorbentHyperSep Poros SAX
TypeSpinTip
Volume (Metric)1 to 10 µL
Unit SizeEach
Showing 10 of 14Show more
Features:
Ideal for small sample volumes; features a 1 to 2μm wide slit at the bottom that permits liquid to pass through while retaining the chromatographic material (20 to 30μm).
  • Filter-free design to eliminate dead volume
  • No contamination from the supporting matrix
  • Available in many popular chemistries
Applications include:
  • Mass spectrometry
  • Desalting
  • Protein purification
  • MALDI
  • Electrophoresis
  • HPCE
  • HPLC
  • CEC
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.