Thermo Scientific™
HyperSep™ SpinTip Microscale SPE Extraction Tips, 10-200μL
Catalog number: 60109-420
Thermo Scientific™
HyperSep™ SpinTip Microscale SPE Extraction Tips, 10-200μL
Catalog number: 60109-420
Get faster sample preparation with minimal sample loss with the revolutionary Thermo Scientific™ HyperSep™ SpinTip Microscale SPE Tips and achieve separation in volumes as low as 100nL.
Catalog Number
60109-420
Unit Size
Pack of 96
Description
Strong Anion Exchanger; 10-200 uL Vol.
Price (USD)
Full specifications
DescriptionStrong Anion Exchanger; 10-200 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Particle Size40 to 60 μm
Product LineHyperSep
SorbentHyperSep Poros SAX
TypeSpinTip
Volume (Metric)10 to 200 µL
Unit SizePack of 96
Catalog Number
Unit Size
Pack of 96
Description
C18; 10-200 uL Vol.
Price (USD)
Full specifications
DescriptionC18; 10-200 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Particle Size40 to 60 μm
Product LineHyperSep
SorbentBioBasic C18
TypeSpinTip
Volume (Metric)10 to 200 µL
Unit SizePack of 96
Catalog Number
Unit Size
Pack of 96
Description
Zirconium Dioxide; 10-200 uL Vol.
Price (USD)
Full specifications
DescriptionZirconium Dioxide; 10-200 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Product LineHyperSep
SorbentHyperSep Zirconium Dioxide
TypeSpinTip
Volume (Metric)10 to 200 µL
Unit SizePack of 96
Catalog Number
Unit Size
Pack of 96
Description
Titanium Dioxide; 10-200 uL Vol.
Price (USD)
Full specifications
DescriptionTitanium Dioxide; 10-200 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Product LineHyperSep
SorbentHyperSep Titanium Dioxide
TypeSpinTip
Volume (Metric)10 to 200 µL
Unit SizePack of 96
Catalog Number
Unit Size
Pack of 96
Description
Strong Cation Exchanger; 10-200 uL Vol.
Price (USD)
Full specifications
DescriptionStrong Cation Exchanger; 10-200 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Particle Size40 to 60 μm
Product LineHyperSep
SorbentHyperSep Poros SCX
TypeSpinTip
Volume (Metric)10 to 200 µL
Unit SizePack of 96
Catalog Number
Unit Size
Pack of 96
Description
Weak Anion Exchanger; 10-200 uL Vol.
Price (USD)
Full specifications
DescriptionWeak Anion Exchanger; 10-200 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Product LineHyperSep
SorbentHyperSep Poros WAX
TypeSpinTip
Volume (Metric)10 to 200 µL
Unit SizePack of 96
Catalog Number
Unit Size
Pack of 96
Description
HyperSep SpinTip; 10-200 uL; Trypsin Material
Price (USD)
Full specifications
DescriptionHyperSep SpinTip; 10-200 uL; Trypsin Material
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Product LineHyperSep
SorbentHyperSep Trypsin
TypeSpinTip
Volume (Metric)10 to 200 µL
Unit SizePack of 96
Catalog Number
Unit Size
Pack of 96
Description
HILIC; 10-200 uL Vol.
Price (USD)
Full specifications
DescriptionHILIC; 10-200 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Product LineHyperSep
SorbentHyperSep HILIC
TypeSpinTip
Volume (Metric)10 to 200 µL
Unit SizePack of 96
Catalog Number
Unit Size
Pack of 96
Description
C18 plus Hypercarb; 10-200 uL Vol.
Price (USD)
Full specifications
DescriptionC18 plus Hypercarb; 10-200 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Product LineHyperSep
SorbentHyperSep Hypercarb and C18 (Mix Mode)
TypeSpinTip
Volume (Metric)10 to 200 µL
Unit SizePack of 96
Catalog Number
Unit Size
Pack of 96
Description
Hypercarb; 10-200 uL Vol.
Price (USD)
Full specifications
DescriptionHypercarb; 10-200 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Particle Size30 μm
Product LineHyperSep
SorbentHyperSep Hypercarb
TypeSpinTip
Volume (Metric)10 to 200 µL
Unit SizePack of 96
Showing 10 of 10
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
60109-420Full specifications
Pack of 96Strong Anion Exchanger; 10-200 uL Vol.Contact Us ›
DescriptionStrong Anion Exchanger; 10-200 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Particle Size40 to 60 μm
Product LineHyperSep
SorbentHyperSep Poros SAX
TypeSpinTip
Volume (Metric)10 to 200 µL
Unit SizePack of 96
60109-412Full specifications
Pack of 96C18; 10-200 uL Vol.Contact Us ›
DescriptionC18; 10-200 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Particle Size40 to 60 μm
Product LineHyperSep
SorbentBioBasic C18
TypeSpinTip
Volume (Metric)10 to 200 µL
Unit SizePack of 96
60109-425Full specifications
Pack of 96Zirconium Dioxide; 10-200 uL Vol.Contact Us ›
DescriptionZirconium Dioxide; 10-200 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Product LineHyperSep
SorbentHyperSep Zirconium Dioxide
TypeSpinTip
Volume (Metric)10 to 200 µL
Unit SizePack of 96
60109-422Full specifications
Pack of 96Titanium Dioxide; 10-200 uL Vol.Contact Us ›
DescriptionTitanium Dioxide; 10-200 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Product LineHyperSep
SorbentHyperSep Titanium Dioxide
TypeSpinTip
Volume (Metric)10 to 200 µL
Unit SizePack of 96
60109-421Full specifications
Pack of 96Strong Cation Exchanger; 10-200 uL Vol.Contact Us ›
DescriptionStrong Cation Exchanger; 10-200 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Particle Size40 to 60 μm
Product LineHyperSep
SorbentHyperSep Poros SCX
TypeSpinTip
Volume (Metric)10 to 200 µL
Unit SizePack of 96
60109-419Full specifications
Pack of 96Weak Anion Exchanger; 10-200 uL Vol.Contact Us ›
DescriptionWeak Anion Exchanger; 10-200 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Product LineHyperSep
SorbentHyperSep Poros WAX
TypeSpinTip
Volume (Metric)10 to 200 µL
Unit SizePack of 96
60109-418Full specifications
Pack of 96HyperSep SpinTip; 10-200 uL; Trypsin MaterialContact Us ›
DescriptionHyperSep SpinTip; 10-200 uL; Trypsin Material
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Product LineHyperSep
SorbentHyperSep Trypsin
TypeSpinTip
Volume (Metric)10 to 200 µL
Unit SizePack of 96
60109-417Full specifications
Pack of 96HILIC; 10-200 uL Vol.Contact Us ›
DescriptionHILIC; 10-200 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Product LineHyperSep
SorbentHyperSep HILIC
TypeSpinTip
Volume (Metric)10 to 200 µL
Unit SizePack of 96
60109-416Full specifications
Pack of 96C18 plus Hypercarb; 10-200 uL Vol.Contact Us ›
DescriptionC18 plus Hypercarb; 10-200 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Product LineHyperSep
SorbentHyperSep Hypercarb and C18 (Mix Mode)
TypeSpinTip
Volume (Metric)10 to 200 µL
Unit SizePack of 96
60109-415Full specifications
Pack of 96Hypercarb; 10-200 uL Vol.Contact Us ›
DescriptionHypercarb; 10-200 uL Vol.
For Use With (Application)Microscale Solid Phase Extraction
For Use With (Equipment)Micropipettes
Particle Size30 μm
Product LineHyperSep
SorbentHyperSep Hypercarb
TypeSpinTip
Volume (Metric)10 to 200 µL
Unit SizePack of 96
Showing 10 of 10
Features:
Ideal for small sample volumes; features a 1 to 2μm wide slit at the bottom that permits liquid to pass through while retaining the chromatographic material (20 to 30μm).
  • Filter-free design to eliminate dead volume
  • No contamination from the supporting matrix
  • Available in many popular chemistries
Applications include:
  • Mass spectrometry
  • Desalting
  • Protein purification
  • MALDI
  • Electrophoresis
  • HPCE
  • HPLC
  • CEC
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.