Thermo Scientific™
BioBasic™ 8 HPLC Columns
Catalog number: 72205-051030
Thermo Scientific™
BioBasic™ 8 HPLC Columns
Catalog number: 72205-051030
Optimized for the separation of a wide range of peptides, Thermo Scientific™ BioBasic™ 8 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules.
Catalog Number
72205-051030
Unit Size
-
Particle Size
5 μm
Diameter (Metric)
1 mm
Length (Metric)
50 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)1 mm
Length (Metric)50 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 9
Pore Size300 Å
Surface Area100 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load5%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
TypeAnalytical Column
Unit Size-
Catalog Number
Unit Size
-
Particle Size
5 μm
Diameter (Metric)
2.1 mm
Length (Metric)
50 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Length (Metric)50 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 9
Pore Size300 Å
Surface Area100 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load5%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
TypeAnalytical Column
Unit Size-
Catalog Number
Unit Size
-
Particle Size
5 μm
Diameter (Metric)
1 mm
Length (Metric)
100 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)1 mm
Length (Metric)100 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 9
Pore Size300 Å
Surface Area100 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load5%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
TypeAnalytical Column
Unit Size-
Catalog Number
Unit Size
-
Particle Size
5 μm
Diameter (Metric)
4.6 mm
Length (Metric)
100 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Length (Metric)100 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 9
Pore Size300 Å
Surface Area100 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load5%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
TypeAnalytical Column
Unit Size-
Catalog Number
Unit Size
-
Particle Size
5 μm
Diameter (Metric)
1 mm
Length (Metric)
150 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)1 mm
Length (Metric)150 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 9
Pore Size300 Å
Surface Area100 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load5%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
TypeAnalytical Column
Unit Size-
Catalog Number
Unit Size
-
Particle Size
5 μm
Diameter (Metric)
2.1 mm
Length (Metric)
150 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Length (Metric)150 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 9
Pore Size300 Å
Surface Area100 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load5%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
TypeAnalytical Column
Unit Size-
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
4.6 mm
Length (Metric)
150 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Length (Metric)150 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 9
Pore Size300 Å
Surface Area100 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load5%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
TypeAnalytical Column
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
4.6 mm
Length (Metric)
250 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Length (Metric)250 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 9
Pore Size300 Å
Surface Area100 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load5%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
TypeAnalytical Column
Unit SizeEach
Catalog Number
Unit Size
-
Particle Size
5 μm
Diameter (Metric)
2.1 mm
Length (Metric)
100 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Length (Metric)100 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 9
Pore Size300 Å
Surface Area100 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load5%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
TypeAnalytical Column
Unit Size-
Catalog Number
Unit Size
Pack of 4
Particle Size
5 μm
Diameter (Metric)
4 mm
Length (Metric)
10 mm
Price (USD)
Full specifications
Column FormatGuard Cartridge, 4 Pk.
Diameter (Metric)4 mm
For Use WithUniguard Direct-Connection and Stand Alone Holder
Length (Metric)10 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 9
Pore Size300 Å
Surface Area100 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load5%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
TypeGuard Cartridge, 4 Pk.
Unit SizePack of 4
Showing 10 of 12Show more
Catalog NumberSpecificationsUnit SizeParticle SizeDiameter (Metric)Length (Metric)Price (USD)
72205-051030Full specifications
-5 μm1 mm50 mmRequest A Quote
Column FormatAnalytical Column
Diameter (Metric)1 mm
Length (Metric)50 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 9
Pore Size300 Å
Surface Area100 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load5%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
TypeAnalytical Column
Unit Size-
72205-052130Full specifications
-5 μm2.1 mm50 mmRequest A Quote
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Length (Metric)50 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 9
Pore Size300 Å
Surface Area100 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load5%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
TypeAnalytical Column
Unit Size-
72205-101030Full specifications
-5 μm1 mm100 mmRequest A Quote
Column FormatAnalytical Column
Diameter (Metric)1 mm
Length (Metric)100 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 9
Pore Size300 Å
Surface Area100 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load5%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
TypeAnalytical Column
Unit Size-
72205-104630Full specifications
-5 μm4.6 mm100 mmRequest A Quote
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Length (Metric)100 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 9
Pore Size300 Å
Surface Area100 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load5%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
TypeAnalytical Column
Unit Size-
72205-151030Full specifications
-5 μm1 mm150 mmRequest A Quote
Column FormatAnalytical Column
Diameter (Metric)1 mm
Length (Metric)150 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 9
Pore Size300 Å
Surface Area100 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load5%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
TypeAnalytical Column
Unit Size-
72205-152130Full specifications
-5 μm2.1 mm150 mmRequest A Quote
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Length (Metric)150 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 9
Pore Size300 Å
Surface Area100 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load5%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
TypeAnalytical Column
Unit Size-
72205-154630Full specifications
Each 5 μm4.6 mm150 mmContact Us ›
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Length (Metric)150 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 9
Pore Size300 Å
Surface Area100 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load5%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
TypeAnalytical Column
Unit SizeEach
72205-254630Full specifications
Each 5 μm4.6 mm250 mmContact Us ›
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Length (Metric)250 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 9
Pore Size300 Å
Surface Area100 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load5%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
TypeAnalytical Column
Unit SizeEach
72205-102130Full specifications
-5 μm2.1 mm100 mmRequest A Quote
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Length (Metric)100 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 9
Pore Size300 Å
Surface Area100 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load5%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
TypeAnalytical Column
Unit Size-
72205-014001Full specifications
Pack of 45 μm4 mm10 mmContact Us ›
Column FormatGuard Cartridge, 4 Pk.
Diameter (Metric)4 mm
For Use WithUniguard Direct-Connection and Stand Alone Holder
Length (Metric)10 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 9
Pore Size300 Å
Surface Area100 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load5%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
TypeGuard Cartridge, 4 Pk.
Unit SizePack of 4
Showing 10 of 12Show more
  • 300Å pore size for improved biomolecule analysis
  • An excellent starting column for protein and peptide analysis
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations
Ordering Information:
Other column dimensions are available, including preparative columns. Please call Customer Service for more information.