Thermo Scientific™

BioBasic™ SEC 120 Size Exclusion Chromatography HPLC Columns

Catalog number: 73405-307846
Thermo Scientific™

BioBasic™ SEC 120 Size Exclusion Chromatography HPLC Columns

Catalog number: 73405-307846

Provide outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 120 Size Exclusion Chromatography HPLC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 100 to 10,000,000 daltons, allowing the analysis of proteins, peptides and other biomolecules.

 
Catalog Number
73405-307846
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
7.8 mm
Length (Metric)
300 mm
Price (USD)
Full specifications
Column FormatAnalytical Column
Diameter (Metric)7.8 mm
For Use WithSize Exclusion Chromatography
Length (Metric)300 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
Pore Size120 Å
USP TypeL33
Stationary PhaseDiol (OH)
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeSize Exclusion
PhaseSize Exclusion
TypeAnalytical Column
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
7.8 mm
Length (Metric)
150 mm
Price (USD)
Full specifications
Column FormatHigh Temperature Column
Diameter (Metric)7.8 mm
For Use WithSize Exclusion Chromatography
Length (Metric)150 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
Pore Size120 Å
USP TypeL33
Stationary PhaseDiol (OH)
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeSize Exclusion
PhaseSize Exclusion
TypeHigh Temperature Column
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
7.8 mm
Length (Metric)
30 mm
Price (USD)
Full specifications
Column FormatGuard Column
Diameter (Metric)7.8 mm
For Use WithSize Exclusion Chromatography
Length (Metric)30 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
Pore Size120 Å
USP TypeL33
Stationary PhaseDiol (OH)
Product LineBioBasic
Packing MaterialSilica
Column TypeSize Exclusion
PhaseSize Exclusion
TypeGuard Column
Unit SizeEach
Showing 3 of 3
Catalog NumberSpecificationsUnit SizeParticle SizeDiameter (Metric)Length (Metric)Price (USD)
73405-307846Full specifications
Each5 μm7.8 mm300 mmContact Us ›
Column FormatAnalytical Column
Diameter (Metric)7.8 mm
For Use WithSize Exclusion Chromatography
Length (Metric)300 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
Pore Size120 Å
USP TypeL33
Stationary PhaseDiol (OH)
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeSize Exclusion
PhaseSize Exclusion
TypeAnalytical Column
Unit SizeEach
73405-157846Full specifications
Each5 μm7.8 mm150 mmContact Us ›
Column FormatHigh Temperature Column
Diameter (Metric)7.8 mm
For Use WithSize Exclusion Chromatography
Length (Metric)150 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
Pore Size120 Å
USP TypeL33
Stationary PhaseDiol (OH)
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeSize Exclusion
PhaseSize Exclusion
TypeHigh Temperature Column
Unit SizeEach
73405-037821Full specifications
Each5 μm7.8 mm30 mmRequest A Quote
Column FormatGuard Column
Diameter (Metric)7.8 mm
For Use WithSize Exclusion Chromatography
Length (Metric)30 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
Pore Size120 Å
USP TypeL33
Stationary PhaseDiol (OH)
Product LineBioBasic
Packing MaterialSilica
Column TypeSize Exclusion
PhaseSize Exclusion
TypeGuard Column
Unit SizeEach
Showing 3 of 3
  • Covers separation of analytes over a wide molecular weight range
  • Long column life and high column efficiencies
  • Simple mechanism of interaction based on molecular size and shape
  • Excellent for sample cleanup
  • Straightforward method development, simple mobile phases
  • 120Å pore size

Ordering Information:

For information on bulk quantities, please inquire.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.