Thermo Scientific™

BioBasic™ PREP SEC 300 Size Exclusion Chromatography HPLC Columns

Catalog number: 73505-037821
Thermo Scientific™

BioBasic™ PREP SEC 300 Size Exclusion Chromatography HPLC Columns

Catalog number: 73505-037821

Provide outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 300 Size Exclusion Chromatography preparative HPLC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 7,000 to 1,200,000 daltons, allowing for the analysis of proteins, peptides and other biomolecules.

 
Catalog Number
73505-037821
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
7.8 mm
Length (Metric)
30 mm
Column Format
Guard Column
Price (USD)
Full specifications
Column FormatGuard Column
Diameter (Metric)7.8 mm
Length (Metric)30 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
Pore Size300 Å
USP TypeL59
Stationary PhaseDiol (OH)
Product LineBioBasic
Packing MaterialSilica
Column TypeSize Exclusion
PhaseSize Exclusion
TypeGuard Column
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
7.8 mm
Length (Metric)
150 mm
Column Format
High Temperature Column
Price (USD)
Full specifications
Column FormatHigh Temperature Column
Diameter (Metric)7.8 mm
Length (Metric)150 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
Pore Size300 Å
USP TypeL59
Stationary PhaseDiol (OH)
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeSize Exclusion
PhaseSize Exclusion
TypeHigh Temperature Column
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
7.8 mm
Length (Metric)
300 mm
Column Format
High Temperature Column
Price (USD)
Full specifications
Column FormatHigh Temperature Column
Diameter (Metric)7.8 mm
Length (Metric)300 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
Pore Size300 Å
USP TypeL59
Stationary PhaseDiol (OH)
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeSize Exclusion
PhaseSize Exclusion
TypeHigh Temperature Column
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
20 mm
Length (Metric)
50 mm
Column Format
Preparative Column, Standard
Price (USD)
Full specifications
Column FormatPreparative Column, Standard
Diameter (Metric)20 mm
Length (Metric)50 mm
Max. Pressure4300 psi (295 bar)
Particle Size5 μm
pH2 to 8
Pore Size300 Å
Temperature60°C
USP TypeL59
Stationary PhaseDiol (OH)
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeSize Exclusion
EndcappedNo
PhaseSize Exclusion
Unit SizeEach
Showing 4 of 4
Catalog NumberSpecificationsUnit SizeParticle SizeDiameter (Metric)Length (Metric)Column FormatPrice (USD)
73505-037821Full specifications
Each5 μm7.8 mm30 mmGuard ColumnContact Us ›
Column FormatGuard Column
Diameter (Metric)7.8 mm
Length (Metric)30 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
Pore Size300 Å
USP TypeL59
Stationary PhaseDiol (OH)
Product LineBioBasic
Packing MaterialSilica
Column TypeSize Exclusion
PhaseSize Exclusion
TypeGuard Column
Unit SizeEach
73505-157846Full specifications
Each 5 μm7.8 mm150 mmHigh Temperature ColumnContact Us ›
Column FormatHigh Temperature Column
Diameter (Metric)7.8 mm
Length (Metric)150 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
Pore Size300 Å
USP TypeL59
Stationary PhaseDiol (OH)
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeSize Exclusion
PhaseSize Exclusion
TypeHigh Temperature Column
Unit SizeEach
73505-307846Full specifications
Each5 μm7.8 mm300 mmHigh Temperature ColumnContact Us ›
Column FormatHigh Temperature Column
Diameter (Metric)7.8 mm
Length (Metric)300 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
Pore Size300 Å
USP TypeL59
Stationary PhaseDiol (OH)
Product LineBioBasic
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeSize Exclusion
PhaseSize Exclusion
TypeHigh Temperature Column
Unit SizeEach
73505-059270AFull specifications
Each5 μm20 mm50 mmPreparative Column, StandardContact Us ›
Column FormatPreparative Column, Standard
Diameter (Metric)20 mm
Length (Metric)50 mm
Max. Pressure4300 psi (295 bar)
Particle Size5 μm
pH2 to 8
Pore Size300 Å
Temperature60°C
USP TypeL59
Stationary PhaseDiol (OH)
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeSize Exclusion
EndcappedNo
PhaseSize Exclusion
Unit SizeEach
Showing 4 of 4

BioBasic™ PREP SEC 300 HPLC Column Benefits:

  • Covers separation of analytes over a wide molecular weight range
  • Long column life and high column efficiencies
  • Simple mechanism of interaction based on molecular size and shape
  • Excellent for sample cleanup
  • Straightforward method development, simple mobile phases
  • 300Å pore size

Ordering Information:

For information on bulk quantities, please inquire.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.