Informational:Regulatory requirements vary by country; products may not be available in your geographic area.
Applied Biosystems™

PrioCHECK™ FMD IPC 3ABC Kit

Catalog number: 7610740
Applied Biosystems™

PrioCHECK™ FMD IPC 3ABC Kit

Catalog number: 7610740
Catalog Number
7610740
Unit Size
60-90 samples
Price (USD)
Catalog NumberUnit SizePrice (USD)
761074060-90 samplesContact Us ›
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
The PrioCHECK® FMD IPC 3ABC kit is a fast, highly sensitive, and cost-effective tool to determine FMDV vaccine purity. The PrioCHECK® FMD IPC 3ABC kit is an in-process control (IPC) filtration-assisted chemiluminometric immunoassay, to detect and quantify 3ABC protein in vaccine-antigen batches regardless of their volume and composition. In-process control testing contributes significantly to saving production costs of vaccines.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Specifications

Immunoassay Kit Grade
Filter
Target Organism Class
Foot and Mouth Disease Virus (FMDV)
Target Species Validated
All Sensitive Animals
Product Line
PrioCHECK™, Prionics™
Product Type
ELISA Kit
Protein Family
Other Proteins
Gene ID (Entrez)
Non-Gene
Target Species
Livestock
Assay Sensitivity
See product documentation
Gene Symbol
Non-Gene
Target Kit Named
FMD IPC 3ABC
Immunoassay Kit Format
Various Other

Contents & Storage

1 filter plate