Thermo Scientific™

MSIA™ D.A.R.T.'S™ for the Versette™ and Novus i Platform

Catalog number: 991PRT11
Thermo Scientific™

MSIA™ D.A.R.T.'S™ for the Versette™ and Novus i Platform

Catalog number: 991PRT11
Improve hybrid MS bio-analysis with Thermo Scientific™ MSIA™ D.A.R.T.'S™ for the Versette™ and Novus i Platform , an exclusive Thermo Fisher Scientific workflow solution for proteins and peptides. Enabled through proprietary MSIA D.A.R.T.'S™ technologies, these workflows have demonstrated pg/mL level detection while providing simultaneous qualitative/quantitative data content from complex biological matrix, such as human plasma. Ideal for biologic and biomarker bio-analysis, MSIA provides innovative enablement in the areas of PKPD, DMPK, ADME, Biotransformation (DAR and payload determination), as well as routine biomarker analysis.
 
Catalog Number
991PRT11
Unit Size
Each
Description
MSIA D.A.R.T.'S, Protein A
Format
Pack of 96 Tips
Price (USD)
Full specifications
DescriptionMSIA D.A.R.T.'S, Protein A
Volume (Metric)300 µL
Storage Requirements2° to 8°C
FormatPack of 96 Tips
Unit SizeEach
Catalog Number
Unit Size
Each
Description
MSIA D.A.R.T.'S, Custom
Format
Pack of 96 Tips
Price (USD)
Full specifications
DescriptionMSIA D.A.R.T.'S, Custom
Volume (Metric)300 µL
Storage Requirements2° to 8°C
FormatPack of 96 Tips
Unit SizeEach
Catalog Number
Unit Size
Each
Description
MSIA D.A.R.T.'S, Protein A
Format
Pack of 24 Tips
Price (USD)
Full specifications
DescriptionMSIA D.A.R.T.'S, Protein A
Volume (Metric)300 µL
Storage Requirements2° to 8°C
FormatPack of 24 Tips
Unit SizeEach
Catalog Number
Unit Size
Each
Description
MSIA D.A.R.T.'S, Protein G
Format
Pack of 96 Tips
Price (USD)
Full specifications
DescriptionMSIA D.A.R.T.'S, Protein G
Volume (Metric)300 µL
Storage Requirements2° to 8°C
FormatPack of 96 Tips
Unit SizeEach
Catalog Number
Unit Size
Each
Description
MSIA D.A.R.T.'S, Protein G
Format
Pack of 24 Tips
Price (USD)
Full specifications
DescriptionMSIA D.A.R.T.'S, Protein G
Volume (Metric)300 µL
Storage Requirements2° to 8°C
FormatPack of 24 Tips
Unit SizeEach
Catalog Number
Unit Size
Case of 96
Description
MSIA D.A.R.T.'S, Protein A/G
Format
Pack of 96 Tips
Price (USD)
Full specifications
DescriptionMSIA D.A.R.T.'S, Protein A/G
Volume (Metric)300 µL
Storage Requirements2° to 8°C
FormatPack of 96 Tips
Unit SizeCase of 96
Catalog Number
Unit Size
Each
Description
MSIA D.A.R.T.'S, Protein A/G
Format
Pack of 24 Tips
Price (USD)
Full specifications
DescriptionMSIA D.A.R.T.'S, Protein A/G
Volume (Metric)300 µL
Storage Requirements2° to 8°C
FormatPack of 24 Tips
Unit SizeEach
Catalog Number
Unit Size
Case of 96
Description
MSIA D.A.R.T.'S, Streptavidin
Format
Pack of 96 Tips
Price (USD)
Full specifications
DescriptionMSIA D.A.R.T.'S, Streptavidin
Volume (Metric)300 µL
FormatPack of 96 Tips
Unit SizeCase of 96
Catalog Number
Unit Size
Each
Description
MSIA D.A.R.T.'S, Streptavidin
Format
Pack of 24 Tips
Price (USD)
Full specifications
DescriptionMSIA D.A.R.T.'S, Streptavidin
Volume (Metric)300 µL
FormatPack of 24 Tips
Unit SizeEach
Catalog Number
Unit Size
Each
Description
MSIA D.A.R.T.'S, Insulin
Format
Pack of 96 Tips
Price (USD)
Full specifications
DescriptionMSIA D.A.R.T.'S, Insulin
Volume (Metric)300 µL
FormatPack of 96 Tips
Unit SizeEach
Showing 10 of 12Show more
Catalog NumberSpecificationsUnit SizeDescriptionFormatPrice (USD)
991PRT11Full specifications
EachMSIA D.A.R.T.'S, Protein APack of 96 TipsRequest A Quote
DescriptionMSIA D.A.R.T.'S, Protein A
Volume (Metric)300 µL
Storage Requirements2° to 8°C
FormatPack of 96 Tips
Unit SizeEach
991CUS02Full specifications
EachMSIA D.A.R.T.'S, CustomPack of 96 TipsRequest A Quote
DescriptionMSIA D.A.R.T.'S, Custom
Volume (Metric)300 µL
Storage Requirements2° to 8°C
FormatPack of 96 Tips
Unit SizeEach
991PRT12Full specifications
EachMSIA D.A.R.T.'S, Protein APack of 24 TipsRequest A Quote
DescriptionMSIA D.A.R.T.'S, Protein A
Volume (Metric)300 µL
Storage Requirements2° to 8°C
FormatPack of 24 Tips
Unit SizeEach
991PRT13Full specifications
EachMSIA D.A.R.T.'S, Protein GPack of 96 TipsRequest A Quote
DescriptionMSIA D.A.R.T.'S, Protein G
Volume (Metric)300 µL
Storage Requirements2° to 8°C
FormatPack of 96 Tips
Unit SizeEach
991PRT14Full specifications
EachMSIA D.A.R.T.'S, Protein GPack of 24 TipsRequest A Quote
DescriptionMSIA D.A.R.T.'S, Protein G
Volume (Metric)300 µL
Storage Requirements2° to 8°C
FormatPack of 24 Tips
Unit SizeEach
991PRT15Full specifications
Case of 96MSIA D.A.R.T.'S, Protein A/GPack of 96 TipsRequest A Quote
DescriptionMSIA D.A.R.T.'S, Protein A/G
Volume (Metric)300 µL
Storage Requirements2° to 8°C
FormatPack of 96 Tips
Unit SizeCase of 96
991PRT16Full specifications
EachMSIA D.A.R.T.'S, Protein A/GPack of 24 TipsRequest A Quote
DescriptionMSIA D.A.R.T.'S, Protein A/G
Volume (Metric)300 µL
Storage Requirements2° to 8°C
FormatPack of 24 Tips
Unit SizeEach
991STR11Full specifications
Case of 96MSIA D.A.R.T.'S, StreptavidinPack of 96 TipsRequest A Quote
DescriptionMSIA D.A.R.T.'S, Streptavidin
Volume (Metric)300 µL
FormatPack of 96 Tips
Unit SizeCase of 96
991STR12Full specifications
EachMSIA D.A.R.T.'S, StreptavidinPack of 24 TipsRequest A Quote
DescriptionMSIA D.A.R.T.'S, Streptavidin
Volume (Metric)300 µL
FormatPack of 24 Tips
Unit SizeEach
991001096Full specifications
EachMSIA D.A.R.T.'S, InsulinPack of 96 TipsRequest A Quote
DescriptionMSIA D.A.R.T.'S, Insulin
Volume (Metric)300 µL
FormatPack of 96 Tips
Unit SizeEach
Showing 10 of 12Show more
Next generation hybrid Mass Spectrometry bio-analytical workflows:
  • Enables analysis of proteins of interest down to femtomole level
  • Streamlined workflow for protein qualitative & quantative bio-analysis
  • Employs a proprietary molecular trapping micro-column technology in an easy to use pipette tip housing format
  • Drives antibody/antigen binding kinetics and greatly improves assaying of low-abundance protein from a biological sample
Enabling workflows and technologies for biologic and biomarker research:
  • Immunoaffinity column captures protein of interest with improved signal-to-noise ratio
  • Superior specificity and assaying characteristics compared to conventional approaches
  • Allows for hybrid assaying down to femtomole levels
  • Streptavidin and Protein A, G & A/G MSIA D.A.R.T.'S provide end user flexibility to analyze their specific targets of interest
  • Custom MSIA D.A.R.T.'S are available upon request
  • Optimized workflow solution Insulin (endogenous and therapeutic forms)

Requires:

300μL pipette such as Thermo Scientific™ Versette™ Liquid Handling Platform, Thermo Scientific™ Finnpipette™ Novus i Multichannel pipetters (listed in user manual)

Recommended for:

Hybrid mass spectrometry based assays

For research use only