Invitrogen™

BacMam LRRK2-GFP Wild Type Reagent

Catalog number: A14170
Invitrogen™

BacMam LRRK2-GFP Wild Type Reagent

Catalog number: A14170
Catalog Number
A14170
Unit Size
15 ml
Price (USD)
Price: 2,260.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A1417015 ml
Price: 2,260.00
Your Price:
-
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
BacMam LRRK2-GFP Wild Type Reagent is a modified insect cell virus (baculovirus) that enables easy, efficient, and titratable overexpression of full-length wild type LRRK2-GFP in your choice of cellular background, including primary neurons and neuroblastoma cell lines.

BacMam LRRK2-GFP Wild Type Reagent Lets You:

• Overexpress full-length LRRK2-GFP in most cellular backgrounds
• Visualize LRRK2 expression
• Conduct the LRRK2 cellular assays you need
• Understand the effects of therapeutically relevant LRRK2 mutations

Use BacMam Technology with GFP for Expression and Visualization of LRRK2
GFP (Green Fluorescent Protein) allows you to visually confirm LRRK2-GFP protein expression and compare localization of wild type with pathological mutants in various cellular backgrounds, especially primary neurons, while BacMam technology utilizes a modified baculovirus to deliver and express genes in mammalian cells with minimal effort and toxicity. BacMam enables efficient, titratable LRRK2-GFP protein expression in various mammalian cellular backgrounds including cell lines that may not be compatible with traditional transfection methods, such as primary neurons and neuroblastoma cell lines.

Expand Your Battery of Cellular Assays for LRRK2
Once full-length LRRK2-GFP has been introduced into your cellular background, the transduced cells allow you the flexibility to conduct many types of LRRK2 experiments, including studies of regulation of physiologically functional pathways (including signaling pathways leading to function) and identification of potential physiological substrates for LRRK2.

Examine LRRK2 Mutations with BacMam Reagents
Use therapeutic mutant reagents G2019S (Cat No A14174) along with the wild type LRRK2-GFP reagent to enable your research into LRRK2 mutations, including exploration of differences in cellular response to stimulus among different mutations.

For research use only. Not intended for human or animal therapeutic or diagnostic use.

Related Links:
LRRK2 tools for advancing Parkinson's disease research
Kinase protein portfolio
LanthaScreen® Eu Kinase Binding Assay
LanthaScreen® Activity Assay
Learn More About BacMam Technology

Specifications

Delivery Type
Baculovirus (BacMam System)
Promoter
None (Promoterless)
Quantity
15 mL
Shipping Condition
Wet Ice

Contents & Storage

Recombinant baculovirus particles suspended in Gibco™ Sf-900™ III SFM media (Cat# 12658) supplemented with 3% heat-inactivated fetal bovine serum (hiFBS) (Cat# 10100).

Store at 4°C, DO NOT FREEZE, KEEP STERILE, protect from light, aliquot upon arrival.