Thermo Scientific™

LanthaScreen™ Eu-anti-GFP Antibody

Catalog number: A14173
Thermo Scientific™

LanthaScreen™ Eu-anti-GFP Antibody

Catalog number: A14173
Catalog Number
A14173
Unit Size
2.5 µg
Price (USD)
Price: 477.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
25 µg
Price (USD)
Price: 968.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A141732.5 µg
Price: 477.00
Your Price:
-
A1419025 µg
Price: 968.00
Your Price:
-
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
These Europium-labeled anti-GFP antibodies bind specifically to GFP-tagged proteins allowing precise interrogation of protein samples. These antibodies can be used with our Kinase Tracers to conduct LanthaScreen™ Eu Kinase Binding Assays.

LanthaScreen™ Eu-anti-GFP Antibody Lets You:

• Indirectly label proteins tagged with GFP
• Measure the right kinase
• Interrogate active and non-activated kinases

Eu-anti-GFP Gives You the Benefits of Indirect Labeling
LanthaScreen™ Europium-labeled anti-GFP antibodies can be used for indirect labeling of proteins tagged with GFP. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein negatively impacts protein activity.

Use Eu-anti-GFP Antibody in the Highly Specific LanthaScreen™ Eu Kinase Binding Assay
When LanthaScreen™ Eu-anti-GFP Antibody is used in the LanthaScreen™ Eu Kinase Binding Assay, any signal is the result of a specific interaction of the Eu-anti-GFP antibody with the kinase of interest. Other kinases or phosphatases present without tag or with a different tag produce no signal. Unlike activity assays, the kinase binding assay can be performed with either active or non-active kinase preparations, making it ideal for detection of Type II inhibitors.

For research use only. Not intended for human or animal therapeutic or diagnostic use.

Specifications

Antibody Conjugate
Europium (Eu)
High-throughput Compatibility
High-throughput Compatible
Label or Dye
Europium
Label Type
Lanthanide Chelates
Mass
2.5 µg
Shipping Condition
Dry Ice
Target Molecule
GFP
Product Line
LanthaScreen™

Contents & Storage

Europium labeled anti-GFP antibody in HEPES buffered saline containing 0.1% Triton X-100.

Store at -20°C