Invitrogen™

ERN1 Recombinant Human Protein

Catalog number: A32889
Invitrogen™

ERN1 Recombinant Human Protein

Catalog number: A32889
Catalog Number
A32889
Unit Size
100 µg
Price (USD)
Price: 2,255.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
10 µg
Price (USD)
Price: 478.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
1 mg
Price (USD)
Price: 6,670.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A32889100 µg
Price: 2,255.00
Your Price:
-
A3097110 µg
Price: 478.00
Your Price:
-
A333741 mg
Price: 6,670.00
Your Price:
-
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
This ERN1 recombinant human protein (468–end) was expressed in insect cells. ERN1 serine/threonine kinase or endoplasmic reticulum to nucleus signaling 1 is the ER to nucleus signaling 1 protein, a human homologue of the yeast Ire1 gene product which possesses intrinsic kinase activity and an endoribonuclease activity and it is important in endoplasmic reticulum-based stress signals. ERN1 controls IRE1 proteolysis in mammalian cells. ERN1 has a critical function in extraembryonic cells that is essential for fetal viability. ERN1 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service.

• Validated for use with the LanthaScreen™ Eu Kinase Binding assay
• Available in multiple pack sizes for convenient use
• Also available in our SelectScreen Kinase Profiling Service

Specifications

Accession Number
NP_001424
Molecular Weight
84.1 kDa
Quantity
100 µg
KinaseFamily
IRE Family
Protein Length
aa 468 - end
Recombinant
Yes
Protein Tag
GST Tag
Shipping Condition
Dry Ice
Expression System
Insect
Protein Form
Recombinant, Active, Catalytic
Form
Liquid
Validated Application
Kinase Assay
Species
Human

Contents & Storage

Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.