Invitrogen™

MASTL Recombinant Human Protein

Catalog number: A32896
Invitrogen™

MASTL Recombinant Human Protein

Catalog number: A32896
Catalog Number
A32896
Unit Size
100 µg
Price (USD)
Price: 2,255.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
10 µg
Price (USD)
Price: 483.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
1 mg
Price (USD)
Price: 7,010.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A32896100 µg
Price: 2,255.00
Your Price:
-
A3097810 µg
Price: 483.00
Your Price:
-
A333811 mg
Price: 7,010.00
Your Price:
-
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
This MASTL recombinant human protein (full length) was expressed in insect cells. MASTL or microtubule associated serine/threonine kinase-like belongs to the MAST family, AGC Ser/Thr protein kinase group. MASTL promotes mitotic progression and cell cycle reentry after DNA damage. Loss of MASTL leads to defects in chromosome condensation, separation, and other aspects of mitotic progression, which is primarily mediated by inhibiting PP2A/B55 delta. Overexpression of MASTL is correlated with cancer progression, poor patient survival, and tumor recurrence in various cancers. MASTL recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service.

• Validated for use with the LanthaScreen™ Eu Kinase Binding assay
• Available in multiple pack sizes for convenient use
• Also available in our SelectScreen Kinase Profiling Service

Specifications

Accession Number
NP_001165774
Molecular Weight
123.6 kDa
Quantity
100 µg
KinaseFamily
MAST Family
Protein Length
Full Length
Recombinant
Yes
Protein Tag
GST Tag
Shipping Condition
Dry Ice
Expression System
Insect
Protein Form
Full Length, Recombinant
Form
Liquid
Validated Application
Kinase Assay
Species
Human

Contents & Storage

Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.