Thermo Scientific™
9mm Clear Glass Screw Thread Vials
Catalog number: C4000-LV3W
Thermo Scientific™
9mm Clear Glass Screw Thread Vials
Catalog number: C4000-LV3W
Nearly eliminate septa push-through and improve sealing. Thermo Scientific™ 9 mm Clear Glass Screw Thread Vials are 12 x 32 mm and built with Advanced Vial Closure System (AVCS). This innovation not only prevents septa push through, but provides a stop point for the cap to prevent over- or under-tightening and provide an optimal, consistent seal every time. Vials are made from superior quality 33 expansion borosilicate clear (Type 1, Class A) glass, and meet all US, EU and JPN Pharmacopia standards. Choose from Thermo Scientific™ Premiere™, Thermo Scientific™ National™ and Thermo Scientific™ Chromacol™ brands.
Catalog Number
C4000-LV3W
Unit Size
Pack of 100
Base Style
Insert Vial
Volume (Metric)
375 μL
Marking Spot
Yes
Price (USD)
Full specifications
Description9mm Target DP MacroVial 200µL, Fused Insert
Diameter (Metric) Opening9 mm
Diameter (Metric) Inner12 mm
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)375 μL
Insert Type200μL Fused Insert
Volume (Metric) Residual<1 μL
Base StyleInsert Vial
Volume (Metric) Total375μL
Vial Type9 mm Target DP Vial
Closure TypeScrew
Height (Metric)32 mm
Quantity100
Closure Size9 mm
Marking SpotYes
Volume (Metric) Usable240μL
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Base Style
Insert Vial
Volume (Metric)
0.3 mL
Marking Spot
Yes
Price (USD)
Full specifications
Description9 mm clear glass screw thread micro+ vial with write-on patch, fused insert, 0.3 mL volume
Diameter (Metric) Opening9 mm
Diameter (Metric) Inner12 mm
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)0.3 mL
Insert Type300μL Fused Insert
Volume (Metric) Residual<3μL
Base StyleInsert Vial
Volume (Metric) Total0.3 mL
Vial Type9 mm Screw Thread Vial
Closure TypeScrew
Height (Metric)32 mm
Closure Size9 mm
Marking SpotYes
Volume (Metric) Usable250μL
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Base Style
Flat Bottom
Volume (Metric)
2.0 mL
Marking Spot
Yes
Price (USD)
Full specifications
Description9 mm clear glass wide mouth screw thread vial with write-on ID patch, 2 ml volume
Diameter (Metric) Opening9 mm
Diameter (Metric) Inner12 mm
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)2.0 mL
Volume (Metric) Residual<170μL
Base StyleFlat Bottom
Volume (Metric) Total2 mL
Vial Type9 mm Screw Thread Vial
Closure TypeScrew
Height (Metric)32 mm
Closure Size9mm
Marking SpotYes
Volume (Metric) Usable1.5 mL
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 125
Base Style
Flat Bottom
Volume (Metric)
4.0 mL
Marking Spot
No
Price (USD)
Full specifications
Description4mL screw top vial, clear, uses 9mm screw caps (not included)
Diameter (Metric) Opening9 mm
Diameter (Metric) Inner15 mm
ColorClear
Diameter (Metric) Outer15 mm
MaterialGlass
Volume (Metric)4.0 mL
Volume (Metric) Residual<500 μL
Base StyleFlat Bottom
Volume (Metric) Total4 mL
Vial Type9 mm Screw Thread Vial
Closure TypeScrew
Height (Metric)46 mm
Quantity125
Closure Size9 mm
Marking SpotNo
Volume (Metric) Usable3.5 mL
Unit SizePack of 125
Catalog Number
Unit Size
Case of 1000
Base Style
Flat Bottom
Volume (Metric)
2.0 mL
Marking Spot
No
Price (USD)
Full specifications
Description9mm Screw Thread Vial
Diameter (Metric) Opening9 mm
Diameter (Metric) Inner12 mm
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)2.0 mL
Volume (Metric) Residual<170 μL
Base StyleFlat Bottom
Volume (Metric) Total2 mL
Vial Type9 mm Screw Thread Vial
Closure TypeScrew
Height (Metric)32 mm
Quantity1000
Closure Size9 mm
Marking SpotNo
Volume (Metric) Usable1.5 mL
For Use With (Equipment)Thermo Scientific, Agilent, PE, Shimadzu, Varian
Unit SizeCase of 1000
Catalog Number
Unit Size
Pack of 100
Base Style
Flat Bottom
Volume (Metric)
2.0 mL
Marking Spot
No
Price (USD)
Full specifications
DescriptionClear Glass
Diameter (Metric) Opening9 mm
Diameter (Metric) Inner12 mm
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)2.0 mL
Volume (Metric) Residual<170 μL
Base StyleFlat Bottom
Product LineSurestop
Volume (Metric) Total2 mL
Vial Type9 mm SureStop Vial (AVCS)
Closure TypeScrew
Height (Metric)32 mm
Quantity100
Closure Size9 mm
Marking SpotNo
Volume (Metric) Usable1.5 mL
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Base Style
Flat Bottom
Volume (Metric)
2.0 mL
Marking Spot
Yes
Price (USD)
Full specifications
DescriptionClear Glass w/ID Patch
Diameter (Metric) Opening9 mm
Diameter (Metric) Inner12 mm
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)2.0 mL
Volume (Metric) Residual<170 μL
Base StyleFlat Bottom
Product LineSurestop
Volume (Metric) Total2 mL
Vial Type9 mm SureStop Vial (AVCS)
Closure TypeScrew
Height (Metric)32 mm
Quantity100
Closure Size9 mm
Marking SpotYes
Volume (Metric) Usable1.5 mL
Unit SizePack of 100
Catalog Number
Unit Size
Pack of,100
Base Style
Flat Bottom
Volume (Metric)
2.0 mL
Marking Spot
Blue
Price (USD)
Full specifications
Description2mL; Clear Glass, With Blue Color Band
Diameter (Metric) Opening9 mm
Diameter (Metric) Inner12 mm
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)2.0 mL
Volume (Metric) Residual<170 μL
Volume (Metric) Working1.8 mL
Base StyleFlat Bottom
Volume (Metric) Total2 mL
Vial Type9 mm Target DP ColorBand Vial
Closure TypeScrew
Height (Metric)32 mm
Quantity100
Closure Size9 mm
Marking SpotBlue
Volume (Metric) Usable1.5 mL
Unit SizePack of,100
Catalog Number
Unit Size
Pack of 100
Base Style
Flat Bottom
Volume (Metric)
2.0 mL
Marking Spot
Green
Price (USD)
Full specifications
Description2mL; Clear Glass, With Green Color Band
Diameter (Metric) Opening9 mm
Diameter (Metric) Inner12 mm
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)2.0 mL
Volume (Metric) Residual<170 μL
Volume (Metric) Working1.8 mL
Base StyleFlat Bottom
Volume (Metric) Total2 mL
Vial Type9 mm Target DP ColorBand Vial
Closure TypeScrew
Height (Metric)32 mm
Quantity100
Closure Size9 mm
Marking SpotGreen
Volume (Metric) Usable1.5 mL
Unit SizePack of 100
Catalog Number
Unit Size
Each
Base Style
Flat Bottom
Volume (Metric)
2.0 mL
Marking Spot
Red
Price (USD)
Full specifications
Description2mL; Clear Glass, With Red Color Band
Diameter (Metric) Opening9 mm
Diameter (Metric) Inner12 mm
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)2.0 mL
Volume (Metric) Residual<170 μL
Volume (Metric) Working1.8 mL
Base StyleFlat Bottom
Volume (Metric) Total2 mL
Vial Type9 mm Target DP ColorBand Vial
Closure TypeScrew
Height (Metric)32 mm
Quantity100
Closure Size9 mm
Marking SpotRed
Volume (Metric) Usable1.5 mL
Unit SizeEach
Showing 10 of 26Show more
Catalog NumberSpecificationsUnit SizeBase StyleVolume (Metric)Marking SpotPrice (USD)
C4000-LV3WFull specifications
Pack of 100Insert Vial375 μLYesContact Us ›
Description9mm Target DP MacroVial 200µL, Fused Insert
Diameter (Metric) Opening9 mm
Diameter (Metric) Inner12 mm
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)375 μL
Insert Type200μL Fused Insert
Volume (Metric) Residual<1 μL
Base StyleInsert Vial
Volume (Metric) Total375μL
Vial Type9 mm Target DP Vial
Closure TypeScrew
Height (Metric)32 mm
Quantity100
Closure Size9 mm
Marking SpotYes
Volume (Metric) Usable240μL
Unit SizePack of 100
60180-507Full specifications
Pack of 100Insert Vial0.3 mLYesContact Us ›
Description9 mm clear glass screw thread micro+ vial with write-on patch, fused insert, 0.3 mL volume
Diameter (Metric) Opening9 mm
Diameter (Metric) Inner12 mm
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)0.3 mL
Insert Type300μL Fused Insert
Volume (Metric) Residual<3μL
Base StyleInsert Vial
Volume (Metric) Total0.3 mL
Vial Type9 mm Screw Thread Vial
Closure TypeScrew
Height (Metric)32 mm
Closure Size9 mm
Marking SpotYes
Volume (Metric) Usable250μL
Unit SizePack of 100
60180-509Full specifications
Pack of 100Flat Bottom2.0 mLYesContact Us ›
Description9 mm clear glass wide mouth screw thread vial with write-on ID patch, 2 ml volume
Diameter (Metric) Opening9 mm
Diameter (Metric) Inner12 mm
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)2.0 mL
Volume (Metric) Residual<170μL
Base StyleFlat Bottom
Volume (Metric) Total2 mL
Vial Type9 mm Screw Thread Vial
Closure TypeScrew
Height (Metric)32 mm
Closure Size9mm
Marking SpotYes
Volume (Metric) Usable1.5 mL
Unit SizePack of 100
60180-510Full specifications
Pack of 125Flat Bottom4.0 mLNoContact Us ›
Description4mL screw top vial, clear, uses 9mm screw caps (not included)
Diameter (Metric) Opening9 mm
Diameter (Metric) Inner15 mm
ColorClear
Diameter (Metric) Outer15 mm
MaterialGlass
Volume (Metric)4.0 mL
Volume (Metric) Residual<500 μL
Base StyleFlat Bottom
Volume (Metric) Total4 mL
Vial Type9 mm Screw Thread Vial
Closure TypeScrew
Height (Metric)46 mm
Quantity125
Closure Size9 mm
Marking SpotNo
Volume (Metric) Usable3.5 mL
Unit SizePack of 125
60180-723Full specifications
Case of 1000Flat Bottom2.0 mLNoContact Us ›
Description9mm Screw Thread Vial
Diameter (Metric) Opening9 mm
Diameter (Metric) Inner12 mm
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)2.0 mL
Volume (Metric) Residual<170 μL
Base StyleFlat Bottom
Volume (Metric) Total2 mL
Vial Type9 mm Screw Thread Vial
Closure TypeScrew
Height (Metric)32 mm
Quantity1000
Closure Size9 mm
Marking SpotNo
Volume (Metric) Usable1.5 mL
For Use With (Equipment)Thermo Scientific, Agilent, PE, Shimadzu, Varian
Unit SizeCase of 1000
C5000-1Full specifications
Pack of 100Flat Bottom2.0 mLNoContact Us ›
DescriptionClear Glass
Diameter (Metric) Opening9 mm
Diameter (Metric) Inner12 mm
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)2.0 mL
Volume (Metric) Residual<170 μL
Base StyleFlat Bottom
Product LineSurestop
Volume (Metric) Total2 mL
Vial Type9 mm SureStop Vial (AVCS)
Closure TypeScrew
Height (Metric)32 mm
Quantity100
Closure Size9 mm
Marking SpotNo
Volume (Metric) Usable1.5 mL
Unit SizePack of 100
C5000-1WFull specifications
Pack of 100Flat Bottom2.0 mLYesContact Us ›
DescriptionClear Glass w/ID Patch
Diameter (Metric) Opening9 mm
Diameter (Metric) Inner12 mm
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)2.0 mL
Volume (Metric) Residual<170 μL
Base StyleFlat Bottom
Product LineSurestop
Volume (Metric) Total2 mL
Vial Type9 mm SureStop Vial (AVCS)
Closure TypeScrew
Height (Metric)32 mm
Quantity100
Closure Size9 mm
Marking SpotYes
Volume (Metric) Usable1.5 mL
Unit SizePack of 100
C4000-1BFull specifications
Pack of,100Flat Bottom2.0 mLBlueContact Us ›
Description2mL; Clear Glass, With Blue Color Band
Diameter (Metric) Opening9 mm
Diameter (Metric) Inner12 mm
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)2.0 mL
Volume (Metric) Residual<170 μL
Volume (Metric) Working1.8 mL
Base StyleFlat Bottom
Volume (Metric) Total2 mL
Vial Type9 mm Target DP ColorBand Vial
Closure TypeScrew
Height (Metric)32 mm
Quantity100
Closure Size9 mm
Marking SpotBlue
Volume (Metric) Usable1.5 mL
Unit SizePack of,100
C4000-1GFull specifications
Pack of 100Flat Bottom2.0 mLGreenContact Us ›
Description2mL; Clear Glass, With Green Color Band
Diameter (Metric) Opening9 mm
Diameter (Metric) Inner12 mm
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)2.0 mL
Volume (Metric) Residual<170 μL
Volume (Metric) Working1.8 mL
Base StyleFlat Bottom
Volume (Metric) Total2 mL
Vial Type9 mm Target DP ColorBand Vial
Closure TypeScrew
Height (Metric)32 mm
Quantity100
Closure Size9 mm
Marking SpotGreen
Volume (Metric) Usable1.5 mL
Unit SizePack of 100
C4000-1RFull specifications
EachFlat Bottom2.0 mLRedRequest A Quote
Description2mL; Clear Glass, With Red Color Band
Diameter (Metric) Opening9 mm
Diameter (Metric) Inner12 mm
ColorClear
Diameter (Metric) Outer12 mm
MaterialGlass
Volume (Metric)2.0 mL
Volume (Metric) Residual<170 μL
Volume (Metric) Working1.8 mL
Base StyleFlat Bottom
Volume (Metric) Total2 mL
Vial Type9 mm Target DP ColorBand Vial
Closure TypeScrew
Height (Metric)32 mm
Quantity100
Closure Size9 mm
Marking SpotRed
Volume (Metric) Usable1.5 mL
Unit SizeEach
Showing 10 of 26Show more
  • 9 mm wide opening vials
  • Easy-on, easy-off convenience with just one turn
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.