Thermo Scientific™
8mm Amber Glass Crimp Top Vials
Catalog number: C4008-742
Thermo Scientific™
8mm Amber Glass Crimp Top Vials
Catalog number: C4008-742
These low-volume sample vials are ideal for 6mm, 7mm and 8mm autosampler trays. Choose from flat or conical base to maximize recovery needed for small-sample injections. Re-usable sleeves support the crimped, sealed vials in the correct position in autosampler carousels or in racks and allow movement of the vials as a unit to injection positions in both GC and HPLC autosamplers. Made of superior quality 51A amber (Type 1, Class B), these vials meet all requirements of US, EU and JPN Pharmacopia. Choose from Thermo ScientificTM PremiereTM, NationalTM, ChromacolTM brands.
Catalog Number
C4008-742
Unit Size
Pack of 100
Closure Size
8 mm
Volume (Metric)
775 μL
Price (USD)
Full specifications
DescriptionAmber Glass Vial, 7 x 40mm, Flat Bottom
Diameter (Metric) Opening8 mm
Diameter (Metric) Inner7 mm
ColorAmber
Diameter (Metric) Outer7 mm
MaterialGlass
Volume (Metric)775 μL
Volume (Metric) Residual<70 μL
Base StyleFlat Base
Volume (Metric) Total775μL
Vial Type8 mm Crimp Top Vial
Closure TypeCrimp
Height (Metric)40 mm
Quantity100
Closure Size8 mm
Marking SpotNo
Volume (Metric) Usable650μL
For Use With (Equipment)Beckman™ 504, CTC/Leap™ Technologies HTS PAL, Thermo Scientific™ AS 200S, Gilson™ 231
Unit SizePack of 100
Catalog Number
02-CTVA
also known as 02-CTV(A)
Unit Size
Each
Closure Size
8 mm
Volume (Metric)
250 μL
Price (USD)
Full specifications
Description0.2mL Sci-Vi Crimp Top Vial
Diameter (Metric) Opening8 mm
Diameter (Metric) Inner6 mm
ColorAmber
Diameter (Metric) Outer6 mm
MaterialGlass
Volume (Metric)250 μL
Volume (Metric) Residual<5 μL
Base StyleConical
Volume (Metric) Total250μL
Vial Type8 mm Crimp Top Vial
Closure TypeCrimp
Height (Metric)32 mm
Quantity500
Closure Size8 mm
Marking SpotNo
Volume (Metric) Usable200μL
Unit SizeEach
Catalog Number
05-CTVA
also known as 05-CTV(A)
Unit Size
Each
Closure Size
8 mm
Volume (Metric)
500 μL
Price (USD)
Full specifications
Description0.5mL Crimp Top Tapered Vial
Diameter (Metric) Opening8 mm
Diameter (Metric) Inner7 mm
ColorAmber
Diameter (Metric) Outer7 mm
MaterialGlass
Volume (Metric)500 μL
Volume (Metric) Residual<5 μL
Base StyleConical
Volume (Metric) Total500μL
Vial Type8 mm Crimp Top Vial
Closure TypeCrimp
Height (Metric)30 mm
Quantity500
Closure Size8 mm
Marking SpotNo
Volume (Metric) Usable450μL
Unit SizeEach
Catalog Number
06-CTVA
also known as 06-CTV(A)
Unit Size
Pack of 500
Closure Size
8 mm
Volume (Metric)
600 μL
Price (USD)
Full specifications
Description0.6mL Crimp Top Tapered Vial
Diameter (Metric) Opening8 mm
Diameter (Metric) Inner7 mm
ColorAmber
Diameter (Metric) Outer7 mm
MaterialGlass
Volume (Metric)600 μL
Volume (Metric) Residual<5 μL
Base StyleConical
Volume (Metric) Total600μL
Vial Type8 mm Crimp Top Vial
Closure TypeCrimp
Height (Metric)32 mm
Quantity500
Closure Size8 mm
Marking SpotNo
Volume (Metric) Usable550μL
Unit SizePack of 500
Catalog Number
07-CPVA
also known as 07-CPV(A)
Unit Size
Pack of,100
Closure Size
8 mm
Volume (Metric)
575 μL
Price (USD)
Full specifications
Description0.7mL Crimp Top Tapered Vial
Diameter (Metric) Opening8 mm
Diameter (Metric) Inner7 mm
ColorAmber
Diameter (Metric) Outer7 mm
MaterialGlass
Volume (Metric)575 μL
Volume (Metric) Residual<5 μL
Base StyleConical
Volume (Metric) Total575μL
Vial Type8 mm Crimp Top Vial
Closure TypeCrimp
Height (Metric)40 mm
Quantity500
Closure Size8 mm
Marking SpotNo
Volume (Metric) Usable450μL
Unit SizePack of,100
Catalog Number
08-CRVA
also known as 08-CRV(A)
Unit Size
Each
Closure Size
8 mm
Volume (Metric)
700 μL
Price (USD)
Full specifications
Description0.8mL Crimp Top Vial
Diameter (Metric) Opening8 mm
Diameter (Metric) Inner7 mm
ColorAmber
Diameter (Metric) Outer7 mm
MaterialGlass
Volume (Metric)700 μL
Volume (Metric) Residual<30 μL
Base StyleRound Bottom
Volume (Metric) Total700μL
Vial Type8 mm Crimp Top Vial
Closure TypeCrimp
Height (Metric)32 mm
Quantity500
Closure Size8 mm
Marking SpotNo
Volume (Metric) Usable600μL
Unit SizeEach
Catalog Number
Unit Size
Pack of 100
Closure Size
8 mm
Volume (Metric)
575 μL
Price (USD)
Full specifications
DescriptionAmber Glass Vial, 7 x 40mm, Conical Bottom
Diameter (Metric) Opening8 mm
Diameter (Metric) Inner7 mm
ColorAmber
Diameter (Metric) Outer7 mm
MaterialGlass
Volume (Metric)575 μL
Volume (Metric) Residual<2 μL
Base StyleConical Base
Volume (Metric) Total575μL
Vial Type8 mm Crimp Top Vial
Closure TypeCrimp
Height (Metric)40 mm
Quantity100
Closure Size8 mm
Marking SpotNo
Volume (Metric) Usable450μL
For Use With (Equipment)Beckman™ 504, CTC/Leap™ Technologies HTS PAL, Thermo Scientific™ AS 200S, Gilson™ 231
Unit SizePack of 100
Showing 7 of 7
Catalog NumberSpecificationsUnit SizeClosure SizeVolume (Metric)Price (USD)
C4008-742Full specifications
Pack of 1008 mm775 μLContact Us ›
DescriptionAmber Glass Vial, 7 x 40mm, Flat Bottom
Diameter (Metric) Opening8 mm
Diameter (Metric) Inner7 mm
ColorAmber
Diameter (Metric) Outer7 mm
MaterialGlass
Volume (Metric)775 μL
Volume (Metric) Residual<70 μL
Base StyleFlat Base
Volume (Metric) Total775μL
Vial Type8 mm Crimp Top Vial
Closure TypeCrimp
Height (Metric)40 mm
Quantity100
Closure Size8 mm
Marking SpotNo
Volume (Metric) Usable650μL
For Use With (Equipment)Beckman™ 504, CTC/Leap™ Technologies HTS PAL, Thermo Scientific™ AS 200S, Gilson™ 231
Unit SizePack of 100
02-CTVA
also known as 02-CTV(A)
Full specifications
Each8 mm250 μLRequest A Quote
Description0.2mL Sci-Vi Crimp Top Vial
Diameter (Metric) Opening8 mm
Diameter (Metric) Inner6 mm
ColorAmber
Diameter (Metric) Outer6 mm
MaterialGlass
Volume (Metric)250 μL
Volume (Metric) Residual<5 μL
Base StyleConical
Volume (Metric) Total250μL
Vial Type8 mm Crimp Top Vial
Closure TypeCrimp
Height (Metric)32 mm
Quantity500
Closure Size8 mm
Marking SpotNo
Volume (Metric) Usable200μL
Unit SizeEach
05-CTVA
also known as 05-CTV(A)
Full specifications
Each8 mm500 μLRequest A Quote
Description0.5mL Crimp Top Tapered Vial
Diameter (Metric) Opening8 mm
Diameter (Metric) Inner7 mm
ColorAmber
Diameter (Metric) Outer7 mm
MaterialGlass
Volume (Metric)500 μL
Volume (Metric) Residual<5 μL
Base StyleConical
Volume (Metric) Total500μL
Vial Type8 mm Crimp Top Vial
Closure TypeCrimp
Height (Metric)30 mm
Quantity500
Closure Size8 mm
Marking SpotNo
Volume (Metric) Usable450μL
Unit SizeEach
06-CTVA
also known as 06-CTV(A)
Full specifications
Pack of 5008 mm600 μLContact Us ›
Description0.6mL Crimp Top Tapered Vial
Diameter (Metric) Opening8 mm
Diameter (Metric) Inner7 mm
ColorAmber
Diameter (Metric) Outer7 mm
MaterialGlass
Volume (Metric)600 μL
Volume (Metric) Residual<5 μL
Base StyleConical
Volume (Metric) Total600μL
Vial Type8 mm Crimp Top Vial
Closure TypeCrimp
Height (Metric)32 mm
Quantity500
Closure Size8 mm
Marking SpotNo
Volume (Metric) Usable550μL
Unit SizePack of 500
07-CPVA
also known as 07-CPV(A)
Full specifications
Pack of,1008 mm575 μLContact Us ›
Description0.7mL Crimp Top Tapered Vial
Diameter (Metric) Opening8 mm
Diameter (Metric) Inner7 mm
ColorAmber
Diameter (Metric) Outer7 mm
MaterialGlass
Volume (Metric)575 μL
Volume (Metric) Residual<5 μL
Base StyleConical
Volume (Metric) Total575μL
Vial Type8 mm Crimp Top Vial
Closure TypeCrimp
Height (Metric)40 mm
Quantity500
Closure Size8 mm
Marking SpotNo
Volume (Metric) Usable450μL
Unit SizePack of,100
08-CRVA
also known as 08-CRV(A)
Full specifications
Each8 mm700 μLRequest A Quote
Description0.8mL Crimp Top Vial
Diameter (Metric) Opening8 mm
Diameter (Metric) Inner7 mm
ColorAmber
Diameter (Metric) Outer7 mm
MaterialGlass
Volume (Metric)700 μL
Volume (Metric) Residual<30 μL
Base StyleRound Bottom
Volume (Metric) Total700μL
Vial Type8 mm Crimp Top Vial
Closure TypeCrimp
Height (Metric)32 mm
Quantity500
Closure Size8 mm
Marking SpotNo
Volume (Metric) Usable600μL
Unit SizeEach
C4008-740Full specifications
Pack of 1008 mm575 μLContact Us ›
DescriptionAmber Glass Vial, 7 x 40mm, Conical Bottom
Diameter (Metric) Opening8 mm
Diameter (Metric) Inner7 mm
ColorAmber
Diameter (Metric) Outer7 mm
MaterialGlass
Volume (Metric)575 μL
Volume (Metric) Residual<2 μL
Base StyleConical Base
Volume (Metric) Total575μL
Vial Type8 mm Crimp Top Vial
Closure TypeCrimp
Height (Metric)40 mm
Quantity100
Closure Size8 mm
Marking SpotNo
Volume (Metric) Usable450μL
For Use With (Equipment)Beckman™ 504, CTC/Leap™ Technologies HTS PAL, Thermo Scientific™ AS 200S, Gilson™ 231
Unit SizePack of 100
Showing 7 of 7
Choose the right vial for your needs
8mm crimp top seal minimizes exposure between sample solvent and septum
Precision-machined (glass or PTFE) sleeves allow the vials to be used in the vast majority of commercial autosamplers
Autosampler compatibility*
Thermo Scientific™
Agilent™
Beckman™
Carlo Erba™
CTC™
Fisons™
PerkinElmer™
Shimadzu™
VWR™ (Merck)/Hitachi™
*See the Thermo Scientific Columns and Consumables Catalog for autosampler compatibility
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.