Thermo Scientific™

9mm Autosampler Inserts

Catalog number: C4010-K630
Thermo Scientific™

9mm Autosampler Inserts

Catalog number: C4010-K630
Clear glass or polypropylene inserts for 9mm HPLC and GC autosampler vials are available in a variety of configurations with usable volumes from 160μL to 3.5mL.
 
Catalog Number
C4010-K630
Unit Size
Pack of 100
Volume (Metric)
375 μL
Material
Glass
Insert Type
Conical
Features
Precision-formed tip, Kimshield deactivated
Price (USD)
Full specifications
Description300µL Polyspring Insert, Kimshield
Diameter (Metric) Inner6 mm
ColorClear
Diameter (Metric) Outer6 mm
MaterialGlass
Volume (Metric)375 μL
FeaturesPrecision-formed tip, Kimshield deactivated
Insert TypeConical
Volume (Metric) Residual<1 μL
Product LineNational WebSeal
Volume (Metric) Total375 µL
Vial TypeInsert
Height (Metric)29 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler
Volume (Metric) Usable300 μL
For Use With (Equipment)Target DP 9 mm Wide Opening Screw Thread Vials
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Volume (Metric)
400 μL
Material
Glass
Insert Type
Pulled Point
Features
Precision-formed tip, rests on vial bottom
Price (USD)
Full specifications
Description350µL Insert
Diameter (Metric) Inner6 mm
ColorClear
Diameter (Metric) Outer6 mm
MaterialGlass
Volume (Metric)400 μL
FeaturesPrecision-formed tip, rests on vial bottom
Insert TypePulled Point
Volume (Metric) Residual<4 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total400 µL
Vial TypeInsert
Height (Metric)31 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler
Volume (Metric) Usable350 μL
For Use With (Equipment)National 11 mm Crimp Top Vials
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Volume (Metric)
375 μL
Material
Glass
Insert Type
Conical
Features
Preferred for viscous solutions
Price (USD)
Full specifications
Description300µL Insert
Diameter (Metric) Inner6 mm
ColorClear
Diameter (Metric) Outer6 mm
MaterialGlass
Volume (Metric)375 μL
FeaturesPreferred for viscous solutions
Insert TypeConical
Volume (Metric) Residual<1 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total375 µL
Vial TypeInsert
Height (Metric)29 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler
Volume (Metric) Usable300 μL
For Use With (Equipment)National 11 mm Crimp Top Vials
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Volume (Metric)
400 μL
Material
Glass
Insert Type
Precision Point
Features
Precision-formed tip, rests on vial bottom
Price (USD)
Full specifications
Description350µL Insert
Diameter (Metric) Inner6 mm
ColorClear
Diameter (Metric) Outer6 mm
MaterialGlass
Volume (Metric)400 μL
FeaturesPrecision-formed tip, rests on vial bottom
Insert TypePrecision Point
Volume (Metric) Residual<2 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total400 µL
Vial TypeInsert
Height (Metric)31 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler
Volume (Metric) Usable350 μL
For Use With (Equipment)National 11 mm Crimp Top Vials
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Volume (Metric)
325 μL
Material
Polypropylene
Insert Type
Conical, Graduated
Features
Conical Base; Graduation Marks
Price (USD)
Full specifications
Description300µL Insert, Graduation Marks
Diameter (Metric) Inner6 mm
ColorClear
Diameter (Metric) Outer6 mm
MaterialPolypropylene
Volume (Metric)325 μL
FeaturesConical Base; Graduation Marks
Insert TypeConical, Graduated
Volume (Metric) Residual<2 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total325 µL
Vial TypeInsert
Height (Metric)30 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler
Closure Size200 μL
Volume (Metric) Usable250 μL
For Use With (Equipment)8 mm HPLC, GC Autosampler Vial
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Volume (Metric)
375 μL
Material
Glass
Insert Type
Conical
Features
Polyspring centers and cushions insert
Price (USD)
Full specifications
Description300µL Polyspring Insert
Diameter (Metric) Inner6 mm
ColorClear
Diameter (Metric) Outer6 mm
MaterialGlass
Volume (Metric)375 μL
FeaturesPolyspring centers and cushions insert
Insert TypeConical
Volume (Metric) Residual<1 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total375 µL
Vial TypeInsert
Height (Metric)29 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler
Volume (Metric) Usable300 μL
For Use With (Equipment)National 11 mm Crimp Top Vials
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 500
Volume (Metric)
500 μL
Material
Glass
Insert Type
Flat Bottom
Features
Insert rests on bottom of vial
Price (USD)
Full specifications
Description400µL MicroSert Insert
Diameter (Metric) Inner6 mm
ColorClear
Diameter (Metric) Outer6 mm
MaterialGlass
Volume (Metric)500 μL
FeaturesInsert rests on bottom of vial
Insert TypeFlat Bottom
Volume (Metric) Residual<25 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total500 μL
Vial TypeInsert
Height (Metric)31 mm
Quantity500
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler
Volume (Metric) Usable400 μL
For Use With (Equipment)National 11 mm Crimp Top Vials
Unit SizePack of 500
Catalog Number
Unit Size
Pack of 500
Volume (Metric)
500 μL
Material
Polypropylene
Insert Type
Features
Molded plastic insert
Price (USD)
Full specifications
Description400µL MicroSert Insert
Diameter (Metric) Inner6 mm
ColorClear
Diameter (Metric) Outer6 mm
MaterialPolypropylene
Volume (Metric)500 μL
FeaturesMolded plastic insert
Volume (Metric) Residual<25 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total500 µL
Vial TypeInsert
Height (Metric)31 mm
Quantity500
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler
Volume (Metric) Usable400 μL
For Use With (Equipment)Target DP Vials
Unit SizePack of 500
Catalog Number
Unit Size
Pack of 100
Volume (Metric)
375 μL
Material
Glass
Insert Type
Conical, Silanized
Features
Insert rests on bottom of vial
Price (USD)
Full specifications
Description300µL Insert, silanized
Diameter (Metric) Inner6 mm
ColorClear
Diameter (Metric) Outer6 mm
MaterialGlass
Volume (Metric)375 μL
FeaturesInsert rests on bottom of vial
Insert TypeConical, Silanized
Volume (Metric) Residual<1 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total375 µL
Vial TypeInsert
Height (Metric)29 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler
Volume (Metric) Usable300 μL
For Use With (Equipment)National 11 mm Crimp Top Vials
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Volume (Metric)
375 μL
Material
Glass
Insert Type
Conical, Silanized
Features
Precision-formed tip, NSC silanized
Price (USD)
Full specifications
Description300µL Polyspring Insert, silanized
Diameter (Metric) Inner6 mm
ColorClear
Diameter (Metric) Outer6 mm
MaterialGlass
Volume (Metric)375 μL
FeaturesPrecision-formed tip, NSC silanized
Insert TypeConical, Silanized
Volume (Metric) Residual<1 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total375 µL
Vial TypeInsert
Height (Metric)29 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler
Volume (Metric) Usable300 μL
For Use With (Equipment)National 11 mm Crimp Top Vials
Unit SizePack of 100
Showing 10 of 12Show more
Catalog NumberSpecificationsUnit SizeVolume (Metric)MaterialInsert TypeFeaturesPrice (USD)
C4010-K630Full specifications
Pack of 100375 μLGlassConicalPrecision-formed tip, Kimshield deactivatedContact Us ›
Description300µL Polyspring Insert, Kimshield
Diameter (Metric) Inner6 mm
ColorClear
Diameter (Metric) Outer6 mm
MaterialGlass
Volume (Metric)375 μL
FeaturesPrecision-formed tip, Kimshield deactivated
Insert TypeConical
Volume (Metric) Residual<1 μL
Product LineNational WebSeal
Volume (Metric) Total375 µL
Vial TypeInsert
Height (Metric)29 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler
Volume (Metric) Usable300 μL
For Use With (Equipment)Target DP 9 mm Wide Opening Screw Thread Vials
Unit SizePack of 100
C4010-627LFull specifications
Pack of 100400 μLGlassPulled PointPrecision-formed tip, rests on vial bottomContact Us ›
Description350µL Insert
Diameter (Metric) Inner6 mm
ColorClear
Diameter (Metric) Outer6 mm
MaterialGlass
Volume (Metric)400 μL
FeaturesPrecision-formed tip, rests on vial bottom
Insert TypePulled Point
Volume (Metric) Residual<4 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total400 µL
Vial TypeInsert
Height (Metric)31 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler
Volume (Metric) Usable350 μL
For Use With (Equipment)National 11 mm Crimp Top Vials
Unit SizePack of 100
C4010-629Full specifications
Pack of 100375 μLGlassConicalPreferred for viscous solutionsContact Us ›
Description300µL Insert
Diameter (Metric) Inner6 mm
ColorClear
Diameter (Metric) Outer6 mm
MaterialGlass
Volume (Metric)375 μL
FeaturesPreferred for viscous solutions
Insert TypeConical
Volume (Metric) Residual<1 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total375 µL
Vial TypeInsert
Height (Metric)29 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler
Volume (Metric) Usable300 μL
For Use With (Equipment)National 11 mm Crimp Top Vials
Unit SizePack of 100
C4010-629LFull specifications
Pack of 100400 μLGlassPrecision PointPrecision-formed tip, rests on vial bottomContact Us ›
Description350µL Insert
Diameter (Metric) Inner6 mm
ColorClear
Diameter (Metric) Outer6 mm
MaterialGlass
Volume (Metric)400 μL
FeaturesPrecision-formed tip, rests on vial bottom
Insert TypePrecision Point
Volume (Metric) Residual<2 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total400 µL
Vial TypeInsert
Height (Metric)31 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler
Volume (Metric) Usable350 μL
For Use With (Equipment)National 11 mm Crimp Top Vials
Unit SizePack of 100
C4010-629PFull specifications
Pack of 100325 μLPolypropyleneConical, GraduatedConical Base; Graduation MarksContact Us ›
Description300µL Insert, Graduation Marks
Diameter (Metric) Inner6 mm
ColorClear
Diameter (Metric) Outer6 mm
MaterialPolypropylene
Volume (Metric)325 μL
FeaturesConical Base; Graduation Marks
Insert TypeConical, Graduated
Volume (Metric) Residual<2 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total325 µL
Vial TypeInsert
Height (Metric)30 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler
Closure Size200 μL
Volume (Metric) Usable250 μL
For Use With (Equipment)8 mm HPLC, GC Autosampler Vial
Unit SizePack of 100
C4010-630Full specifications
Pack of 100375 μLGlassConicalPolyspring centers and cushions insertContact Us ›
Description300µL Polyspring Insert
Diameter (Metric) Inner6 mm
ColorClear
Diameter (Metric) Outer6 mm
MaterialGlass
Volume (Metric)375 μL
FeaturesPolyspring centers and cushions insert
Insert TypeConical
Volume (Metric) Residual<1 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total375 µL
Vial TypeInsert
Height (Metric)29 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler
Volume (Metric) Usable300 μL
For Use With (Equipment)National 11 mm Crimp Top Vials
Unit SizePack of 100
C4011-631Full specifications
Pack of 500500 μLGlassFlat BottomInsert rests on bottom of vialContact Us ›
Description400µL MicroSert Insert
Diameter (Metric) Inner6 mm
ColorClear
Diameter (Metric) Outer6 mm
MaterialGlass
Volume (Metric)500 μL
FeaturesInsert rests on bottom of vial
Insert TypeFlat Bottom
Volume (Metric) Residual<25 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total500 μL
Vial TypeInsert
Height (Metric)31 mm
Quantity500
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler
Volume (Metric) Usable400 μL
For Use With (Equipment)National 11 mm Crimp Top Vials
Unit SizePack of 500
C4011-631PFull specifications
Pack of 500500 μLPolypropyleneMolded plastic insertContact Us ›
Description400µL MicroSert Insert
Diameter (Metric) Inner6 mm
ColorClear
Diameter (Metric) Outer6 mm
MaterialPolypropylene
Volume (Metric)500 μL
FeaturesMolded plastic insert
Volume (Metric) Residual<25 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total500 µL
Vial TypeInsert
Height (Metric)31 mm
Quantity500
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler
Volume (Metric) Usable400 μL
For Use With (Equipment)Target DP Vials
Unit SizePack of 500
C4010-S629Full specifications
Pack of 100375 μLGlassConical, SilanizedInsert rests on bottom of vialContact Us ›
Description300µL Insert, silanized
Diameter (Metric) Inner6 mm
ColorClear
Diameter (Metric) Outer6 mm
MaterialGlass
Volume (Metric)375 μL
FeaturesInsert rests on bottom of vial
Insert TypeConical, Silanized
Volume (Metric) Residual<1 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total375 µL
Vial TypeInsert
Height (Metric)29 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler
Volume (Metric) Usable300 μL
For Use With (Equipment)National 11 mm Crimp Top Vials
Unit SizePack of 100
C4010-S630Full specifications
Pack of 100375 μLGlassConical, SilanizedPrecision-formed tip, NSC silanizedContact Us ›
Description300µL Polyspring Insert, silanized
Diameter (Metric) Inner6 mm
ColorClear
Diameter (Metric) Outer6 mm
MaterialGlass
Volume (Metric)375 μL
FeaturesPrecision-formed tip, NSC silanized
Insert TypeConical, Silanized
Volume (Metric) Residual<1 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total375 µL
Vial TypeInsert
Height (Metric)29 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler
Volume (Metric) Usable300 μL
For Use With (Equipment)National 11 mm Crimp Top Vials
Unit SizePack of 100
Showing 10 of 12Show more
  • Pulled point inserts are an economical choice for noncritical applications
  • Nominal O.D. of 6mm to fit Target wide-opening vials

For use with:

  • Target LoVial Wide-Opening vials
  • Snap-It™ Wide-Opening Snap Cap vials
  • Target DP™ vials
  • Target Wide-Opening vials
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.