Thermo Scientific™

11 mm Autosampler Vial Crimp Caps

Catalog number: C4011-1A
Thermo Scientific™

11 mm Autosampler Vial Crimp Caps

Catalog number: C4011-1A

Minimize contamination from handling with convenient, pre-assembled Thermo Scientific™ 11 mm Autosampler Vial Crimp Caps, compatible with most HPLC and GC autosamplers. Choose from Thermo Scientific™, National™ or Chromacol™ brands.

 
Catalog Number
C4011-1A
Unit Size
Case of 1000
Closure Color
Silver
Septum
Clear PTFE/Natural Red Rubber
Price (USD)
Full specifications
Closure ColorSilver
DescriptionPTFE/Red Rubber Septa; Silver Aluminum Seal; 1000/Cs.
ColorSilver, Clear, Red
MaterialPTFE, Rubber
SeptumClear PTFE/Natural Red Rubber
Closure MaterialAluminum
Closure TypeCrimp
Quantity1000
Thickness (Metric)1.0 mm
Closure Size11 mm
For Use With (Equipment)12x32 Crimp Vials
Unit SizeCase of 1000
Catalog Number
Unit Size
Case of 1000
Closure Color
Silver
Septum
Clear PTFE/Rubber
Price (USD)
Full specifications
Closure ColorSilver
DescriptionFor use w/Thermo Scientific , PE, Claris, Shimadzu, Varian
ColorSilver, Clear, Clear Septa
MaterialPTFE, Rubber
Volume (Metric)2 mL
SeptumClear PTFE/Rubber
Closure MaterialAluminum
Closure TypeCrimp
Quantity1000
Thickness (Metric)1.0 mm
Closure Size11 mm
For Use With (Equipment)Thermo Scientific, PE, Claris, Shimadzu, and Varian Vials
Unit SizeCase of 1000
Catalog Number
Unit Size
Pack of 100
Closure Color
Silver
Septum
Clear PTFE/Natural Red Rubber
Price (USD)
Full specifications
Closure ColorSilver
DescriptionPTFE/Red Rubber Septa; Silver Aluminum Seal; 100/Pk.
ColorSilver, Clear, Red
MaterialPTFE, Rubber
SeptumClear PTFE/Natural Red Rubber
Closure MaterialAluminum
Closure TypeCrimp
Quantity100
Thickness (Metric)1.0 mm
Closure Size11 mm
For Use With (Equipment)12x32 Crimp Vials
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Closure Color
Blue
Septum
Clear PTFE/Natural Red Rubber
Price (USD)
Full specifications
Closure ColorBlue
DescriptionPTFE/Red Rubber Septa; Blue Aluminum Seal
ColorBlue, Clear, Red
MaterialPTFE, Rubber
SeptumClear PTFE/Natural Red Rubber
Closure MaterialAluminum
Closure TypeCrimp
Quantity100
Thickness (Metric)1.0 mm
Closure Size11 mm
For Use With (Equipment)12x32 Crimp Vials
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Closure Color
Green
Septum
Clear PTFE/Natural Red Rubber
Price (USD)
Full specifications
Closure ColorGreen
DescriptionPTFE/Red Rubber Septa; Green Aluminum Seal
ColorGreen, Clear, Red
MaterialPTFE, Rubber
SeptumClear PTFE/Natural Red Rubber
Closure MaterialAluminum
Closure TypeCrimp
Quantity100
Thickness (Metric)1.0 mm
Closure Size11 mm
For Use With (Equipment)12x32 Crimp Vials
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Closure Color
Red
Septum
Clear PTFE/Natural Red Rubber
Price (USD)
Full specifications
Closure ColorRed
DescriptionPTFE/Red Rubber Septa; Red Aluminum Seal
ColorRed, Clear
MaterialPTFE, Rubber
SeptumClear PTFE/Natural Red Rubber
Closure MaterialAluminum
Closure TypeCrimp
Quantity100
Thickness (Metric)1.0 mm
Closure Size11 mm
For Use With (Equipment)12x32 Crimp Vials
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Closure Color
Yellow
Septum
Clear PTFE/Natural Red Rubber
Price (USD)
Full specifications
Closure ColorYellow
Description11mm Crimp Cap, 5.5mm hole
ColorYellow, Clear, Red
MaterialPTFE, Rubber
SeptumClear PTFE/Natural Red Rubber
Closure MaterialAluminum
Closure TypeCrimp
Quantity100
Thickness (Metric)1.0 mm
Closure Size11 mm
For Use With (Equipment)12x32 Crimp Vials
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Closure Color
Silver
Septum
Red PTFE/White Silicone/Red PTFE
Price (USD)
Full specifications
Closure ColorSilver
DescriptionPTFE/Silicone/PTFE Septa; Silver Aluminum Seal
ColorSilver, Red, White
MaterialPTFE, Silicone
SeptumRed PTFE/White Silicone/Red PTFE
Material (Septa)Silicone/PTFE
Closure MaterialAluminum
Closure TypeCrimp
Quantity100
Thickness (Metric)1.0 mm
Closure Size11 mm
For Use With (Equipment)12x32 Crimp Vials
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Closure Color
Silver
Septum
Red PTFE/White Silicone
Price (USD)
Full specifications
Closure ColorSilver
DescriptionPTFE/Silicone Septa; Silver Aluminum Seal
ColorSilver, Red, White
MaterialPTFE, Silicone
SeptumRed PTFE/White Silicone
Material (Septa)Silicone/PTFE
Closure MaterialAluminum
Closure TypeCrimp
Quantity100
Thickness (Metric)1.3 mm
Closure Size11 mm
For Use With (Equipment)12x32 Crimp Vials
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Closure Color
Blue
Septum
Red PTFE/White Silicone
Price (USD)
Full specifications
Closure ColorBlue
DescriptionRed PTFE/White Silicone Septa; Blue Aluminum Seal
ColorBlue, Red, White
MaterialPTFE, Silicone
SeptumRed PTFE/White Silicone
Material (Septa)Silicone/PTFE
Closure MaterialAluminum
Closure TypeCrimp
Quantity100
Thickness (Metric)1.3 mm
Closure Size11 mm
For Use With (Equipment)12x32 Crimp Vials
Unit SizePack of 100
Showing 10 of 40Show more
Catalog NumberSpecificationsUnit SizeClosure ColorSeptumPrice (USD)
C4011-1AFull specifications
Case of 1000SilverClear PTFE/Natural Red RubberContact Us ›
Closure ColorSilver
DescriptionPTFE/Red Rubber Septa; Silver Aluminum Seal; 1000/Cs.
ColorSilver, Clear, Red
MaterialPTFE, Rubber
SeptumClear PTFE/Natural Red Rubber
Closure MaterialAluminum
Closure TypeCrimp
Quantity1000
Thickness (Metric)1.0 mm
Closure Size11 mm
For Use With (Equipment)12x32 Crimp Vials
Unit SizeCase of 1000
60180-705Full specifications
Case of 1000SilverClear PTFE/RubberContact Us ›
Closure ColorSilver
DescriptionFor use w/Thermo Scientific , PE, Claris, Shimadzu, Varian
ColorSilver, Clear, Clear Septa
MaterialPTFE, Rubber
Volume (Metric)2 mL
SeptumClear PTFE/Rubber
Closure MaterialAluminum
Closure TypeCrimp
Quantity1000
Thickness (Metric)1.0 mm
Closure Size11 mm
For Use With (Equipment)Thermo Scientific, PE, Claris, Shimadzu, and Varian Vials
Unit SizeCase of 1000
C4011-1APFull specifications
Pack of 100SilverClear PTFE/Natural Red RubberContact Us ›
Closure ColorSilver
DescriptionPTFE/Red Rubber Septa; Silver Aluminum Seal; 100/Pk.
ColorSilver, Clear, Red
MaterialPTFE, Rubber
SeptumClear PTFE/Natural Red Rubber
Closure MaterialAluminum
Closure TypeCrimp
Quantity100
Thickness (Metric)1.0 mm
Closure Size11 mm
For Use With (Equipment)12x32 Crimp Vials
Unit SizePack of 100
C4011-98BFull specifications
Pack of 100BlueClear PTFE/Natural Red RubberContact Us ›
Closure ColorBlue
DescriptionPTFE/Red Rubber Septa; Blue Aluminum Seal
ColorBlue, Clear, Red
MaterialPTFE, Rubber
SeptumClear PTFE/Natural Red Rubber
Closure MaterialAluminum
Closure TypeCrimp
Quantity100
Thickness (Metric)1.0 mm
Closure Size11 mm
For Use With (Equipment)12x32 Crimp Vials
Unit SizePack of 100
C4011-98GFull specifications
Pack of 100GreenClear PTFE/Natural Red RubberContact Us ›
Closure ColorGreen
DescriptionPTFE/Red Rubber Septa; Green Aluminum Seal
ColorGreen, Clear, Red
MaterialPTFE, Rubber
SeptumClear PTFE/Natural Red Rubber
Closure MaterialAluminum
Closure TypeCrimp
Quantity100
Thickness (Metric)1.0 mm
Closure Size11 mm
For Use With (Equipment)12x32 Crimp Vials
Unit SizePack of 100
C4011-98RFull specifications
Pack of 100RedClear PTFE/Natural Red RubberContact Us ›
Closure ColorRed
DescriptionPTFE/Red Rubber Septa; Red Aluminum Seal
ColorRed, Clear
MaterialPTFE, Rubber
SeptumClear PTFE/Natural Red Rubber
Closure MaterialAluminum
Closure TypeCrimp
Quantity100
Thickness (Metric)1.0 mm
Closure Size11 mm
For Use With (Equipment)12x32 Crimp Vials
Unit SizePack of 100
C4011-98YFull specifications
Pack of 100YellowClear PTFE/Natural Red RubberContact Us ›
Closure ColorYellow
Description11mm Crimp Cap, 5.5mm hole
ColorYellow, Clear, Red
MaterialPTFE, Rubber
SeptumClear PTFE/Natural Red Rubber
Closure MaterialAluminum
Closure TypeCrimp
Quantity100
Thickness (Metric)1.0 mm
Closure Size11 mm
For Use With (Equipment)12x32 Crimp Vials
Unit SizePack of 100
C4011-2AFull specifications
Pack of 100SilverRed PTFE/White Silicone/Red PTFEContact Us ›
Closure ColorSilver
DescriptionPTFE/Silicone/PTFE Septa; Silver Aluminum Seal
ColorSilver, Red, White
MaterialPTFE, Silicone
SeptumRed PTFE/White Silicone/Red PTFE
Material (Septa)Silicone/PTFE
Closure MaterialAluminum
Closure TypeCrimp
Quantity100
Thickness (Metric)1.0 mm
Closure Size11 mm
For Use With (Equipment)12x32 Crimp Vials
Unit SizePack of 100
C4011-4AFull specifications
Pack of 100SilverRed PTFE/White SiliconeContact Us ›
Closure ColorSilver
DescriptionPTFE/Silicone Septa; Silver Aluminum Seal
ColorSilver, Red, White
MaterialPTFE, Silicone
SeptumRed PTFE/White Silicone
Material (Septa)Silicone/PTFE
Closure MaterialAluminum
Closure TypeCrimp
Quantity100
Thickness (Metric)1.3 mm
Closure Size11 mm
For Use With (Equipment)12x32 Crimp Vials
Unit SizePack of 100
C4011-4BFull specifications
Pack of 100BlueRed PTFE/White SiliconeContact Us ›
Closure ColorBlue
DescriptionRed PTFE/White Silicone Septa; Blue Aluminum Seal
ColorBlue, Red, White
MaterialPTFE, Silicone
SeptumRed PTFE/White Silicone
Material (Septa)Silicone/PTFE
Closure MaterialAluminum
Closure TypeCrimp
Quantity100
Thickness (Metric)1.3 mm
Closure Size11 mm
For Use With (Equipment)12x32 Crimp Vials
Unit SizePack of 100
Showing 10 of 40Show more

Aluminum crimp seals with prefitted septa fit all 11 mm crimp top and snap cap vials. Must be applied with a crimping tool.

Ordering Information:
Shipped in sealed polybags to prevent contamination during transport.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.