Thermo Scientific™
Manual Crimpers, Decrimpers and Decrimping Pliers
Catalog number: C4020-100
Thermo Scientific™
Manual Crimpers, Decrimpers and Decrimping Pliers
Catalog number: C4020-100
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers, also for use with National or Chromacol vials.
Catalog Number
C4020-100
Unit Size
Each 1
Description
Manual Crimper, Standard
Product Line
National
Size
20 mm
Price (USD)
Full specifications
For Use With (Application)Industrial Chromatography
For Use With (Equipment)20 mm Standard Seal, National Vial, Chromacol Vial
Product LineNational
Size20 mm
TypeManual Crimper
DescriptionManual Crimper, Standard
No. per Pack1 Ea.
Unit SizeEach 1
Catalog Number
Unit Size
Each
Description
Manual Crimper, Standard
Product Line
Chromacol
Size
8 mm
Price (USD)
Full specifications
For Use With (Application)Industrial Chromatography
For Use With (Equipment)8mm Standard Seal, National Vial, Chromacol Vial
Product LineChromacol
Size8 mm
TypeManual Crimper
DescriptionManual Crimper, Standard
No. per Pack1 Ea.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Manual Crimper, Standard
Product Line
Chromacol
Size
11 mm
Price (USD)
Full specifications
For Use With (Application)Industrial Chromatography
For Use With (Equipment)11 mm Standard Seal, National Vial, Chromacol Vial
Product LineChromacol
Size11 mm
TypeManual Crimper
DescriptionManual Crimper, Standard
No. per Pack1 Ea.
Unit SizeEach
Catalog Number
Unit Size
-
Description
Manual Crimper, Standard
Product Line
Chromacol
Size
20 mm
Price (USD)
Full specifications
For Use With (Application)Industrial Chromatography
For Use With (Equipment)20 mm Standard Seal, National Vial, Chromacol Vial
Product LineChromacol
Size20 mm
TypeManual Crimper
DescriptionManual Crimper, Standard
No. per Pack1 Ea.
Unit Size-
Catalog Number
Unit Size
Each
Description
Manual Crimper, Standard
Product Line
National
Size
8 mm
Price (USD)
Full specifications
For Use With (Application)Industrial Chromatography
For Use With (Equipment)8mm Standard Seal, National Vial, Chromacol Vial
Product LineNational
Size8 mm
TypeManual Crimper
DescriptionManual Crimper, Standard
No. per Pack1 Ea.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Manual Crimper, Standard
Product Line
National
Size
11 mm
Price (USD)
Full specifications
For Use With (Application)Industrial Chromatography
For Use With (Equipment)11 mm Standard Seal, National Vial, Chromacol Vial
Product LineNational
Size11 mm
TypeManual Crimper
DescriptionManual Crimper, Standard
No. per Pack1 Ea.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Manual Crimper, Standard
Product Line
National
Size
13 mm
Price (USD)
Full specifications
For Use With (Application)Industrial Chromatography
For Use With (Equipment)13 mm Standard Seal, National Vial, Chromacol Vial
Product LineNational
Size13 mm
TypeManual Crimper
DescriptionManual Crimper, Standard
No. per Pack1 Ea.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Manual Crimper, Flip-Off
Product Line
National
Size
13 mm
Price (USD)
Full specifications
For Use With (Application)Industrial Chromatography
For Use With (Equipment)National Vial, Chromacol Vial, Repeatable Crimp, Remove Seal, Attach Seals to Chromatography, Serum Vial
Product LineNational
Size13 mm
TypeManual Crimper
DescriptionManual Crimper, Flip-Off
No. per Pack1 Ea.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Manual Crimper, Standard
Product Line
National
Size
28 mm
Price (USD)
Full specifications
For Use With (Application)Industrial Chromatography
For Use With (Equipment)National Vial, Chromacol Vial, Repeatable Crimp, Remove Seal, Attach Seals to Chromatography, Serum Vial
Product LineNational
Size28 mm
TypeManual Crimper
DescriptionManual Crimper, Standard
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Manual Crimper, Standard
Product Line
National
Size
30 mm
Price (USD)
Full specifications
For Use With (Application)Industrial Chromatography
For Use With (Equipment)National Vial, Chromacol Vial
Product LineNational
Size30 mm
TypePrecision Crimper
DescriptionManual Crimper, Standard
Unit SizeEach
Showing 10 of 26Show more
Catalog NumberSpecificationsUnit SizeDescriptionProduct LineSizePrice (USD)
C4020-100Full specifications
Each 1Manual Crimper, StandardNational20 mmContact Us ›
For Use With (Application)Industrial Chromatography
For Use With (Equipment)20 mm Standard Seal, National Vial, Chromacol Vial
Product LineNational
Size20 mm
TypeManual Crimper
DescriptionManual Crimper, Standard
No. per Pack1 Ea.
Unit SizeEach 1
CR-8CFull specifications
EachManual Crimper, StandardChromacol8 mmRequest A Quote
For Use With (Application)Industrial Chromatography
For Use With (Equipment)8mm Standard Seal, National Vial, Chromacol Vial
Product LineChromacol
Size8 mm
TypeManual Crimper
DescriptionManual Crimper, Standard
No. per Pack1 Ea.
Unit SizeEach
CR-11CFull specifications
EachManual Crimper, StandardChromacol11 mmRequest A Quote
For Use With (Application)Industrial Chromatography
For Use With (Equipment)11 mm Standard Seal, National Vial, Chromacol Vial
Product LineChromacol
Size11 mm
TypeManual Crimper
DescriptionManual Crimper, Standard
No. per Pack1 Ea.
Unit SizeEach
CR-20CFull specifications
-Manual Crimper, StandardChromacol20 mmRequest A Quote
For Use With (Application)Industrial Chromatography
For Use With (Equipment)20 mm Standard Seal, National Vial, Chromacol Vial
Product LineChromacol
Size20 mm
TypeManual Crimper
DescriptionManual Crimper, Standard
No. per Pack1 Ea.
Unit Size-
C4008-100Full specifications
EachManual Crimper, StandardNational8 mmContact Us ›
For Use With (Application)Industrial Chromatography
For Use With (Equipment)8mm Standard Seal, National Vial, Chromacol Vial
Product LineNational
Size8 mm
TypeManual Crimper
DescriptionManual Crimper, Standard
No. per Pack1 Ea.
Unit SizeEach
C4012-100Full specifications
Each Manual Crimper, StandardNational11 mmContact Us ›
For Use With (Application)Industrial Chromatography
For Use With (Equipment)11 mm Standard Seal, National Vial, Chromacol Vial
Product LineNational
Size11 mm
TypeManual Crimper
DescriptionManual Crimper, Standard
No. per Pack1 Ea.
Unit SizeEach
C4013-100Full specifications
EachManual Crimper, StandardNational13 mmContact Us ›
For Use With (Application)Industrial Chromatography
For Use With (Equipment)13 mm Standard Seal, National Vial, Chromacol Vial
Product LineNational
Size13 mm
TypeManual Crimper
DescriptionManual Crimper, Standard
No. per Pack1 Ea.
Unit SizeEach
C4013-100FFull specifications
EachManual Crimper, Flip-OffNational13 mmContact Us ›
For Use With (Application)Industrial Chromatography
For Use With (Equipment)National Vial, Chromacol Vial, Repeatable Crimp, Remove Seal, Attach Seals to Chromatography, Serum Vial
Product LineNational
Size13 mm
TypeManual Crimper
DescriptionManual Crimper, Flip-Off
No. per Pack1 Ea.
Unit SizeEach
C4028-100Full specifications
EachManual Crimper, StandardNational28 mmContact Us ›
For Use With (Application)Industrial Chromatography
For Use With (Equipment)National Vial, Chromacol Vial, Repeatable Crimp, Remove Seal, Attach Seals to Chromatography, Serum Vial
Product LineNational
Size28 mm
TypeManual Crimper
DescriptionManual Crimper, Standard
Unit SizeEach
C4030-100Full specifications
EachManual Crimper, StandardNational30 mmContact Us ›
For Use With (Application)Industrial Chromatography
For Use With (Equipment)National Vial, Chromacol Vial
Product LineNational
Size30 mm
TypePrecision Crimper
DescriptionManual Crimper, Standard
Unit SizeEach
Showing 10 of 26Show more
Manual Crimpers
Lightweight, precision crimpers attach seals to chromatography or serum vials. Available for standard aluminum or flip-off seals.
  • Factory set to properly crimp standard-thickness seals
  • Adjustable handle setting screw allows proper crimps with septa of varying thickness
Manual Decrimpers
Forged pliers feature a fitted gripping surface designed to allow removal of the listed sizes of seals.
  • Simply grip pliers around top of seal, squeeze and pry upward to remove
Manual Decrimping Pliers
Place decrimping head squarely on seal and squeeze.
  • Minimizes damage to vial rim with chromatography or serum vials
  • Removes standard aluminum seals and many steel seals
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.