Thermo Scientific™
9mm Certified Clear Wide Opening Screw Thread Kits
Catalog number: CERT5000-79
Thermo Scientific™
9mm Certified Clear Wide Opening Screw Thread Kits
Catalog number: CERT5000-79
Use certified clean, low particle, low background vials for your LC/MS, GC/MS and chromatography high sensitivity applications. Thermo Scientific™ 9mm Wide Opening Screw Thread Certified Vial Kits are fully lot-tested by multiple methods for 15 critical parameters. Every pack comes with a Certificate of Conformance. Manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) glass, vials are available in assembled and unassembled kits. An assortment of wide opening 9mm vials and a selection of closures are available to suit your application.
Catalog Number
CERT5000-79
Unit Size
Pack of 100
Description
Convenience Kit, Certified 9mm Wide Opening High Recovery Screw Vial, 1.7mL
Closure Color
Gray
Septum
Bonded Clear PTFE/Clear Silicone
Price (USD)
Full specifications
Closure ColorGray
DescriptionConvenience Kit, Certified 9mm Wide Opening High Recovery Screw Vial, 1.7mL
ColorClear
MaterialGlass, PTFE, Silicone
Pre-slitYes
SeptumBonded Clear PTFE/Clear Silicone
No. per Pack100
Product LineLC-GC CERT
Marking SpotNo
Unit SizePack of 100
Catalog Number
Unit Size
Each
Description
Convenience Kit, Certified 9mm Wide Opening Total Recovery Screw Vial, 1/10µL Reservoir
Closure Color
Gray
Septum
Bonded Clear PTFE/Clear Silicone
Price (USD)
Full specifications
Closure ColorGray
DescriptionConvenience Kit, Certified 9mm Wide Opening Total Recovery Screw Vial, 1/10µL Reservoir
ColorClear
MaterialGlass, PTFE, Silicone
Pre-slitYes
SeptumBonded Clear PTFE/Clear Silicone
No. per Pack100
Closure TypeScrew Thread
Marking SpotNo
Unit SizeEach
Catalog Number
Unit Size
Pack of 100
Description
Convenience Kit, Certified 9mm Wide Opening SureStop Vial, 2mL
Closure Color
Blue
Septum
Ivory PTFE/Red Rubber
Price (USD)
Full specifications
Closure ColorBlue
DescriptionConvenience Kit, Certified 9mm Wide Opening SureStop Vial, 2mL
ColorClear
MaterialGlass, PTFE, Rubber
Pre-slitNo
SeptumIvory PTFE/Red Rubber
No. per Pack100
Product LineLC-GC CERT
Marking SpotYes
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Description
Convenience Kit, Certified 9mm Wide Opening SureStop Vial, 2mL
Closure Color
Blue
Septum
Red PTFE/White Silicone
Price (USD)
Full specifications
Closure ColorBlue
DescriptionConvenience Kit, Certified 9mm Wide Opening SureStop Vial, 2mL
ColorClear
MaterialGlass, PTFE, Silicone
Pre-slitNo
SeptumRed PTFE/White Silicone
No. per Pack100
Product LineLC-GC CERT
Marking SpotYes
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Description
Convenience Kit, Certified 9mm Wide Opening SureStop Vial, 2mL
Closure Color
Blue
Septum
Blue PTFE/White Silicone
Price (USD)
Full specifications
Closure ColorBlue
DescriptionConvenience Kit, Certified 9mm Wide Opening SureStop Vial, 2mL
ColorClear
MaterialGlass, PTFE, Silicone
Pre-slitYes
SeptumBlue PTFE/White Silicone
No. per Pack100
Product LineLC-GC CERT
Marking SpotYes
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Description
Convenience Kit, Certified 9mm Wide Opening Screw Vial, 2mL
Closure Color
Blue
Septum
Ivory PTFE/Red Rubber
Price (USD)
Full specifications
Closure ColorBlue
DescriptionConvenience Kit, Certified 9mm Wide Opening Screw Vial, 2mL
ColorClear
MaterialGlass, PTFE, Rubber
Pre-slitNo
SeptumIvory PTFE/Red Rubber
No. per Pack100
Product LineLC-GC CERT
Closure TypeScrew Thread
Marking SpotNo
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Description
Convenience Kit, Certified 9mm Wide Opening Screw Vial, 2mL
Closure Color
Blue
Septum
Ivory PTFE/Red Rubber
Price (USD)
Full specifications
Closure ColorBlue
DescriptionConvenience Kit, Certified 9mm Wide Opening Screw Vial, 2mL
ColorClear
MaterialGlass, PTFE, Rubber
Pre-slitNo
SeptumIvory PTFE/Red Rubber
No. per Pack100
Product LineLC-GC CERT
Closure TypeScrew Thread
Marking SpotYes
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Description
Convenience Kit, Certified 9mm Wide Opening Screw Vial, 2mL
Closure Color
Blue
Septum
Red PTFE/White Silicone
Price (USD)
Full specifications
Closure ColorBlue
DescriptionConvenience Kit, Certified 9mm Wide Opening Screw Vial, 2mL
ColorClear
MaterialGlass, PTFE, Silicone
Pre-slitNo
SeptumRed PTFE/White Silicone
No. per Pack100
Product LineLC-GC CERT
Closure TypeScrew Thread
Marking SpotYes
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Description
Convenience Kit, 9mm Wide Opening 1.7mLHigh Recovery Screw Vial with 30μL Reservoir
Closure Color
Blue
Septum
Bonded Clear PTFE/Clear Silicone
Price (USD)
Full specifications
Closure ColorBlue
DescriptionConvenience Kit, 9mm Wide Opening 1.7mLHigh Recovery Screw Vial with 30μL Reservoir
ColorClear
MaterialGlass, PTFE, Silicone
Pre-slitNo
SeptumBonded Clear PTFE/Clear Silicone
No. per Pack100
Product LineMass Spec Certified
Marking SpotNo
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Description
Convenience Kit, 9mm Wide Opening 1.5mLTotal Recovery Screw Vial with 10μL Reservoir
Closure Color
Blue
Septum
Bonded Clear PTFE/Clear Silicone
Price (USD)
Full specifications
Closure ColorBlue
DescriptionConvenience Kit, 9mm Wide Opening 1.5mLTotal Recovery Screw Vial with 10μL Reservoir
ColorClear
MaterialGlass, PTFE, Silicone
Pre-slitNo
SeptumBonded Clear PTFE/Clear Silicone
No. per Pack100
Product LineMass Spec Certified
Closure TypeScrew Thread
Marking SpotNo
Unit SizePack of 100
Showing 10 of 21Show more
Catalog NumberSpecificationsUnit SizeDescriptionClosure ColorSeptumPrice (USD)
CERT5000-79Full specifications
Pack of 100Convenience Kit, Certified 9mm Wide Opening High Recovery Screw Vial, 1.7mLGrayBonded Clear PTFE/Clear SiliconeContact Us ›
Closure ColorGray
DescriptionConvenience Kit, Certified 9mm Wide Opening High Recovery Screw Vial, 1.7mL
ColorClear
MaterialGlass, PTFE, Silicone
Pre-slitYes
SeptumBonded Clear PTFE/Clear Silicone
No. per Pack100
Product LineLC-GC CERT
Marking SpotNo
Unit SizePack of 100
CERT4000-993Full specifications
EachConvenience Kit, Certified 9mm Wide Opening Total Recovery Screw Vial, 1/10µL ReservoirGrayBonded Clear PTFE/Clear SiliconeRequest A Quote
Closure ColorGray
DescriptionConvenience Kit, Certified 9mm Wide Opening Total Recovery Screw Vial, 1/10µL Reservoir
ColorClear
MaterialGlass, PTFE, Silicone
Pre-slitYes
SeptumBonded Clear PTFE/Clear Silicone
No. per Pack100
Closure TypeScrew Thread
Marking SpotNo
Unit SizeEach
CERT5000-580WFull specifications
Pack of 100Convenience Kit, Certified 9mm Wide Opening SureStop Vial, 2mLBlueIvory PTFE/Red RubberContact Us ›
Closure ColorBlue
DescriptionConvenience Kit, Certified 9mm Wide Opening SureStop Vial, 2mL
ColorClear
MaterialGlass, PTFE, Rubber
Pre-slitNo
SeptumIvory PTFE/Red Rubber
No. per Pack100
Product LineLC-GC CERT
Marking SpotYes
Unit SizePack of 100
CERT5000-578WFull specifications
Pack of 100Convenience Kit, Certified 9mm Wide Opening SureStop Vial, 2mLBlueRed PTFE/White SiliconeContact Us ›
Closure ColorBlue
DescriptionConvenience Kit, Certified 9mm Wide Opening SureStop Vial, 2mL
ColorClear
MaterialGlass, PTFE, Silicone
Pre-slitNo
SeptumRed PTFE/White Silicone
No. per Pack100
Product LineLC-GC CERT
Marking SpotYes
Unit SizePack of 100
CERT5000-593WFull specifications
Pack of 100Convenience Kit, Certified 9mm Wide Opening SureStop Vial, 2mLBlueBlue PTFE/White SiliconeContact Us ›
Closure ColorBlue
DescriptionConvenience Kit, Certified 9mm Wide Opening SureStop Vial, 2mL
ColorClear
MaterialGlass, PTFE, Silicone
Pre-slitYes
SeptumBlue PTFE/White Silicone
No. per Pack100
Product LineLC-GC CERT
Marking SpotYes
Unit SizePack of 100
CERT5000-80Full specifications
Pack of 100Convenience Kit, Certified 9mm Wide Opening Screw Vial, 2mLBlueIvory PTFE/Red RubberContact Us ›
Closure ColorBlue
DescriptionConvenience Kit, Certified 9mm Wide Opening Screw Vial, 2mL
ColorClear
MaterialGlass, PTFE, Rubber
Pre-slitNo
SeptumIvory PTFE/Red Rubber
No. per Pack100
Product LineLC-GC CERT
Closure TypeScrew Thread
Marking SpotNo
Unit SizePack of 100
CERT5000-80WFull specifications
Pack of 100Convenience Kit, Certified 9mm Wide Opening Screw Vial, 2mLBlueIvory PTFE/Red RubberContact Us ›
Closure ColorBlue
DescriptionConvenience Kit, Certified 9mm Wide Opening Screw Vial, 2mL
ColorClear
MaterialGlass, PTFE, Rubber
Pre-slitNo
SeptumIvory PTFE/Red Rubber
No. per Pack100
Product LineLC-GC CERT
Closure TypeScrew Thread
Marking SpotYes
Unit SizePack of 100
CERT5000-92WFull specifications
Pack of 100Convenience Kit, Certified 9mm Wide Opening Screw Vial, 2mLBlueRed PTFE/White SiliconeContact Us ›
Closure ColorBlue
DescriptionConvenience Kit, Certified 9mm Wide Opening Screw Vial, 2mL
ColorClear
MaterialGlass, PTFE, Silicone
Pre-slitNo
SeptumRed PTFE/White Silicone
No. per Pack100
Product LineLC-GC CERT
Closure TypeScrew Thread
Marking SpotYes
Unit SizePack of 100
MSCERT5000-32Full specifications
Pack of 100Convenience Kit, 9mm Wide Opening 1.7mLHigh Recovery Screw Vial with 30μL ReservoirBlueBonded Clear PTFE/Clear SiliconeContact Us ›
Closure ColorBlue
DescriptionConvenience Kit, 9mm Wide Opening 1.7mLHigh Recovery Screw Vial with 30μL Reservoir
ColorClear
MaterialGlass, PTFE, Silicone
Pre-slitNo
SeptumBonded Clear PTFE/Clear Silicone
No. per Pack100
Product LineMass Spec Certified
Marking SpotNo
Unit SizePack of 100
MSCERT5000-33TRFull specifications
Pack of 100Convenience Kit, 9mm Wide Opening 1.5mLTotal Recovery Screw Vial with 10μL ReservoirBlueBonded Clear PTFE/Clear SiliconeContact Us ›
Closure ColorBlue
DescriptionConvenience Kit, 9mm Wide Opening 1.5mLTotal Recovery Screw Vial with 10μL Reservoir
ColorClear
MaterialGlass, PTFE, Silicone
Pre-slitNo
SeptumBonded Clear PTFE/Clear Silicone
No. per Pack100
Product LineMass Spec Certified
Closure TypeScrew Thread
Marking SpotNo
Unit SizePack of 100
Showing 10 of 21Show more
Unassembled kits
  • 100 vials and 100 caps with pre-assembled septa
  • Reusable two-compartment trays protect vials and closures while keeping matching supplies together
  • Clear trays make it easy to keep track of available supplies without opening containers
Assembled kits
  • 100 vials with pre-attached caps and septa
  • Packaged in convenient vial trays with clear covers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.