Thermo Scientific™

Diluflux™ Automated Gravimetric Dilutor

Catalog number: DB4000A
Thermo Scientific™

Diluflux™ Automated Gravimetric Dilutor

Catalog number: DB4000A
Accurately dispense broth medium from a Dry-Bag or bottle into suitable containers or into pre-weighed food bags using the Thermo Scientific™ Diluflux™ Automated Gravimetric Dilutor.
 
Catalog Number
DB4000A
Unit Size
-
Description
Diluflux Automated Gravimetric Dilutor
Includes
Automated Dilutor Dual Pump unit only
Shipping Weight (Metric)
9.0 kg
Material
Stainless Steel (Case)
Price (USD)
Full specifications
Shipping Weight (Metric)9.0 kg
DescriptionDiluflux Automated Gravimetric Dilutor
IncludesAutomated Dilutor Dual Pump unit only
For Use WithDry-Bags
Electrical Requirements100/127 V 220/240 V 50/60 Hz
MaterialStainless Steel (Case)
Voltage100/127 and 220/240 V
Frequency50/60 Hz
Overall Height (English)49cm
Overall Length (English)36cm
Overall Width (English)38cm
Unit Size-
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeDescriptionIncludesShipping Weight (Metric)MaterialPrice (USD)
DB4000AFull specifications
- Diluflux Automated Gravimetric DilutorAutomated Dilutor Dual Pump unit only9.0 kgStainless Steel (Case)Request A Quote
Shipping Weight (Metric)9.0 kg
DescriptionDiluflux Automated Gravimetric Dilutor
IncludesAutomated Dilutor Dual Pump unit only
For Use WithDry-Bags
Electrical Requirements100/127 V 220/240 V 50/60 Hz
MaterialStainless Steel (Case)
Voltage100/127 and 220/240 V
Frequency50/60 Hz
Overall Height (English)49cm
Overall Length (English)36cm
Overall Width (English)38cm
Unit Size-
Showing 1 of 1
The easy-to-use gravimetric diluter allows the simultaneous use of two diluents by having dual dispensing heads that are fed by dual peristaltic pumps mounted into the unit.

Dilution time: < 20 secs. for 250mL

Dilution factor: 1/2 to 1/99

Accuracy :
< 0.1g for sample weighing
> 99% for liquid mass dispensing (for a sample weight > 1g)

Contact us to arrange a demonstration of the Auto Diluter and its accessories.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.