Informational:During checkout, this product may be reported as "Out of Stock" with an Estimated Availability date. The Actual Availability date for this product may be earlier. Send inquiries regarding Actual Availability date, including the catalog number, to discoverysciences@lifetech.com.
Thermo Scientific™

GeneBLAzer™ CALCRL:RAMP3-CRE-bla FreeStyle™ 293F Cells

Catalog number: K1439
Thermo Scientific™

GeneBLAzer™ CALCRL:RAMP3-CRE-bla FreeStyle™ 293F Cells

Catalog number: K1439
Catalog Number
K1439
Unit Size
8 x 106 cells
Price (USD)
Price: 8,690.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K14398 x 106 cells
Price: 8,690.00
Your Price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The GeneBLAzer® CALCRL:RAMP3-CRE-bla FreeStyle™ 293F cells contain the human Amylin Receptor 2 (CALCRL:RAMP3), (Accession # NM_005795.3 NM_005856.1) stably integrated into the CellSensor® CRE-bla Freestyle™ 293F cell line. CellSensor® CRE-bla Freestyle™ 293F cells (Cat # K1130) contain a beta-lactamase (bla) reporter gene under control of the cAMP response element (CRE).

CALCRL:RAMP3-CRE-bla FreeStyle™ 293F cells are functionally validated for Z'-factor and EC50 concentrations of Adrenomedullin(1-52). In addition, CALCRL:RAMP3-CRE-bla FreeStyle™ 293F cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section.

Specifications

Accession Number
NM_005855.2
Assay Entry
Cell-based beta lactamase reporter gene
Cell State
Dividing Cells
Detection Method
Fluorescent
Gene Symbol
CALCRL:RAMP3
Population Status
Single Cell Clone Origin
Product Line
Freestyle™, GeneBLAzer™
Reporter Gene
BLA (Beta-Lactamase)
Target Entry
CALCRL:RAMP3
Cell Line
FreeStyle™ 293-F Cells
System Type
GeneBLAzer™
Druggable Target
GPCRs

Contents & Storage

(LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
Includes:
• GeneBLAzer™ CALCRL:RAMP3-CRE-bla Freestyle™ 293F cells
• GeneBLAzer™ CALCRL:RAMP3-CRE-bla Freestyle™ 293F Assay Protocol
• Certificate of Analysis
The GeneBLAzer™ CALCRL:RAMP3-CRE-bla Freestyle™ 293F cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.