Informational:During checkout, this product may be reported as "Out of Stock" with an Estimated Availability date. The Actual Availability date for this product may be earlier. Send inquiries regarding Actual Availability date, including the catalog number, to discoverysciences@lifetech.com.
Thermo Scientific™

GeneBLAzer™ CNR1-Gα15-NFAT-bla CHO-K1 Cells

Catalog number: K1529
Thermo Scientific™

GeneBLAzer™ CNR1-Gα15-NFAT-bla CHO-K1 Cells

Catalog number: K1529
Catalog Number
K1529
Unit Size
2 × 10^6 cells
Price (USD)
Price: 8,690.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K15292 × 10^6 cells
Price: 8,690.00
Your Price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
GeneBLAzer® CNR1-Gα15-NFAT-bla CHO-K1 cells contain the human Cannabinoid Receptor 1 (CNR1) stably integrated into the GeneBLAzer® Gα15-NFAT-bla CHO-K1 cell line. GeneBLAzer® Gα15-NFAT-bla CHO-K1 cells (Cat. no. K1537) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element and the Promiscuous G protein, Gα15.

The GeneBLAzer® CNR1-Gα15-NFAT-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of CP-55,940. In addition, GeneBLAzer® CNR1-Gα15-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.

The cannabinoid receptors are a family of GPCRs named after their endogenous ligand Delta9-tetrahydrocannabinol (Delta9-THC). Delta9-THC is the psychoactive component of Cannabis sativa (marijuana). CNR1 is present in many tissues throughout the body. The highest concentrations of cannabinoid receptors are in the neurons of the brain, the hippocampus, cerebral cortex, basal ganglia, and the cerebellum. Low-level expression occurs in the lung, testis, and uterus, as well as vascular tissue. Delta9-THC and CP55,940 are endogenous agonists of the CNR1 pathway. Several Delta9-THC and CP55,940 analogs have been synthesized, including HU 210 and WIN 55,212-2. The CNR1 and CNR2 receptors have equal affinity for cannabinoid agonists, but several antagonists that are subtype selective have been synthesized. Examples of these antagonists are AM251, LY-320135, and SR 141716A.

For Academic pricing please login or contact us at discoverysciences@invitrogen.com.

Specifications

Accession Number
NM_016083
Assay Entry
Cell-based beta lactamase reporter gene
Cell State
Dividing Cells
Detection Method
Fluorescent
For Use With (Application)
FRET
Gene Symbol
CNR1
Population Status
Single Cell Clone Origin
Product Line
GeneBLAzer™
Reporter Gene
BLA (Beta-Lactamase)
Target Entry
CNR1
Cell Line
CHO-K1
System Type
GeneBLAzer™
Druggable Target
GPCRs

Contents & Storage

Contents:
K1529 GeneBLAzer™ CNR1-Gα15-NFAT-bla CHO-K1 cells
(LiveBLAzer™-FRET B⁄G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
Includes:
• GeneBLAzer™ CNR1-Gα15-NFAT-bla CHO-K1 cells
• GeneBLAzer™ CNR1-Gα15-NFAT-bla CHO-K1 Assay Protocol
• Certificate of Analysis

Storage:
The GeneBLAzer™ CNR1-Gα15-NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.