Informational:During checkout, this product may be reported as "Out of Stock" with an Estimated Availability date. The Actual Availability date for this product may be earlier. Send inquiries regarding Actual Availability date, including the catalog number, to discoverysciences@lifetech.com.

Tango™ OPRK1-bla U2OS Cells

Catalog number: K1576

Tango™ OPRK1-bla U2OS Cells

Catalog number: K1576
Catalog Number
K1576
Unit Size
2 × 10^6 cells
Price (USD)
Price: 8,450.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K15762 × 10^6 cells
Price: 8,450.00
Your Price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Tango™ OPRK1-bla U2OS cells contain the human Opiod Receptor Kappa 1 (OPRK1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin⁄TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.

The Tango™ OPRK1-bla U2OS cells cells have been functionally validated for Z' factor and EC50 concentrations of established ligands. In addition, Tango™ OPRK1-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary for each product.

Specifications

Accession Number
NM_000912
Assay Entry
Cell-based beta-arrestin recruitment
Cell State
Dividing Cells
Cytoplasmic Tail
Tail from AVPR2 GPCR
Detection Method
Fluorescent
Gene Symbol
OPRK1
Population Status
Pool of Stable Cells
Product Line
Tango™
Reporter Gene
BLA (Beta-Lactamase)
Target Entry
OPRK1
Cell Line
U2OS
System Type
Tango™
Druggable Target
GPCRs

Contents & Storage

K1576 Tango™ OPRK1-bla U2OS cells
(LiveBLAzer™-FRET B⁄G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
Includes:
• Tango™ OPRK1-bla U2OS cells
• Tango™ OPRK1-bla U2OS Assay Protocol
• Certificate of Analysis

Storage:
The Tango™ OPRK1-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.