Informational:During checkout, this product may be reported as "Out of Stock" with an Estimated Availability date. The Actual Availability date for this product may be earlier. Send inquiries regarding Actual Availability date, including the catalog number, to discoverysciences@lifetech.com.
Thermo Scientific™

GeneBLAzer™ AR-UAS-bla Griptite™ Cells

Catalog number: K1698
Thermo Scientific™

GeneBLAzer™ AR-UAS-bla Griptite™ Cells

Catalog number: K1698
Catalog Number
K1698
Unit Size
2 x 106 cells
Price (USD)
Price: 8,600.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K16982 x 106 cells
Price: 8,600.00
Your Price:
-
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
AR-UAS-bla GripTite™ 293 cells contain the ligand-binding domain (LBD) of the rat Androgen receptor (AR) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer®UAS-bla GripTite™ 293 cell line. GeneBLAzer®UAS-bla GripTite™ cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-AR (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates. AR-UAS-bla GripTite™ 293 cells are functionally validated for Z' and EC50 concentrations of R1881. Cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.

Specifications

Alternate Agonists
Docosahexaenoic Acid, Phytanic Acid, All-trans-retinoic acid
Antagonists
RO 41-5253
Assay Entry
Cell-based beta lactamase reporter gene
Cell State
Dividing Cells
Detection Method
Fluorescent
Primary Agonist
9-cis-retinoic acid
Product Line
GeneBLAzer™
Reporter Gene
BLA (Beta-Lactamase)
Shipping Condition
Room Temperature
Target Entry
AR, NR3C4
System Type
GeneBLAzer™
Druggable Target
Nuclear Receptors

Contents & Storage

Shipping: Dry IceStorage: -80°C/Liquid NitrogenContains: AR-UAS-bla GripTite™ cells (K1698)