Informational:During checkout, this product may be reported as "Out of Stock" with an Estimated Availability date. The Actual Availability date for this product may be earlier. Send inquiries regarding Actual Availability date, including the catalog number, to discoverysciences@lifetech.com.
Thermo Scientific™

GeneBLAzer™ AVPR2-CRE-bla CHO-K1 Cells

Catalog number: K1718
Thermo Scientific™

GeneBLAzer™ AVPR2-CRE-bla CHO-K1 Cells

Catalog number: K1718
Catalog Number
K1718
Unit Size
2 × 10^6 cells
Price (USD)
Price: 8,690.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K17182 × 10^6 cells
Price: 8,690.00
Your Price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
GeneBLAzer® AVPR2 CRE-bla CHO-K1 cells contain the human Arginine Vasopressin 2 (AVPR2) receptor stably integrated into the CellSensor® CRE-bla CHO-K1 cell line. CellSensor® CRE-bla CHO-K1 cells (Cat. no. K1535) contain a beta-lactamase reporter gene under control of the Cyclic AMP Response Element (CRE) response element.

The GeneBLAzer® AVPR2 CRE-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of dDAVP. In addition, GeneBLAzer® AVPR2 CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.

Agonists and antagonists of the AVPR2 receptors are in various stages of clinical trials for the treatment of hypertension and as diuretics.

For Academic pricing please login or contact us at discoverysciences@invitrogen.com.

Specifications

Accession Number
NM_000054
Assay Entry
Cell-based beta lactamase reporter gene
Cell State
Dividing Cells
Detection Method
Fluorescent
For Use With (Application)
FRET
Gene Symbol
AVPR2
Population Status
Single Cell Clone Origin
Product Line
GeneBLAzer™
Reporter Gene
BLA (Beta-Lactamase)
Target Entry
AVPR2
Cell Line
CHO-K1
System Type
GeneBLAzer™
Druggable Target
GPCRs

Contents & Storage

Contents:
K1718 GeneBLAzer™ AVPR2 CRE-bla CHO-K1 cells
(LiveBLAzer™-FRET B⁄G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
Includes:
• GeneBLAzer™ AVPR2 CRE-bla CHO-K1 cells
• GeneBLAzer™ AVPR2 CRE-bla CHO-K1 Assay Protocol
• Certificate of Analysis

Storage:
The GeneBLAzer™ AVPR2 CRE-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.