Informational:During checkout, this product may be reported as "Out of Stock" with an Estimated Availability date. The Actual Availability date for this product may be earlier. Send inquiries regarding Actual Availability date, including the catalog number, to discoverysciences@lifetech.com.
Thermo Scientific™

GeneBLAzer™ Bradykinin-B2-NFAT-bla CHO-K1 Cells

Catalog number: K1790
Thermo Scientific™

GeneBLAzer™ Bradykinin-B2-NFAT-bla CHO-K1 Cells

Catalog number: K1790
Catalog Number
K1790
Unit Size
8 x 106 cells
Price (USD)
Price: 8,690.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K17908 x 106 cells
Price: 8,690.00
Your Price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
GeneBLAzer® Bradykinin-B2-NFAT-bla CHO-K1 cells contain the human Bradykinin Receptor B2 (B2) stably integrated into the CellSensor® CRE-bla CHO-K1 cell line. CellSensor® NFAT-bla CHO-K1 cells (Cat. no. K1534) contain a beta-lactamase reporter gene under control of the Cyclic AMP Response Element (CRE) response element.

The Bradykinin-B2-NFAT-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of Bradykinin. In addition, GeneBLAzer® Bradykinin-B2-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.

For Academic pricing please login or contact us at discoverysciences@invitrogen.com.

Specifications

Accession Number
NM_000623.2
Assay Entry
Cell-based beta lactamase reporter gene
Cell State
Dividing Cells
Detection Method
Fluorescent
For Use With (Application)
FRET
Gene Symbol
BK2
Population Status
Single Cell Clone Origin
Product Line
GeneBLAzer™
Readout
End Point
Reporter Gene
BLA (Beta-Lactamase)
Target Entry
BDKRB2
Cell Line
CHO-K1
System Type
GeneBLAzer™
Druggable Target
GPCRs

Contents & Storage

Contents:
K1790 GeneBLAzer™ Bradykinin-B2-NFAT-bla CHO-K1 cells
(LiveBLAzer™-FRET B⁄G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
Includes:
• GeneBLAzer™ Bradykinin-B2-NFAT-bla CHO-K1 cells
• GeneBLAzer™ Bradykinin-B2-NFAT-bla CHO-K1 Assay Protocol
• Certificate of Analysis

Storage:
The GeneBLAzer™ Bradykinin-B2-NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.