Informational:During checkout, this product may be reported as "Out of Stock" with an Estimated Availability date. The Actual Availability date for this product may be earlier. Send inquiries regarding Actual Availability date, including the catalog number, to discoverysciences@lifetech.com.

GeneBLAzer™ GnRHR-NFAT-bla CHO-K1 Cells

Catalog number: K1854

GeneBLAzer™ GnRHR-NFAT-bla CHO-K1 Cells

Catalog number: K1854
Catalog Number
K1854
Unit Size
1 vial
Price (USD)
Price: 8,610.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K18541 vial
Price: 8,610.00
Your Price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
GeneBLAzer® GnRHR-NFAT-bla CHO-K1 cells contain the human gonadotropin-releasing hormone receptor (GnRHR), (Accession # NM_001012763) stably integrated into the CellSensor® NFAT-bla HEK 293T cell line. CellSensor® NFAT-bla HEK 293T (Cat No. K1538) cells contain a beta-lactamase reporter gene under control of the NFAT Response Element. GeneBLAzer® GnRHR-NFAT-bla CHO-K1 cells are functionally validated for Z’-factor and EC50 concentrations of Luteinizing Hormone-Releasing Hormone (LH-RH). In addition, GeneBLAzer® GnRHR-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions.

Specifications

Accession Number
NM_000406.2
Assay Entry
Cell-based beta lactamase reporter gene
Cell State
Dividing Cells
Detection Method
Fluorescent
For Use With (Application)
FRET
Human Protein Class
G-Protein Coupled Receptor (GPCR)
Population Status
Single Cell Clone Origin
Product Line
GeneBLAzer™
Readout
End Point
Target Entry
GnRHR
Cell Line
CHO-K1
System Type
GeneBLAzer™
Druggable Target
GPCRs

Contents & Storage

GeneBLAzer™ GnRHR-NFAT-bla CHO-K1 cells are shipped on dry ice, 8 x 10^6 cells⁄vial. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.