Thermo Scientific™
Compound Discoverer™ Software
Catalog number: OPTON-30790
Thermo Scientific™
Compound Discoverer™ Software
Catalog number: OPTON-30790
Streamline and customize high-resolution accurate-mass data analysis for small molecule workflows with Thermo Scientific™ Compound Discoverer™ software. Compound Discoverer software simplifies and reduces processing, enabling you to go from data to insights faster. Leverage easy to use pre-built workflow templates or fully customizable node-based workflows, integrated compound identification capabilities, and statistical analysis tools for streamlined data analyses to deliver confident results to drive your decision making.
Catalog Number
OPTON-30790
Unit Size
-
Description
Compound Discoverer 2.1 Upgrade From SIEVE (All Versions). The installer will check for a valid SIEVE license.
Includes
Compound Discoverer 2.1 software
Price (USD)
Full specifications
TypeSoftware Upgrade
Software2.1
DescriptionCompound Discoverer 2.1 Upgrade From SIEVE (All Versions). The installer will check for a valid SIEVE license.
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesCompound Discoverer 2.1 software
Unit Size-
Catalog Number
Unit Size
-
Description
Compound Discoverer 2.1 (One License)
Includes
Compound Discoverer 2.1 software (One License)
Price (USD)
Full specifications
TypeSoftware
Software2.1
DescriptionCompound Discoverer 2.1 (One License)
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesCompound Discoverer 2.1 software (One License)
Unit Size-
Catalog Number
Unit Size
-
Description
Compound Discoverer 2.1 with Visualization
Includes
Compound Discoverer 2.1 software and 27 in. HD monitor
Price (USD)
Full specifications
TypeSoftware
Software2.1
DescriptionCompound Discoverer 2.1 with Visualization
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesCompound Discoverer 2.1 software and 27 in. HD monitor
Unit Size-
Catalog Number
Unit Size
-
Description
Compound Discoverer 2.1 (2-5 licenses)
Includes
Compound Discoverer 2.1 software, Order quantity must be between 2 to 5.
Price (USD)
Full specifications
TypeSoftware
Software2.1
DescriptionCompound Discoverer 2.1 (2-5 licenses)
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesCompound Discoverer 2.1 software, Order quantity must be between 2 to 5.
Unit Size-
Catalog Number
Unit Size
-
Description
Compound Discoverer 2.1 Upgrade From 1.0. The installer will check for a valid 1.0 license.
Includes
Compound Discoverer 2.1 software
Price (USD)
Full specifications
TypeSoftware Upgrade
Software2.1
DescriptionCompound Discoverer 2.1 Upgrade From 1.0. The installer will check for a valid 1.0 license.
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesCompound Discoverer 2.1 software
Unit Size-
Catalog Number
Unit Size
-
Description
Compound Discoverer 2.1 Upgrade From 2.0. The installer will check for a valid 2.0 license.
Includes
Compound Discoverer 2.1 software
Price (USD)
Full specifications
TypeSoftware Upgrade
Software2.1
DescriptionCompound Discoverer 2.1 Upgrade From 2.0. The installer will check for a valid 2.0 license.
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesCompound Discoverer 2.1 software
Unit Size-
Catalog Number
Unit Size
-
Description
Compound Discoverer Software with 27 in. Monitor
Includes
Mass Frontier Software 7.0 SR3 and 27 in. Monitor
Price (USD)
Full specifications
TypeSoftware
Software2
DescriptionCompound Discoverer Software with 27 in. Monitor
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesMass Frontier Software 7.0 SR3 and 27 in. Monitor
Unit Size-
Catalog Number
Unit Size
-
Description
Compound Discoverer Software
Includes
Mass Frontier Software 7.0 SR3
Price (USD)
Full specifications
TypeSoftware
Software2
DescriptionCompound Discoverer Software
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesMass Frontier Software 7.0 SR3
Unit Size-
Catalog Number
Unit Size
-
Description
Compound Discoverer Software Upgrade from SIEVE (All Versions)
Includes
Upgrade with Mass Frontier Software 7.0 SR3
Price (USD)
Full specifications
TypeSoftware Upgrade
Software2
DescriptionCompound Discoverer Software Upgrade from SIEVE (All Versions)
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesUpgrade with Mass Frontier Software 7.0 SR3
Unit Size-
Catalog Number
Unit Size
-
Description
Compound Discoverer Software Upgrade from 1.0
Includes
Upgrade Only
Price (USD)
Full specifications
TypeSoftware Upgrade
Software2
DescriptionCompound Discoverer Software Upgrade from 1.0
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesUpgrade Only
Unit Size-
Showing 10 of 19Show more
Catalog NumberSpecificationsUnit SizeDescriptionIncludesPrice (USD)
OPTON-30790Full specifications
- Compound Discoverer 2.1 Upgrade From SIEVE (All Versions). The installer will check for a valid SIEVE license.Compound Discoverer 2.1 softwareRequest A Quote
TypeSoftware Upgrade
Software2.1
DescriptionCompound Discoverer 2.1 Upgrade From SIEVE (All Versions). The installer will check for a valid SIEVE license.
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesCompound Discoverer 2.1 software
Unit Size-
OPTON-30783Full specifications
- Compound Discoverer 2.1 (One License)Compound Discoverer 2.1 software (One License)Request A Quote
TypeSoftware
Software2.1
DescriptionCompound Discoverer 2.1 (One License)
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesCompound Discoverer 2.1 software (One License)
Unit Size-
OPTON-30782Full specifications
- Compound Discoverer 2.1 with VisualizationCompound Discoverer 2.1 software and 27 in. HD monitorRequest A Quote
TypeSoftware
Software2.1
DescriptionCompound Discoverer 2.1 with Visualization
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesCompound Discoverer 2.1 software and 27 in. HD monitor
Unit Size-
OPTON-30784Full specifications
- Compound Discoverer 2.1 (2-5 licenses)Compound Discoverer 2.1 software, Order quantity must be between 2 to 5.Request A Quote
TypeSoftware
Software2.1
DescriptionCompound Discoverer 2.1 (2-5 licenses)
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesCompound Discoverer 2.1 software, Order quantity must be between 2 to 5.
Unit Size-
OPTON-30791Full specifications
- Compound Discoverer 2.1 Upgrade From 1.0. The installer will check for a valid 1.0 license.Compound Discoverer 2.1 softwareRequest A Quote
TypeSoftware Upgrade
Software2.1
DescriptionCompound Discoverer 2.1 Upgrade From 1.0. The installer will check for a valid 1.0 license.
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesCompound Discoverer 2.1 software
Unit Size-
OPTON-30792Full specifications
- Compound Discoverer 2.1 Upgrade From 2.0. The installer will check for a valid 2.0 license.Compound Discoverer 2.1 softwareRequest A Quote
TypeSoftware Upgrade
Software2.1
DescriptionCompound Discoverer 2.1 Upgrade From 2.0. The installer will check for a valid 2.0 license.
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesCompound Discoverer 2.1 software
Unit Size-
OPTON-30560Full specifications
- Compound Discoverer Software with 27 in. MonitorMass Frontier Software 7.0 SR3 and 27 in. MonitorRequest A Quote
TypeSoftware
Software2
DescriptionCompound Discoverer Software with 27 in. Monitor
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesMass Frontier Software 7.0 SR3 and 27 in. Monitor
Unit Size-
OPTON-30561Full specifications
- Compound Discoverer SoftwareMass Frontier Software 7.0 SR3Request A Quote
TypeSoftware
Software2
DescriptionCompound Discoverer Software
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesMass Frontier Software 7.0 SR3
Unit Size-
OPTON-30570Full specifications
- Compound Discoverer Software Upgrade from SIEVE (All Versions)Upgrade with Mass Frontier Software 7.0 SR3Request A Quote
TypeSoftware Upgrade
Software2
DescriptionCompound Discoverer Software Upgrade from SIEVE (All Versions)
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesUpgrade with Mass Frontier Software 7.0 SR3
Unit Size-
OPTON-30571Full specifications
- Compound Discoverer Software Upgrade from 1.0Upgrade OnlyRequest A Quote
TypeSoftware Upgrade
Software2
DescriptionCompound Discoverer Software Upgrade from 1.0
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesUpgrade Only
Unit Size-
Showing 10 of 19Show more
Know more unknowns
  • Identify compounds using automatic multiple database and spectral library search tools including mzCloud™, Chemspider™, KEGG, and BioCyc, and local database search tools such as the mzVault™ spectral library or mass lists.
  • Access the latest "auto-processed" mzCloud libraries and utilize both DIA and DDA scans for searching.
  • Determine elemental composition using high resolution fine isotopes, MS/MS, and MSn data.
  • Enables data analysis of high resolution, accurate mass Orbitrap mass spectrometers coupled to either liquid chromatography (LC) or gas chromatography (GC) instrumentation.
Workflow flexibility to match analytical requirements
Compound Discoverer software offers complete flexibility, making it easy to assemble a workflow that matches the analytical requirements of your analysis.
  • Workflows are modular and assembled from a suite of advanced algorithms (nodes).
  • Customizable workflow templates for different applications are included with the software for ease-of-use.
  • Workflows can be enhanced with custom nodes to meet the needs of your most challenging research.
Advanced data processing and visualization
  • Increase certainty using multi-factorial Peak Quality factors to reveal, identify, and quantify more compounds with confidence.
  • Biological pathway analysis with high quality data processing and statistical analysis—support for stable isotope labeling (“flux analysis”) workflows to analyze differences in samples.
  • Find real differences between sample sets with heat map-like tables, interactive PCA, partial least squares discriminant analysis (PLS-DA), etc.
  • Visualization of spectral trees along with fragmentation annotations and mzLogic algorithm for ranking chemical database hits using spectral library similarities for structural elucidation.
  • Molecular networks to explore relationships between compounds.