Informational:For order quantities > 1, multiple lots may be used to fulfill the order. If you require a single lot, please email discoverysciences@lifetech.com to ensure your needs are accommodated

88 more kinases now available to order! See the complete list.

PRKCQ (PKC theta) Recombinant Human Protein

Catalog number: P2996

PRKCQ (PKC theta) Recombinant Human Protein

Catalog number: P2996
Catalog Number
P2996
Unit Size
5 µg
Price (USD)
Price: 392.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
100 µg
Price (USD)
Price: 1,646.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
1 mg
Price (USD)
Price: 6,930.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
P29965 µg
Price: 392.00
Your Price:
-
PR4143B100 µg
Price: 1,646.00
Your Price:
-
PV36051 mg
Price: 6,930.00
Your Price:
-
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
The PKC family of serine⁄threonine kinases, including PRKCQ (PKC theta), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent.

Specifications

Accession Number
NP_006248
Gene ID (Entrez)
5584, 5588
Molecular Weight
84.4 kDa
Quantity
5μg
Gene Alias
PRKCQ (PKC theta)
KinaseFamily
PKC Kinase Family
Recombinant
Yes
Protein Tag
His Tag
Assay Entry
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Kinase Group
Ser⁄Thr Kinases: AGC Group
Shipping Condition
Dry Ice
System Compatibility
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate
Target Entry
PRKCQ
Target Gene
protein kinase C, theta
Expression System
Baculovirus
Protein Family
Kinases & Inhibitors
Protein Form
Full Length, Recombinant
Form
Liquid
Protein Subtype
Serine⁄Threonine Kinases
Validated Application
Kinase Assay
Species
Human
Research Category
Signal Transduction

Contents & Storage

Storage Buffer: 20mM HEPES, pH 7.5, 150mM NaCl, 0.5mM EGTA, 0.5mM EDTA, 0.05% NP-40, 5mM DTT, 20% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.