Informational:For order quantities > 1, multiple lots may be used to fulfill the order. If you require a single lot, please email discoverysciences@lifetech.com.

During checkout, this product will be reported as "Out of Stock" with an estimated availability date. Often, the actual availability date will be earlier. Send inquiries regarding actual availability date to discoverysciences@lifetech.com.

88 more kinases now available to order! See the complete list.
Thermo Scientific™

MST4 Recombinant Human Protein

Catalog number: PR7067A
Thermo Scientific™

MST4 Recombinant Human Protein

Catalog number: PR7067A
Catalog Number
PR7067A
Unit Size
100 µg
Price (USD)
Price: 1,646.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
1 mg
Price (USD)
Price: 7,010.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
10 µg
Price (USD)
Price: 480.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PR7067A100 µg
Price: 1,646.00
Your Price:
-
PV40241 mg
Price: 7,010.00
Your Price:
-
PV369010 µg
Price: 480.00
Your Price:
-
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
MST4 is member of the Ste20-like kinase family of serine⁄threonine kinases and is overexpressed during prostate cancer development.

Specifications

Accession Number
NP_057626.2
Molecular Weight
75.7 kDa
Quantity
100 µg
Gene Alias
MST4
KinaseFamily
STE20 Kinase Family
Recombinant
Yes
Protein Tag
GST Tag
Kinase Group
Ser⁄Thr Kinases: STE Group
Shipping Condition
Dry Ice
Expression System
Baculovirus
Protein Form
Full Length
Form
Frozen
Protein Subtype
Serine⁄Threonine Kinases
Validated Application
Kinase Assay
Species
Human
Research Category
Drug Development

Contents & Storage

Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.