Informational:For order quantities > 1, multiple lots may be used to fulfill the order. If you require a single lot, please email discoverysciences@lifetech.com.

During checkout, this product will be reported as "Out of Stock" with an estimated availability date. Often, the actual availability date will be earlier. Send inquiries regarding actual availability date to discoverysciences@lifetech.com.

88 more kinases now available to order! See the complete list.

RIPK2 Recombinant Human Protein

Catalog number: PR7829A

RIPK2 Recombinant Human Protein

Catalog number: PR7829A
Catalog Number
PR7829A
Unit Size
100 µg
Price (USD)
Price: 1,646.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
1 mg
Price (USD)
Price: 7,010.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
10 µg
Price (USD)
Price: 481.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PR7829A100 µg
Price: 1,646.00
Your Price:
-
PV43251 mg
Price: 7,010.00
Your Price:
-
PV421310 µg
Price: 481.00
Your Price:
-
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
RIPK2 is a serine⁄threonine kinase that is involved in cell activation, proliferation, differentiation, and apoptosis.

Specifications

Accession Number
NP_003812
Molecular Weight
38.9 kDa
Quantity
100μg
Gene Alias
RIPK2, RICK, RIP2
KinaseFamily
RIPK Kinase Family
Recombinant
Yes
Protein Tag
His Tag
Kinase Group
Ser⁄Thr Kinases: TKL Group
Shipping Condition
Dry Ice
Expression System
Baculovirus
Protein Form
Catalytic
Form
Frozen
Validated Application
Kinase Assay
Species
Human
Research Category
Drug Development

Contents & Storage

Storage Buffer: 50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% Glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.